Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247563502-247563938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCSAML
Entrez Gene ID
148823
Gene Name
germinal center associated signaling and motility like
Gene Aliases
C1orf150
UniGene
Hs.732113
Gene Chromosome Location
Chr.1: 247507058-247577690 on Build GRCh38
Transcripts
NM_001281834.1 NM_001281835.1 NM_001281836.1 NM_001281837.1 NM_001281838.1 NM_001281853.1 NM_145278.4 XM_017000399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977415 c.139+3A>G p.? Chr.1: 247563642-247563642 on Build GRCh38
319049 c.103A>C p.T35P Chr.1: 247563603-247563603 on Build GRCh38
3486686 c.94G>A p.E32K Chr.1: 247563594-247563594 on Build GRCh38
228846 c.121C>T p.Q41* Chr.1: 247563621-247563621 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773198074 Chr.1: 247563652-247563652 on Build GRCh38
rs1050194946 Chr.1: 247563853-247563853 on Build GRCh38
rs766014359 Chr.1: 247563543-247563543 on Build GRCh38
rs756560835 Chr.1: 247563559-247563559 on Build GRCh38
rs772900215 Chr.1: 247563542-247563542 on Build GRCh38
rs983528281 Chr.1: 247563586-247563586 on Build GRCh38
rs75349326 Chr.1: 247563816-247563816 on Build GRCh38
rs766637691 Chr.1: 247563893-247563893 on Build GRCh38
rs1044209943 Chr.1: 247563734-247563734 on Build GRCh38
rs1003401723 Chr.1: 247563583-247563583 on Build GRCh38
rs778227267 Chr.1: 247563563-247563563 on Build GRCh38
rs908699208 Chr.1: 247563762-247563762 on Build GRCh38
rs759190883 Chr.1: 247563680-247563680 on Build GRCh38
rs551859615 Chr.1: 247563697-247563697 on Build GRCh38
rs748561925 Chr.1: 247563794-247563794 on Build GRCh38
rs773882797 Chr.1: 247563675-247563675 on Build GRCh38
rs145479022 Chr.1: 247563634-247563634 on Build GRCh38
rs1054298612 Chr.1: 247563747-247563747 on Build GRCh38
rs1021392686 Chr.1: 247563821-247563821 on Build GRCh38
rs1009382782 Chr.1: 247563852-247563852 on Build GRCh38
rs375418538 Chr.1: 247563554-247563554 on Build GRCh38
rs916968384 Chr.1: 247563682-247563682 on Build GRCh38
rs567950767 Chr.1: 247563866-247563866 on Build GRCh38
rs143019925 Chr.1: 247563768-247563768 on Build GRCh38
rs753900029 Chr.1: 247563569-247563569 on Build GRCh38
rs769892254 Chr.1: 247563646-247563646 on Build GRCh38
rs928977529 Chr.1: 247563578-247563578 on Build GRCh38
rs560106218 Chr.1: 247563732-247563732 on Build GRCh38
rs779061895 Chr.1: 247563580-247563580 on Build GRCh38
rs768574277 Chr.1: 247563621-247563621 on Build GRCh38
rs372097410 Chr.1: 247563551-247563551 on Build GRCh38
rs140629777 Chr.1: 247563605-247563605 on Build GRCh38
rs777229119 Chr.1: 247563673-247563673 on Build GRCh38
rs946794468 Chr.1: 247563881-247563881 on Build GRCh38
rs769453108 Chr.1: 247563541-247563541 on Build GRCh38
rs890515905 Chr.1: 247563832-247563832 on Build GRCh38
rs7523951 Chr.1: 247563868-247563868 on Build GRCh38
rs527859045 Chr.1: 247563800-247563800 on Build GRCh38
rs533177067 Chr.1: 247563600-247563600 on Build GRCh38
rs935860949 Chr.1: 247563744-247563744 on Build GRCh38
rs983236084 Chr.1: 247563725-247563725 on Build GRCh38
rs868035820 Chr.1: 247563627-247563627 on Build GRCh38
rs964870073 Chr.1: 247563889-247563889 on Build GRCh38
rs748810788 Chr.1: 247563674-247563674 on Build GRCh38
rs951775840 Chr.1: 247563712-247563712 on Build GRCh38
rs757352296 Chr.1: 247563577-247563577 on Build GRCh38
rs753236750 Chr.1: 247563556-247563556 on Build GRCh38
rs905693634 Chr.1: 247563743-247563743 on Build GRCh38

View Full Product Details