Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 11869950-11870478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER2
Entrez Gene ID
254427
Gene Name
proline and serine rich 2
Gene Aliases
C10orf47
UniGene
Hs.435775
Gene Chromosome Location
Chr.10: 11823341-11872277 on Build GRCh38
Transcripts
NM_153256.3 XM_011519438.2 XM_017016028.1 NR_038222.1

Gene Symbol
PROSER2-AS1
Entrez Gene ID
219731
Gene Name
PROSER2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.576787
Gene Chromosome Location
Chr.10: 11849608-11894710 on Build GRCh38
Transcripts
NM_153256.3 XM_011519438.2 XM_017016028.1 NR_038222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6402537 c.1141C>A p.Q381K Chr.10: 11870239-11870239 on Build GRCh38
4388879 c.1179G>C p.Q393H Chr.10: 11870277-11870277 on Build GRCh38
4422282 c.1282C>G p.L428V Chr.10: 11870380-11870380 on Build GRCh38
6387599 c.1210C>A p.P404T Chr.10: 11870308-11870308 on Build GRCh38
6393607 c.1118C>A p.A373D Chr.10: 11870216-11870216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021056369 Chr.10: 11870249-11870249 on Build GRCh38
rs907876079 Chr.10: 11870310-11870310 on Build GRCh38
rs889337897 Chr.10: 11870299-11870299 on Build GRCh38
rs781037914 Chr.10: 11870442-11870442 on Build GRCh38
rs1001800199 Chr.10: 11869970-11869970 on Build GRCh38
rs200622168 Chr.10: 11870182-11870182 on Build GRCh38
rs775645388 Chr.10: 11870236-11870236 on Build GRCh38
rs972142168 Chr.10: 11870193-11870193 on Build GRCh38
rs565718892 Chr.10: 11870358-11870358 on Build GRCh38
rs879092693 Chr.10: 11870269-11870269 on Build GRCh38
rs1037947381 Chr.10: 11870439-11870439 on Build GRCh38
rs372530346 Chr.10: 11870252-11870252 on Build GRCh38
rs770720122 Chr.10: 11870228-11870228 on Build GRCh38
rs1037838529 Chr.10: 11870202-11870202 on Build GRCh38
rs11257369 Chr.10: 11870320-11870320 on Build GRCh38
rs983806386 Chr.10: 11870456-11870456 on Build GRCh38
rs750874391 Chr.10: 11870392-11870393 on Build GRCh38
rs182610629 Chr.10: 11870453-11870453 on Build GRCh38
rs892890108 Chr.10: 11869983-11869983 on Build GRCh38
rs753039020 Chr.10: 11870396-11870396 on Build GRCh38
rs541809225 Chr.10: 11870001-11870001 on Build GRCh38
rs143145956 Chr.10: 11870337-11870337 on Build GRCh38
rs766758903 Chr.10: 11870357-11870357 on Build GRCh38
rs774712871 Chr.10: 11870411-11870411 on Build GRCh38
rs764378465 Chr.10: 11870393-11870393 on Build GRCh38
rs959435648 Chr.10: 11870362-11870362 on Build GRCh38
rs758487000 Chr.10: 11870412-11870412 on Build GRCh38
rs1055599616 Chr.10: 11870259-11870259 on Build GRCh38
rs949406657 Chr.10: 11870064-11870064 on Build GRCh38
rs767757734 Chr.10: 11870126-11870126 on Build GRCh38
rs942094074 Chr.10: 11870026-11870026 on Build GRCh38
rs559537224 Chr.10: 11870180-11870180 on Build GRCh38
rs534844986 Chr.10: 11870372-11870372 on Build GRCh38
rs991795634 Chr.10: 11870365-11870365 on Build GRCh38
rs986715372 Chr.10: 11870016-11870016 on Build GRCh38
rs772055425 Chr.10: 11870207-11870207 on Build GRCh38
rs767929082 Chr.10: 11870328-11870328 on Build GRCh38
rs1051891414 Chr.10: 11870354-11870354 on Build GRCh38
rs1008216195 Chr.10: 11870307-11870307 on Build GRCh38
rs541938628 Chr.10: 11870230-11870230 on Build GRCh38
rs17857360 Chr.10: 11870066-11870066 on Build GRCh38
rs780196455 Chr.10: 11870152-11870152 on Build GRCh38
rs959124016 Chr.10: 11870416-11870416 on Build GRCh38
rs1051554933 Chr.10: 11870424-11870424 on Build GRCh38
rs966446053 Chr.10: 11870251-11870251 on Build GRCh38
rs1038230029 Chr.10: 11870332-11870332 on Build GRCh38
rs1016567124 Chr.10: 11870314-11870314 on Build GRCh38
rs972022903 Chr.10: 11870036-11870036 on Build GRCh38
rs1024817136 Chr.10: 11870441-11870441 on Build GRCh38
rs1041551128 Chr.10: 11870402-11870402 on Build GRCh38
rs990725879 Chr.10: 11870276-11870276 on Build GRCh38
rs770912396 Chr.10: 11870203-11870203 on Build GRCh38
rs12253600 Chr.10: 11870432-11870432 on Build GRCh38
rs116295162 Chr.10: 11870298-11870298 on Build GRCh38
rs947245895 Chr.10: 11870272-11870272 on Build GRCh38
rs919253966 Chr.10: 11870062-11870062 on Build GRCh38
rs756510918 Chr.10: 11870150-11870150 on Build GRCh38
rs746976664 Chr.10: 11870223-11870223 on Build GRCh38
rs752312597 Chr.10: 11870114-11870114 on Build GRCh38
rs866081852 Chr.10: 11870256-11870256 on Build GRCh38
rs953773844 Chr.10: 11870153-11870153 on Build GRCh38
rs1011174772 Chr.10: 11869984-11869984 on Build GRCh38
rs12253554 Chr.10: 11870333-11870333 on Build GRCh38
rs745789964 Chr.10: 11870445-11870445 on Build GRCh38
rs748361539 Chr.10: 11870196-11870196 on Build GRCh38
rs914402996 Chr.10: 11870266-11870266 on Build GRCh38
rs983412094 Chr.10: 11870301-11870301 on Build GRCh38
rs997166361 Chr.10: 11870149-11870149 on Build GRCh38
rs772881601 Chr.10: 11870220-11870220 on Build GRCh38
rs775942718 Chr.10: 11869987-11869987 on Build GRCh38
rs1011290795 Chr.10: 11870107-11870107 on Build GRCh38
rs191542752 Chr.10: 11870382-11870382 on Build GRCh38
rs909272880 Chr.10: 11870024-11870024 on Build GRCh38
rs1041435507 Chr.10: 11870275-11870275 on Build GRCh38
rs942112438 Chr.10: 11870190-11870190 on Build GRCh38
rs574328858 Chr.10: 11870037-11870037 on Build GRCh38
rs74657303 Chr.10: 11870229-11870229 on Build GRCh38
rs890776314 Chr.10: 11870235-11870235 on Build GRCh38
rs569917088 Chr.10: 11870304-11870304 on Build GRCh38
rs963523357 Chr.10: 11870322-11870322 on Build GRCh38
rs375509412 Chr.10: 11870115-11870115 on Build GRCh38
rs1007685232 Chr.10: 11870157-11870157 on Build GRCh38
rs746023256 Chr.10: 11870214-11870214 on Build GRCh38
rs17851505 Chr.10: 11870145-11870145 on Build GRCh38
rs568187783 Chr.10: 11870426-11870426 on Build GRCh38
rs749278816 Chr.10: 11870162-11870162 on Build GRCh38
rs777642908 Chr.10: 11870419-11870419 on Build GRCh38
rs902217744 Chr.10: 11870388-11870388 on Build GRCh38
rs577089253 Chr.10: 11870454-11870454 on Build GRCh38
rs201537600 Chr.10: 11870278-11870278 on Build GRCh38
rs1032175966 Chr.10: 11870254-11870254 on Build GRCh38
rs577124442 Chr.10: 11870132-11870132 on Build GRCh38
rs1056053177 Chr.10: 11870078-11870078 on Build GRCh38
rs767899335 Chr.10: 11870214-11870215 on Build GRCh38
rs919365541 Chr.10: 11870208-11870209 on Build GRCh38
rs953559219 Chr.10: 11870002-11870002 on Build GRCh38
rs937162484 Chr.10: 11870076-11870076 on Build GRCh38
rs888775436 Chr.10: 11870429-11870429 on Build GRCh38
rs914518606 Chr.10: 11870381-11870381 on Build GRCh38
rs371990658 Chr.10: 11870168-11870168 on Build GRCh38
rs762446484 Chr.10: 11870094-11870107 on Build GRCh38
rs902953841 Chr.10: 11870277-11870282 on Build GRCh38
rs976300593 Chr.10: 11869998-11869998 on Build GRCh38
rs1030563248 Chr.10: 11870231-11870231 on Build GRCh38
rs967049644 Chr.10: 11869988-11869988 on Build GRCh38
rs892850130 Chr.10: 11870099-11870099 on Build GRCh38
rs751558508 Chr.10: 11869973-11869999 on Build GRCh38

View Full Product Details