Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13496688-13497200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND7
Entrez Gene ID
222389
Gene Name
BEN domain containing 7
Gene Aliases
C10orf30
UniGene
Hs.498740
Gene Chromosome Location
Chr.10: 13438484-13528772 on Build GRCh38
Transcripts
NM_001100912.1 NM_152751.2 XM_005252402.4 XM_011519390.2 XM_011519391.2 XM_011519392.2 XM_011519393.2 XM_011519394.2 XM_011519395.2 XM_011519396.2 XM_011519397.2 XM_011519398.2 XM_011519399.2 XM_011519400.2 XM_011519401.2 XM_011519402.2 XM_011519403.2 XM_011519404.2 XM_011519405.2 XM_011519406.2 XM_011519407.1 XM_011519408.2 XM_011519410.2 XM_011519411.2 XM_011519412.1 XM_011519413.1 XM_011519414.1 XM_017015915.1 XM_017015916.1 XM_017015917.1 XM_017015918.1 XM_017015919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5050050 c.387C>A p.C129* Chr.10: 13496833-13496833 on Build GRCh38
4409810 c.400A>C p.M134L Chr.10: 13496781-13496781 on Build GRCh38
6611304 c.387C>T p.C129C Chr.10: 13496833-13496833 on Build GRCh38
4819206 c.370C>G p.L124V Chr.10: 13496811-13496811 on Build GRCh38
7258133 c.569C>T p.A190V Chr.10: 13496768-13496768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778593853 Chr.10: 13496843-13496843 on Build GRCh38
rs533512054 Chr.10: 13496956-13496956 on Build GRCh38
rs1052227551 Chr.10: 13496941-13496942 on Build GRCh38
rs569476559 Chr.10: 13497119-13497119 on Build GRCh38
rs754764345 Chr.10: 13496828-13496828 on Build GRCh38
rs765602890 Chr.10: 13496904-13496906 on Build GRCh38
rs773472970 Chr.10: 13496919-13496919 on Build GRCh38
rs538415309 Chr.10: 13497132-13497132 on Build GRCh38
rs79146913 Chr.10: 13496969-13496969 on Build GRCh38
rs1048020101 Chr.10: 13497145-13497145 on Build GRCh38
rs777635118 Chr.10: 13496862-13496862 on Build GRCh38
rs780036951 Chr.10: 13496733-13496733 on Build GRCh38
rs976517796 Chr.10: 13496713-13496713 on Build GRCh38
rs568550072 Chr.10: 13496824-13496824 on Build GRCh38
rs990127859 Chr.10: 13497112-13497112 on Build GRCh38
rs936589499 Chr.10: 13496891-13496891 on Build GRCh38
rs564556041 Chr.10: 13496950-13496950 on Build GRCh38
rs917941270 Chr.10: 13496814-13496814 on Build GRCh38
rs1048059573 Chr.10: 13497066-13497066 on Build GRCh38
rs1041216497 Chr.10: 13496959-13496959 on Build GRCh38
rs766478507 Chr.10: 13496937-13496937 on Build GRCh38
rs553462328 Chr.10: 13497159-13497159 on Build GRCh38
rs551113663 Chr.10: 13497135-13497135 on Build GRCh38
rs889561102 Chr.10: 13497154-13497154 on Build GRCh38
rs746501385 Chr.10: 13496876-13496876 on Build GRCh38
rs573267971 Chr.10: 13497169-13497169 on Build GRCh38
rs1019429860 Chr.10: 13497038-13497038 on Build GRCh38
rs907480130 Chr.10: 13496858-13496858 on Build GRCh38
rs143961403 Chr.10: 13496742-13496742 on Build GRCh38
rs987995311 Chr.10: 13497139-13497140 on Build GRCh38
rs376636504 Chr.10: 13496928-13496929 on Build GRCh38
rs774586682 Chr.10: 13496822-13496822 on Build GRCh38
rs771560506 Chr.10: 13496750-13496750 on Build GRCh38
rs763351028 Chr.10: 13496787-13496787 on Build GRCh38
rs1037294951 Chr.10: 13497137-13497137 on Build GRCh38
rs879048922 Chr.10: 13496930-13496930 on Build GRCh38
rs776950071 Chr.10: 13496791-13496791 on Build GRCh38
rs924614836 Chr.10: 13496724-13496724 on Build GRCh38
rs780673593 Chr.10: 13496901-13496901 on Build GRCh38
rs71503087 Chr.10: 13496929-13496930 on Build GRCh38
rs1038743411 Chr.10: 13496957-13496957 on Build GRCh38
rs1015111812 Chr.10: 13497109-13497109 on Build GRCh38
rs5783327 Chr.10: 13496945-13496945 on Build GRCh38
rs369417947 Chr.10: 13496747-13496747 on Build GRCh38
rs750500731 Chr.10: 13496825-13496825 on Build GRCh38
rs74902034 Chr.10: 13496946-13496946 on Build GRCh38
rs746309655 Chr.10: 13496758-13496758 on Build GRCh38
rs776193350 Chr.10: 13496785-13496785 on Build GRCh38
rs769010519 Chr.10: 13496906-13496906 on Build GRCh38
rs995101532 Chr.10: 13496963-13496963 on Build GRCh38
rs767128380 Chr.10: 13497138-13497138 on Build GRCh38
rs373057266 Chr.10: 13496904-13496904 on Build GRCh38
rs577318059 Chr.10: 13497142-13497142 on Build GRCh38
rs913803398 Chr.10: 13497140-13497141 on Build GRCh38
rs547118089 Chr.10: 13496998-13496998 on Build GRCh38
rs200443723 Chr.10: 13496851-13496851 on Build GRCh38
rs896818139 Chr.10: 13496954-13496954 on Build GRCh38
rs753617899 Chr.10: 13496740-13496740 on Build GRCh38
rs761956779 Chr.10: 13496823-13496823 on Build GRCh38
rs200063820 Chr.10: 13496927-13496927 on Build GRCh38
rs201032751 Chr.10: 13496826-13496826 on Build GRCh38
rs768939349 Chr.10: 13496795-13496795 on Build GRCh38
rs145969876 Chr.10: 13497134-13497134 on Build GRCh38
rs965697477 Chr.10: 13497065-13497065 on Build GRCh38
rs549622001 Chr.10: 13497106-13497106 on Build GRCh38
rs992621834 Chr.10: 13496752-13496752 on Build GRCh38
rs760715122 Chr.10: 13496923-13496923 on Build GRCh38
rs765661345 Chr.10: 13497120-13497120 on Build GRCh38
rs750305082 Chr.10: 13496722-13496722 on Build GRCh38
rs183702190 Chr.10: 13496748-13496748 on Build GRCh38
rs566830472 Chr.10: 13497004-13497004 on Build GRCh38
rs376961432 Chr.10: 13496896-13496896 on Build GRCh38
rs376343391 Chr.10: 13496759-13496759 on Build GRCh38
rs925236989 Chr.10: 13496834-13496834 on Build GRCh38
rs748695280 Chr.10: 13497053-13497053 on Build GRCh38
rs888034976 Chr.10: 13497071-13497071 on Build GRCh38
rs777433434 Chr.10: 13496753-13496753 on Build GRCh38
rs953771844 Chr.10: 13496995-13496995 on Build GRCh38
rs371244608 Chr.10: 13496927-13496928 on Build GRCh38
rs774586896 Chr.10: 13496913-13496913 on Build GRCh38
rs796484540 Chr.10: 13496927-13496928 on Build GRCh38
rs756053856 Chr.10: 13496726-13496726 on Build GRCh38
rs948772258 Chr.10: 13497122-13497122 on Build GRCh38
rs111810942 Chr.10: 13496792-13496792 on Build GRCh38
rs767609116 Chr.10: 13496721-13496721 on Build GRCh38
rs759953358 Chr.10: 13497076-13497076 on Build GRCh38
rs376304759 Chr.10: 13496928-13496928 on Build GRCh38
rs1037145101 Chr.10: 13496947-13496947 on Build GRCh38
rs969491151 Chr.10: 13497127-13497127 on Build GRCh38
rs766281842 Chr.10: 13496827-13496827 on Build GRCh38
rs748615774 Chr.10: 13496917-13496917 on Build GRCh38
rs796919689 Chr.10: 13496929-13496930 on Build GRCh38
rs59561773 Chr.10: 13496945-13496946 on Build GRCh38
rs533598327 Chr.10: 13497156-13497156 on Build GRCh38
rs535865055 Chr.10: 13497007-13497007 on Build GRCh38
rs1044984318 Chr.10: 13497126-13497126 on Build GRCh38
rs948886169 Chr.10: 13496788-13496788 on Build GRCh38
rs1026694699 Chr.10: 13496979-13496979 on Build GRCh38
rs956031652 Chr.10: 13497110-13497110 on Build GRCh38

View Full Product Details