Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16520703-16521203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL3
Entrez Gene ID
389941
Gene Name
complement C1q like 3
Gene Aliases
C1QTNF13, C1ql, CTRP13, K100
UniGene
Hs.676792
Gene Chromosome Location
Chr.10: 16513743-16522005 on Build GRCh38
Transcripts
NM_001010908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198506 c.163C>A p.L55M Chr.10: 16520903-16520903 on Build GRCh38
4956294 c.86G>A p.R29H Chr.10: 16520980-16520980 on Build GRCh38
4667222 c.166C>T p.P56S Chr.10: 16520900-16520900 on Build GRCh38
1128088 c.124A>T p.S42C Chr.10: 16520942-16520942 on Build GRCh38
7247341 c.348C>T p.I116I Chr.10: 16520718-16520718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749672191 Chr.10: 16520898-16520898 on Build GRCh38
rs1040828252 Chr.10: 16521074-16521074 on Build GRCh38
rs961268494 Chr.10: 16520721-16520721 on Build GRCh38
rs752202297 Chr.10: 16520779-16520779 on Build GRCh38
rs748214615 Chr.10: 16520917-16520917 on Build GRCh38
rs762617699 Chr.10: 16520966-16520966 on Build GRCh38
rs372732009 Chr.10: 16521027-16521027 on Build GRCh38
rs1007839791 Chr.10: 16521036-16521036 on Build GRCh38
rs559546423 Chr.10: 16521128-16521128 on Build GRCh38
rs770900487 Chr.10: 16521013-16521013 on Build GRCh38
rs752924544 Chr.10: 16521003-16521003 on Build GRCh38
rs773247485 Chr.10: 16520750-16520750 on Build GRCh38
rs541527618 Chr.10: 16521021-16521021 on Build GRCh38
rs926228779 Chr.10: 16520946-16520946 on Build GRCh38
rs867958674 Chr.10: 16520926-16520926 on Build GRCh38
rs755247100 Chr.10: 16520909-16520909 on Build GRCh38
rs763620914 Chr.10: 16520967-16520967 on Build GRCh38
rs776478174 Chr.10: 16520928-16520928 on Build GRCh38
rs770791299 Chr.10: 16520925-16520925 on Build GRCh38
rs759396446 Chr.10: 16520937-16520937 on Build GRCh38
rs972964643 Chr.10: 16520748-16520748 on Build GRCh38
rs561906962 Chr.10: 16520773-16520773 on Build GRCh38
rs761269820 Chr.10: 16521079-16521079 on Build GRCh38
rs769013505 Chr.10: 16520727-16520727 on Build GRCh38
rs1040513288 Chr.10: 16521170-16521170 on Build GRCh38
rs992967712 Chr.10: 16520895-16520895 on Build GRCh38
rs773907762 Chr.10: 16521066-16521066 on Build GRCh38
rs748304206 Chr.10: 16520741-16520741 on Build GRCh38
rs767991784 Chr.10: 16520855-16520855 on Build GRCh38
rs889414432 Chr.10: 16521142-16521142 on Build GRCh38
rs938752601 Chr.10: 16520820-16520820 on Build GRCh38
rs773193390 Chr.10: 16520920-16520920 on Build GRCh38
rs757946024 Chr.10: 16520814-16520814 on Build GRCh38
rs777847445 Chr.10: 16521005-16521005 on Build GRCh38
rs994033893 Chr.10: 16521117-16521117 on Build GRCh38
rs775174561 Chr.10: 16520959-16520959 on Build GRCh38
rs376506829 Chr.10: 16521024-16521024 on Build GRCh38
rs753122423 Chr.10: 16521106-16521106 on Build GRCh38
rs772227170 Chr.10: 16520919-16520919 on Build GRCh38
rs760549017 Chr.10: 16520755-16520755 on Build GRCh38
rs759135086 Chr.10: 16520767-16520767 on Build GRCh38
rs777928946 Chr.10: 16521113-16521113 on Build GRCh38
rs1008229399 Chr.10: 16521155-16521155 on Build GRCh38
rs613731 Chr.10: 16520999-16520999 on Build GRCh38
rs888388929 Chr.10: 16520788-16520788 on Build GRCh38
rs1036466379 Chr.10: 16521006-16521006 on Build GRCh38
rs766154179 Chr.10: 16520760-16520760 on Build GRCh38
rs760147940 Chr.10: 16521095-16521095 on Build GRCh38
rs746894408 Chr.10: 16520924-16520924 on Build GRCh38
rs538734279 Chr.10: 16520936-16520936 on Build GRCh38
rs930691294 Chr.10: 16521046-16521046 on Build GRCh38
rs764842265 Chr.10: 16520948-16520948 on Build GRCh38
rs768207857 Chr.10: 16521060-16521060 on Build GRCh38
rs756611028 Chr.10: 16520876-16520876 on Build GRCh38
rs761230566 Chr.10: 16520985-16520985 on Build GRCh38
rs765629093 Chr.10: 16521102-16521102 on Build GRCh38
rs529048732 Chr.10: 16520997-16520997 on Build GRCh38
rs754346866 Chr.10: 16521094-16521094 on Build GRCh38
rs754222263 Chr.10: 16520989-16520989 on Build GRCh38
rs758693983 Chr.10: 16521004-16521004 on Build GRCh38
rs1027205144 Chr.10: 16521126-16521126 on Build GRCh38
rs762526467 Chr.10: 16521038-16521038 on Build GRCh38
rs780400568 Chr.10: 16520894-16520894 on Build GRCh38
rs747120177 Chr.10: 16521012-16521012 on Build GRCh38
rs779316337 Chr.10: 16520916-16520916 on Build GRCh38
rs763814215 Chr.10: 16520857-16520857 on Build GRCh38
rs980112357 Chr.10: 16520778-16520778 on Build GRCh38
rs776290777 Chr.10: 16520766-16520766 on Build GRCh38
rs766862356 Chr.10: 16520987-16520987 on Build GRCh38
rs750895215 Chr.10: 16520863-16520863 on Build GRCh38
rs751037130 Chr.10: 16520982-16520982 on Build GRCh38
rs758637214 Chr.10: 16521112-16521112 on Build GRCh38
rs772102503 Chr.10: 16520746-16520746 on Build GRCh38
rs913301413 Chr.10: 16520912-16520912 on Build GRCh38
rs774558933 Chr.10: 16520740-16520740 on Build GRCh38
rs766886131 Chr.10: 16521084-16521084 on Build GRCh38

View Full Product Details