Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18675398-18675902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5B
Entrez Gene ID
221079
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
UniGene
Hs.25362
Gene Chromosome Location
Chr.10: 18659335-18681639 on Build GRCh38
Transcripts
NM_178815.3 XM_005252400.1 XM_005252401.4

SNP Detail

SNP ID Chromosome Location
rs41306326 Chr.10: 18675720-18675720 on Build GRCh38
rs557548208 Chr.10: 18675527-18675527 on Build GRCh38
rs1031431674 Chr.10: 18675750-18675750 on Build GRCh38
rs1012068736 Chr.10: 18675639-18675639 on Build GRCh38
rs778075547 Chr.10: 18675567-18675567 on Build GRCh38
rs1004725315 Chr.10: 18675472-18675472 on Build GRCh38
rs968055089 Chr.10: 18675470-18675470 on Build GRCh38
rs1031547090 Chr.10: 18675512-18675512 on Build GRCh38
rs549912994 Chr.10: 18675780-18675780 on Build GRCh38
rs781163161 Chr.10: 18675661-18675661 on Build GRCh38
rs997508124 Chr.10: 18675590-18675590 on Build GRCh38
rs1016469740 Chr.10: 18675474-18675477 on Build GRCh38
rs144065833 Chr.10: 18675623-18675623 on Build GRCh38
rs565219193 Chr.10: 18675742-18675742 on Build GRCh38
rs766796661 Chr.10: 18675585-18675585 on Build GRCh38
rs920304692 Chr.10: 18675848-18675848 on Build GRCh38
rs180984404 Chr.10: 18675874-18675874 on Build GRCh38
rs1016111248 Chr.10: 18675622-18675622 on Build GRCh38
rs147084208 Chr.10: 18675570-18675570 on Build GRCh38
rs757313104 Chr.10: 18675628-18675628 on Build GRCh38
rs556684317 Chr.10: 18675807-18675807 on Build GRCh38
rs776840031 Chr.10: 18675444-18675444 on Build GRCh38
rs1056622800 Chr.10: 18675629-18675629 on Build GRCh38
rs908097282 Chr.10: 18675619-18675619 on Build GRCh38
rs748306549 Chr.10: 18675532-18675532 on Build GRCh38
rs184135456 Chr.10: 18675566-18675566 on Build GRCh38
rs886525974 Chr.10: 18675617-18675617 on Build GRCh38
rs570744792 Chr.10: 18675511-18675511 on Build GRCh38
rs963342514 Chr.10: 18675497-18675497 on Build GRCh38
rs964575608 Chr.10: 18675818-18675818 on Build GRCh38
rs999457775 Chr.10: 18675748-18675748 on Build GRCh38
rs886407530 Chr.10: 18675878-18675878 on Build GRCh38
rs564413124 Chr.10: 18675735-18675735 on Build GRCh38
rs576234767 Chr.10: 18675536-18675536 on Build GRCh38
rs531796051 Chr.10: 18675777-18675777 on Build GRCh38
rs974583284 Chr.10: 18675823-18675823 on Build GRCh38
rs895342819 Chr.10: 18675633-18675633 on Build GRCh38
rs981777069 Chr.10: 18675670-18675670 on Build GRCh38
rs923722871 Chr.10: 18675528-18675528 on Build GRCh38
rs576356491 Chr.10: 18675540-18675540 on Build GRCh38
rs1018779718 Chr.10: 18675809-18675809 on Build GRCh38
rs947597104 Chr.10: 18675767-18675767 on Build GRCh38
rs749328983 Chr.10: 18675543-18675543 on Build GRCh38
rs989666068 Chr.10: 18675697-18675697 on Build GRCh38
rs964437383 Chr.10: 18675549-18675549 on Build GRCh38
rs1043755892 Chr.10: 18675731-18675731 on Build GRCh38
rs1051679899 Chr.10: 18675605-18675605 on Build GRCh38
rs746002775 Chr.10: 18675693-18675693 on Build GRCh38
rs188486034 Chr.10: 18675680-18675680 on Build GRCh38
rs914981243 Chr.10: 18675749-18675749 on Build GRCh38
rs947568503 Chr.10: 18675564-18675564 on Build GRCh38
rs552685326 Chr.10: 18675694-18675694 on Build GRCh38
rs1012314761 Chr.10: 18675438-18675438 on Build GRCh38
rs568434246 Chr.10: 18675788-18675788 on Build GRCh38
rs955967275 Chr.10: 18675762-18675762 on Build GRCh38
rs773925032 Chr.10: 18675650-18675650 on Build GRCh38
rs192086217 Chr.10: 18675545-18675545 on Build GRCh38
rs535717272 Chr.10: 18675868-18675868 on Build GRCh38
rs368759340 Chr.10: 18675551-18675551 on Build GRCh38
rs923580135 Chr.10: 18675677-18675677 on Build GRCh38
rs1047431893 Chr.10: 18675613-18675613 on Build GRCh38
rs554907790 Chr.10: 18675515-18675515 on Build GRCh38
rs138497131 Chr.10: 18675583-18675583 on Build GRCh38
rs970600458 Chr.10: 18675669-18675669 on Build GRCh38
rs1023594698 Chr.10: 18675658-18675658 on Build GRCh38
rs560568630 Chr.10: 18675594-18675594 on Build GRCh38
rs1030414246 Chr.10: 18675428-18675428 on Build GRCh38
rs1039171032 Chr.10: 18675778-18675778 on Build GRCh38
rs933533509 Chr.10: 18675789-18675789 on Build GRCh38

View Full Product Details