Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35248156-35248701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNY
Entrez Gene ID
219771
Gene Name
cyclin Y
Gene Aliases
C10orf9, CBCP1, CCNX, CFP1
UniGene
Hs.14745
Gene Chromosome Location
Chr.10: 35246981-35572670 on Build GRCh38
Transcripts
NM_181698.3 XM_011519358.1 XM_011519360.2 XM_011519361.1 XM_011519362.1 XM_011519363.1 XM_011519364.1 XM_011519365.2 XM_011519366.1

SNP Detail

SNP ID Chromosome Location
rs899155380 Chr.10: 35248268-35248268 on Build GRCh38
rs761798045 Chr.10: 35248371-35248371 on Build GRCh38
rs555099187 Chr.10: 35248464-35248464 on Build GRCh38
rs1051949822 Chr.10: 35248419-35248419 on Build GRCh38
rs969618169 Chr.10: 35248614-35248614 on Build GRCh38
rs1053719572 Chr.10: 35248657-35248657 on Build GRCh38
rs113863235 Chr.10: 35248372-35248372 on Build GRCh38
rs779538259 Chr.10: 35248400-35248400 on Build GRCh38
rs1051463372 Chr.10: 35248358-35248358 on Build GRCh38
rs542305610 Chr.10: 35248609-35248609 on Build GRCh38
rs111277493 Chr.10: 35248214-35248214 on Build GRCh38
rs943669175 Chr.10: 35248483-35248483 on Build GRCh38
rs1038359646 Chr.10: 35248489-35248490 on Build GRCh38
rs895923948 Chr.10: 35248556-35248556 on Build GRCh38
rs1024741749 Chr.10: 35248362-35248362 on Build GRCh38
rs533550336 Chr.10: 35248520-35248520 on Build GRCh38
rs991715014 Chr.10: 35248496-35248496 on Build GRCh38
rs371826901 Chr.10: 35248500-35248500 on Build GRCh38
rs760464686 Chr.10: 35248561-35248561 on Build GRCh38
rs932969764 Chr.10: 35248351-35248351 on Build GRCh38
rs963863075 Chr.10: 35248518-35248518 on Build GRCh38
rs539371058 Chr.10: 35248369-35248369 on Build GRCh38
rs780323353 Chr.10: 35248366-35248366 on Build GRCh38
rs190442231 Chr.10: 35248381-35248381 on Build GRCh38
rs909359646 Chr.10: 35248509-35248509 on Build GRCh38
rs958914697 Chr.10: 35248465-35248465 on Build GRCh38
rs987912776 Chr.10: 35248390-35248390 on Build GRCh38
rs1035693073 Chr.10: 35248439-35248439 on Build GRCh38
rs58119251 Chr.10: 35248370-35248370 on Build GRCh38
rs573663087 Chr.10: 35248543-35248543 on Build GRCh38
rs774364243 Chr.10: 35248294-35248294 on Build GRCh38
rs1052606102 Chr.10: 35248194-35248194 on Build GRCh38
rs899190672 Chr.10: 35248512-35248512 on Build GRCh38
rs112605534 Chr.10: 35248188-35248188 on Build GRCh38
rs1002820959 Chr.10: 35248410-35248410 on Build GRCh38
rs910683396 Chr.10: 35248480-35248480 on Build GRCh38
rs551790049 Chr.10: 35248199-35248199 on Build GRCh38
rs775955018 Chr.10: 35248575-35248575 on Build GRCh38
rs972288750 Chr.10: 35248541-35248541 on Build GRCh38
rs1008199781 Chr.10: 35248229-35248229 on Build GRCh38
rs1002165723 Chr.10: 35248672-35248672 on Build GRCh38
rs933098693 Chr.10: 35248552-35248552 on Build GRCh38
rs772663962 Chr.10: 35248570-35248570 on Build GRCh38
rs556638270 Chr.10: 35248328-35248328 on Build GRCh38
rs769197859 Chr.10: 35248379-35248379 on Build GRCh38
rs991615132 Chr.10: 35248225-35248225 on Build GRCh38
rs1014254718 Chr.10: 35248580-35248580 on Build GRCh38
rs889036348 Chr.10: 35248365-35248365 on Build GRCh38
rs370488336 Chr.10: 35248657-35248658 on Build GRCh38
rs187253962 Chr.10: 35248249-35248249 on Build GRCh38
rs1052043712 Chr.10: 35248224-35248224 on Build GRCh38
rs981430980 Chr.10: 35248189-35248189 on Build GRCh38
rs570064009 Chr.10: 35248341-35248341 on Build GRCh38
rs941760771 Chr.10: 35248270-35248270 on Build GRCh38
rs571108318 Chr.10: 35248505-35248505 on Build GRCh38
rs933026632 Chr.10: 35248404-35248404 on Build GRCh38
rs928877388 Chr.10: 35248205-35248205 on Build GRCh38

View Full Product Details