Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68747185-68747710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR1
Entrez Gene ID
55749
Gene Name
cell division cycle and apoptosis regulator 1
Gene Aliases
-
UniGene
Hs.49853
Gene Chromosome Location
Chr.10: 68721144-68792377 on Build GRCh38
Transcripts
NM_001282959.1 NM_001282960.1 NM_018237.3 NR_104262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1474720 c.637C>T p.Q213* Chr.10: 68747377-68747377 on Build GRCh38
919796 c.817C>T p.Q273* Chr.10: 68747557-68747557 on Build GRCh38
6287712 c.747G>T p.Q249H Chr.10: 68747487-68747487 on Build GRCh38
427797 c.813G>T p.Q271H Chr.10: 68747553-68747553 on Build GRCh38
5613266 c.646A>G p.T216A Chr.10: 68747386-68747386 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755484882 Chr.10: 68747592-68747592 on Build GRCh38
rs888239393 Chr.10: 68747370-68747372 on Build GRCh38
rs947619247 Chr.10: 68747571-68747571 on Build GRCh38
rs879372080 Chr.10: 68747268-68747268 on Build GRCh38
rs374007238 Chr.10: 68747214-68747214 on Build GRCh38
rs771299236 Chr.10: 68747416-68747416 on Build GRCh38
rs756942213 Chr.10: 68747377-68747377 on Build GRCh38
rs1041956649 Chr.10: 68747576-68747576 on Build GRCh38
rs773431432 Chr.10: 68747285-68747285 on Build GRCh38
rs780817748 Chr.10: 68747521-68747521 on Build GRCh38
rs754607534 Chr.10: 68747517-68747517 on Build GRCh38
rs778091068 Chr.10: 68747569-68747569 on Build GRCh38
rs770194485 Chr.10: 68747394-68747394 on Build GRCh38
rs534110119 Chr.10: 68747335-68747335 on Build GRCh38
rs781562064 Chr.10: 68747391-68747391 on Build GRCh38
rs373840226 Chr.10: 68747505-68747505 on Build GRCh38
rs1055415634 Chr.10: 68747407-68747407 on Build GRCh38
rs765066868 Chr.10: 68747477-68747477 on Build GRCh38
rs377035961 Chr.10: 68747596-68747596 on Build GRCh38
rs1015223397 Chr.10: 68747350-68747350 on Build GRCh38
rs771364994 Chr.10: 68747585-68747585 on Build GRCh38
rs766676573 Chr.10: 68747304-68747304 on Build GRCh38
rs746555945 Chr.10: 68747375-68747375 on Build GRCh38
rs748463374 Chr.10: 68747264-68747264 on Build GRCh38
rs1025182703 Chr.10: 68747529-68747529 on Build GRCh38
rs938657858 Chr.10: 68747605-68747605 on Build GRCh38
rs777153318 Chr.10: 68747226-68747226 on Build GRCh38
rs374047363 Chr.10: 68747333-68747333 on Build GRCh38
rs754415028 Chr.10: 68747386-68747386 on Build GRCh38
rs760262668 Chr.10: 68747338-68747338 on Build GRCh38
rs779408655 Chr.10: 68747594-68747594 on Build GRCh38
rs774673583 Chr.10: 68747427-68747427 on Build GRCh38
rs5030885 Chr.10: 68747218-68747218 on Build GRCh38
rs749268976 Chr.10: 68747327-68747327 on Build GRCh38
rs139960975 Chr.10: 68747672-68747672 on Build GRCh38
rs763717234 Chr.10: 68747349-68747351 on Build GRCh38
rs775427935 Chr.10: 68747443-68747443 on Build GRCh38
rs751147625 Chr.10: 68747359-68747360 on Build GRCh38
rs751152525 Chr.10: 68747508-68747508 on Build GRCh38
rs536354817 Chr.10: 68747614-68747614 on Build GRCh38
rs551781208 Chr.10: 68747278-68747278 on Build GRCh38
rs750259008 Chr.10: 68747484-68747484 on Build GRCh38
rs767766595 Chr.10: 68747440-68747440 on Build GRCh38
rs920997556 Chr.10: 68747445-68747445 on Build GRCh38
rs773665000 Chr.10: 68747409-68747409 on Build GRCh38
rs3216071 Chr.10: 68747339-68747340 on Build GRCh38
rs1018344732 Chr.10: 68747681-68747681 on Build GRCh38
rs749649063 Chr.10: 68747410-68747410 on Build GRCh38
rs749700291 Chr.10: 68747577-68747577 on Build GRCh38
rs145396018 Chr.10: 68747474-68747474 on Build GRCh38
rs958583895 Chr.10: 68747299-68747299 on Build GRCh38
rs368798069 Chr.10: 68747617-68747617 on Build GRCh38
rs35776267 Chr.10: 68747541-68747541 on Build GRCh38
rs779635915 Chr.10: 68747368-68747368 on Build GRCh38
rs199831400 Chr.10: 68747428-68747428 on Build GRCh38
rs1057239860 Chr.10: 68747618-68747618 on Build GRCh38
rs762363755 Chr.10: 68747340-68747340 on Build GRCh38
rs540519590 Chr.10: 68747640-68747640 on Build GRCh38
rs769177445 Chr.10: 68747223-68747223 on Build GRCh38
rs1012792713 Chr.10: 68747625-68747625 on Build GRCh38
rs894543745 Chr.10: 68747623-68747623 on Build GRCh38
rs911209223 Chr.10: 68747300-68747300 on Build GRCh38
rs772154720 Chr.10: 68747612-68747612 on Build GRCh38
rs556091058 Chr.10: 68747638-68747638 on Build GRCh38
rs758099346 Chr.10: 68747361-68747361 on Build GRCh38
rs201058682 Chr.10: 68747359-68747359 on Build GRCh38
rs758152383 Chr.10: 68747493-68747493 on Build GRCh38
rs377059355 Chr.10: 68747356-68747356 on Build GRCh38
rs752117213 Chr.10: 68747526-68747526 on Build GRCh38
rs763300893 Chr.10: 68747290-68747290 on Build GRCh38
rs774620268 Chr.10: 68747321-68747321 on Build GRCh38
rs1002966204 Chr.10: 68747262-68747262 on Build GRCh38

View Full Product Details