Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73821467-73821868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2G
Entrez Gene ID
818
Gene Name
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases
CAMK, CAMK-II, CAMKG
UniGene
Hs.523045
Gene Chromosome Location
Chr.10: 73812501-73874591 on Build GRCh38
Transcripts
NM_001204492.1 NM_001222.3 NM_001320898.1 NM_172169.2 NM_172170.4 NM_172171.2 NM_172173.2 XM_005270195.1 XM_005270197.1 XM_005270198.1 XM_005270199.1 XM_005270200.1 XM_005270201.1 XM_005270203.1 XM_005270205.1 XM_006717993.1 XM_006717996.1 XM_006717997.1 XM_006717998.1 XM_011540225.2 XM_017016716.1 XM_017016717.1 XM_017016718.1 XM_017016719.1 XM_017016720.1 XM_017016721.1 XM_017016722.1 XM_017016723.1 XM_017016724.1 XM_017016725.1 XM_017016726.1 XM_017016727.1 XM_017016728.1 XM_017016729.1 XM_017016730.1 XM_017016731.1 XM_017016732.1 XM_017016733.1 XM_017016734.1 XM_017016735.1 XM_017016736.1 XM_017016737.1 XM_017016738.1 XM_017016739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3738723 c.1116G>C p.E372D Chr.10: 73821719-73821719 on Build GRCh38
390026 c.1036-2A>T p.? Chr.10: 73821732-73821732 on Build GRCh38
6376522 c.1141G>T p.E381* Chr.10: 73821694-73821694 on Build GRCh38
6376523 c.1168G>T p.E390* Chr.10: 73821694-73821694 on Build GRCh38
3738724 c.1047G>C p.E349D Chr.10: 73821719-73821719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043810848 Chr.10: 73821804-73821804 on Build GRCh38
rs987045536 Chr.10: 73821823-73821823 on Build GRCh38
rs569992176 Chr.10: 73821533-73821533 on Build GRCh38
rs753933434 Chr.10: 73821747-73821747 on Build GRCh38
rs149999234 Chr.10: 73821710-73821710 on Build GRCh38
rs368788909 Chr.10: 73821758-73821758 on Build GRCh38
rs199947330 Chr.10: 73821646-73821646 on Build GRCh38
rs1025881089 Chr.10: 73821573-73821573 on Build GRCh38
rs139256535 Chr.10: 73821684-73821684 on Build GRCh38
rs946797621 Chr.10: 73821745-73821745 on Build GRCh38
rs778200735 Chr.10: 73821673-73821674 on Build GRCh38
rs914046302 Chr.10: 73821734-73821734 on Build GRCh38
rs758834975 Chr.10: 73821532-73821532 on Build GRCh38
rs146801903 Chr.10: 73821521-73821521 on Build GRCh38
rs577533660 Chr.10: 73821766-73821766 on Build GRCh38
rs769509185 Chr.10: 73821670-73821670 on Build GRCh38
rs758656005 Chr.10: 73821642-73821642 on Build GRCh38
rs553043141 Chr.10: 73821756-73821756 on Build GRCh38
rs537382697 Chr.10: 73821626-73821626 on Build GRCh38
rs747559332 Chr.10: 73821676-73821676 on Build GRCh38
rs1012427563 Chr.10: 73821601-73821601 on Build GRCh38
rs959891066 Chr.10: 73821581-73821581 on Build GRCh38
rs951163406 Chr.10: 73821579-73821579 on Build GRCh38
rs747553597 Chr.10: 73821661-73821661 on Build GRCh38
rs774341357 Chr.10: 73821698-73821698 on Build GRCh38
rs960153222 Chr.10: 73821567-73821567 on Build GRCh38
rs768345970 Chr.10: 73821648-73821648 on Build GRCh38
rs778957814 Chr.10: 73821635-73821635 on Build GRCh38
rs770964600 Chr.10: 73821732-73821732 on Build GRCh38
rs976077533 Chr.10: 73821631-73821631 on Build GRCh38
rs780853310 Chr.10: 73821654-73821654 on Build GRCh38
rs199893895 Chr.10: 73821711-73821711 on Build GRCh38
rs922910876 Chr.10: 73821632-73821632 on Build GRCh38
rs780563403 Chr.10: 73821774-73821774 on Build GRCh38
rs780753128 Chr.10: 73821645-73821645 on Build GRCh38
rs773119496 Chr.10: 73821493-73821493 on Build GRCh38
rs750825887 Chr.10: 73821636-73821636 on Build GRCh38
rs1014122647 Chr.10: 73821572-73821572 on Build GRCh38
rs983805703 Chr.10: 73821594-73821594 on Build GRCh38
rs1010899518 Chr.10: 73821490-73821490 on Build GRCh38
rs909570091 Chr.10: 73821624-73821624 on Build GRCh38
rs772709641 Chr.10: 73821671-73821671 on Build GRCh38
rs750592046 Chr.10: 73821764-73821764 on Build GRCh38
rs762855777 Chr.10: 73821678-73821678 on Build GRCh38

View Full Product Details