Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84184594-84185095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf99
Entrez Gene ID
387695
Gene Name
chromosome 10 open reading frame 99
Gene Aliases
AP-57, CSBF, UNQ1833
UniGene
Hs.298713
Gene Chromosome Location
Chr.10: 84173764-84185294 on Build GRCh38
Transcripts
NM_207373.2 XM_005269811.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3738749 c.229G>A p.A77T Chr.10: 84184749-84184749 on Build GRCh38
6613971 c.171G>A p.V57V Chr.10: 84184691-84184691 on Build GRCh38
3807832 c.210C>A p.R70R Chr.10: 84184730-84184730 on Build GRCh38
1686634 c.161-1G>A p.? Chr.10: 84184680-84184680 on Build GRCh38
7357963 c.209G>A p.R70H Chr.10: 84184729-84184729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016923166 Chr.10: 84184837-84184837 on Build GRCh38
rs779725187 Chr.10: 84184684-84184684 on Build GRCh38
rs144656005 Chr.10: 84184729-84184729 on Build GRCh38
rs369868550 Chr.10: 84184938-84184938 on Build GRCh38
rs567088504 Chr.10: 84184716-84184716 on Build GRCh38
rs776558772 Chr.10: 84184693-84184693 on Build GRCh38
rs755117983 Chr.10: 84184630-84184630 on Build GRCh38
rs372484418 Chr.10: 84184653-84184653 on Build GRCh38
rs763251984 Chr.10: 84184763-84184764 on Build GRCh38
rs943147673 Chr.10: 84184975-84184975 on Build GRCh38
rs76221724 Chr.10: 84184760-84184760 on Build GRCh38
rs934297962 Chr.10: 84184750-84184750 on Build GRCh38
rs933795384 Chr.10: 84184730-84184730 on Build GRCh38
rs534541807 Chr.10: 84184654-84184654 on Build GRCh38
rs375704720 Chr.10: 84184803-84184803 on Build GRCh38
rs551092841 Chr.10: 84185046-84185046 on Build GRCh38
rs757787208 Chr.10: 84184747-84184747 on Build GRCh38
rs142856023 Chr.10: 84184948-84184948 on Build GRCh38
rs770750974 Chr.10: 84184798-84184798 on Build GRCh38
rs768157409 Chr.10: 84184771-84184771 on Build GRCh38
rs761669467 Chr.10: 84184696-84184696 on Build GRCh38
rs754017818 Chr.10: 84184813-84184813 on Build GRCh38
rs752771747 Chr.10: 84184633-84184633 on Build GRCh38
rs554662594 Chr.10: 84184909-84184909 on Build GRCh38
rs901678421 Chr.10: 84185000-84185000 on Build GRCh38
rs750264971 Chr.10: 84185033-84185033 on Build GRCh38
rs924446351 Chr.10: 84184945-84184945 on Build GRCh38
rs963142760 Chr.10: 84184843-84184843 on Build GRCh38
rs981678612 Chr.10: 84184922-84184922 on Build GRCh38
rs368243249 Chr.10: 84184634-84184634 on Build GRCh38
rs750789544 Chr.10: 84184754-84184754 on Build GRCh38
rs148511886 Chr.10: 84184943-84184943 on Build GRCh38
rs761950888 Chr.10: 84184817-84184817 on Build GRCh38
rs771670299 Chr.10: 84184678-84184678 on Build GRCh38
rs914243066 Chr.10: 84184970-84184970 on Build GRCh38
rs746614434 Chr.10: 84184768-84184768 on Build GRCh38
rs779355423 Chr.10: 84184672-84184672 on Build GRCh38
rs779631100 Chr.10: 84184766-84184766 on Build GRCh38
rs188597583 Chr.10: 84184632-84184632 on Build GRCh38
rs760916391 Chr.10: 84184810-84184810 on Build GRCh38
rs748108923 Chr.10: 84184783-84184783 on Build GRCh38
rs569315082 Chr.10: 84185050-84185050 on Build GRCh38
rs540069094 Chr.10: 84184968-84184968 on Build GRCh38
rs984136015 Chr.10: 84184647-84184647 on Build GRCh38
rs772836540 Chr.10: 84184721-84184721 on Build GRCh38
rs901670601 Chr.10: 84184801-84184801 on Build GRCh38
rs778490744 Chr.10: 84184636-84184636 on Build GRCh38
rs574882684 Chr.10: 84184835-84184835 on Build GRCh38
rs760895947 Chr.10: 84184742-84184742 on Build GRCh38
rs145785254 Chr.10: 84184746-84184746 on Build GRCh38
rs997325345 Chr.10: 84185009-84185009 on Build GRCh38
rs746566628 Chr.10: 84184686-84184686 on Build GRCh38
rs111940595 Chr.10: 84184675-84184675 on Build GRCh38
rs762789784 Chr.10: 84184728-84184728 on Build GRCh38
rs76313553 Chr.10: 84184919-84184919 on Build GRCh38
rs561880717 Chr.10: 84184973-84184973 on Build GRCh38
rs368942414 Chr.10: 84184790-84184790 on Build GRCh38
rs764303294 Chr.10: 84184812-84184812 on Build GRCh38
rs556583336 Chr.10: 84184832-84184832 on Build GRCh38
rs374889526 Chr.10: 84184687-84184687 on Build GRCh38
rs916112330 Chr.10: 84184892-84184892 on Build GRCh38
rs762724524 Chr.10: 84184797-84184797 on Build GRCh38
rs764213323 Chr.10: 84184743-84184743 on Build GRCh38
rs991530128 Chr.10: 84184891-84184891 on Build GRCh38
rs757711245 Chr.10: 84184669-84184669 on Build GRCh38
rs753280528 Chr.10: 84184917-84184917 on Build GRCh38
rs909858465 Chr.10: 84184858-84184858 on Build GRCh38
rs752964088 Chr.10: 84184731-84184731 on Build GRCh38
rs372322598 Chr.10: 84184794-84184794 on Build GRCh38
rs951430151 Chr.10: 84184828-84184828 on Build GRCh38
rs779538405 Chr.10: 84184752-84184752 on Build GRCh38
rs761931328 Chr.10: 84184928-84184928 on Build GRCh38
rs111744038 Chr.10: 84184929-84184929 on Build GRCh38
rs745555542 Chr.10: 84184677-84184677 on Build GRCh38
rs894247230 Chr.10: 84185002-84185002 on Build GRCh38
rs79810437 Chr.10: 84184999-84184999 on Build GRCh38

View Full Product Details