Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84204443-84204946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR1
Entrez Gene ID
92211
Gene Name
cadherin related family member 1
Gene Aliases
CORD15, PCDH21, PRCAD, RP65
UniGene
Hs.137556
Gene Chromosome Location
Chr.10: 84194635-84219621 on Build GRCh38
Transcripts
NM_001171971.2 NM_033100.3 XM_011540337.1 XM_011540338.1 XM_011540339.1 XM_011540340.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5913009 c.784-7C>T p.? Chr.10: 84204520-84204520 on Build GRCh38
1585152 c.839G>A p.R280Q Chr.10: 84204582-84204582 on Build GRCh38
4671036 c.789G>A p.S263S Chr.10: 84204532-84204532 on Build GRCh38
4559438 c.804G>A p.V268V Chr.10: 84204547-84204547 on Build GRCh38
5913010 c.784-7C>T p.? Chr.10: 84204520-84204520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117206971 Chr.10: 84204881-84204881 on Build GRCh38
rs958148889 Chr.10: 84204893-84204893 on Build GRCh38
rs757929412 Chr.10: 84204654-84204654 on Build GRCh38
rs759703580 Chr.10: 84204629-84204629 on Build GRCh38
rs756751690 Chr.10: 84204531-84204531 on Build GRCh38
rs768356867 Chr.10: 84204601-84204601 on Build GRCh38
rs923704864 Chr.10: 84204889-84204889 on Build GRCh38
rs907212691 Chr.10: 84204617-84204617 on Build GRCh38
rs372612164 Chr.10: 84204561-84204561 on Build GRCh38
rs191858417 Chr.10: 84204581-84204581 on Build GRCh38
rs559556471 Chr.10: 84204718-84204718 on Build GRCh38
rs746907059 Chr.10: 84204597-84204597 on Build GRCh38
rs201505995 Chr.10: 84204567-84204567 on Build GRCh38
rs777305743 Chr.10: 84204526-84204526 on Build GRCh38
rs945010413 Chr.10: 84204806-84204806 on Build GRCh38
rs984736133 Chr.10: 84204714-84204714 on Build GRCh38
rs764474008 Chr.10: 84204643-84204643 on Build GRCh38
rs766297600 Chr.10: 84204878-84204878 on Build GRCh38
rs766061790 Chr.10: 84204862-84204862 on Build GRCh38
rs1039018822 Chr.10: 84204803-84204803 on Build GRCh38
rs1045786614 Chr.10: 84204887-84204887 on Build GRCh38
rs766440621 Chr.10: 84204520-84204520 on Build GRCh38
rs768949250 Chr.10: 84204502-84204511 on Build GRCh38
rs762067029 Chr.10: 84204646-84204646 on Build GRCh38
rs762944021 Chr.10: 84204517-84204517 on Build GRCh38
rs372127366 Chr.10: 84204521-84204521 on Build GRCh38
rs372723405 Chr.10: 84204626-84204626 on Build GRCh38
rs575893385 Chr.10: 84204571-84204571 on Build GRCh38
rs764952003 Chr.10: 84204647-84204647 on Build GRCh38
rs1016932100 Chr.10: 84204826-84204826 on Build GRCh38
rs779501094 Chr.10: 84204656-84204656 on Build GRCh38
rs745363286 Chr.10: 84204536-84204536 on Build GRCh38
rs774702123 Chr.10: 84204628-84204628 on Build GRCh38
rs568374773 Chr.10: 84204914-84204914 on Build GRCh38
rs570370727 Chr.10: 84204859-84204859 on Build GRCh38
rs762835814 Chr.10: 84204840-84204840 on Build GRCh38
rs775316579 Chr.10: 84204510-84204510 on Build GRCh38
rs74145747 Chr.10: 84204687-84204687 on Build GRCh38
rs893154857 Chr.10: 84204500-84204500 on Build GRCh38
rs768428118 Chr.10: 84204550-84204550 on Build GRCh38
rs138512311 Chr.10: 84204627-84204627 on Build GRCh38
rs777477465 Chr.10: 84204479-84204479 on Build GRCh38
rs201384219 Chr.10: 84204524-84204524 on Build GRCh38
rs766487238 Chr.10: 84204559-84204559 on Build GRCh38
rs889707130 Chr.10: 84204774-84204774 on Build GRCh38
rs774162981 Chr.10: 84204497-84204497 on Build GRCh38
rs1017819424 Chr.10: 84204726-84204726 on Build GRCh38
rs773423395 Chr.10: 84204514-84204514 on Build GRCh38
rs765497847 Chr.10: 84204513-84204513 on Build GRCh38
rs779248066 Chr.10: 84204475-84204475 on Build GRCh38
rs4606427 Chr.10: 84204741-84204741 on Build GRCh38
rs959339386 Chr.10: 84204632-84204632 on Build GRCh38
rs776537064 Chr.10: 84204624-84204624 on Build GRCh38
rs543254581 Chr.10: 84204576-84204576 on Build GRCh38
rs936505817 Chr.10: 84204734-84204734 on Build GRCh38
rs943353871 Chr.10: 84204784-84204784 on Build GRCh38
rs760455211 Chr.10: 84204511-84204511 on Build GRCh38
rs949226250 Chr.10: 84204467-84204467 on Build GRCh38
rs746817205 Chr.10: 84204540-84204540 on Build GRCh38
rs368184418 Chr.10: 84204480-84204480 on Build GRCh38
rs748905962 Chr.10: 84204528-84204528 on Build GRCh38
rs997575984 Chr.10: 84204813-84204813 on Build GRCh38
rs775389758 Chr.10: 84204539-84204539 on Build GRCh38
rs776355721 Chr.10: 84204716-84204716 on Build GRCh38
rs530132075 Chr.10: 84204738-84204738 on Build GRCh38
rs772213107 Chr.10: 84204639-84204639 on Build GRCh38
rs577851360 Chr.10: 84204476-84204476 on Build GRCh38
rs375409928 Chr.10: 84204522-84204522 on Build GRCh38
rs553516475 Chr.10: 84204493-84204493 on Build GRCh38
rs1006388400 Chr.10: 84204822-84204822 on Build GRCh38
rs977913396 Chr.10: 84204688-84204688 on Build GRCh38
rs763184270 Chr.10: 84204558-84204558 on Build GRCh38
rs201649854 Chr.10: 84204582-84204582 on Build GRCh38
rs768106524 Chr.10: 84204523-84204523 on Build GRCh38
rs760554147 Chr.10: 84204816-84204816 on Build GRCh38
rs1024573648 Chr.10: 84204869-84204869 on Build GRCh38
rs1026219407 Chr.10: 84204683-84204683 on Build GRCh38
rs754274450 Chr.10: 84204645-84204645 on Build GRCh38
rs759431602 Chr.10: 84204505-84204505 on Build GRCh38
rs200640039 Chr.10: 84204566-84204566 on Build GRCh38
rs901938831 Chr.10: 84204804-84204804 on Build GRCh38
rs778441075 Chr.10: 84204532-84204532 on Build GRCh38
rs750024181 Chr.10: 84204653-84204653 on Build GRCh38
rs771880291 Chr.10: 84204506-84204506 on Build GRCh38
rs56780713 Chr.10: 84204487-84204487 on Build GRCh38
rs924991271 Chr.10: 84204825-84204825 on Build GRCh38
rs779967616 Chr.10: 84204596-84204596 on Build GRCh38
rs936222026 Chr.10: 84204839-84204839 on Build GRCh38
rs773335171 Chr.10: 84204551-84204551 on Build GRCh38
rs886634640 Chr.10: 84204821-84204821 on Build GRCh38
rs746489628 Chr.10: 84204642-84204642 on Build GRCh38
rs753458279 Chr.10: 84204579-84204579 on Build GRCh38
rs886047326 Chr.10: 84204591-84204591 on Build GRCh38
rs144092243 Chr.10: 84204895-84204895 on Build GRCh38
rs959618414 Chr.10: 84204844-84204844 on Build GRCh38
rs975826922 Chr.10: 84204775-84204775 on Build GRCh38
rs773999311 Chr.10: 84204571-84204571 on Build GRCh38

View Full Product Details