Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89007609-89008108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAS
Entrez Gene ID
355
Gene Name
Fas cell surface death receptor
Gene Aliases
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
UniGene
Hs.244139
Gene Chromosome Location
Chr.10: 88968429-89017061 on Build GRCh38
Transcripts
NM_000043.5 NM_001320619.1 NM_152871.3 NM_152872.3 NR_028033.3 NR_028034.3 NR_028035.3 NR_028036.3 NR_135313.1 NR_135314.1 NR_135315.1 XM_006717819.3 XM_011539764.2 XM_011539765.2 XM_011539766.2 XM_011539767.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880916 c.218G>A p.C73Y Chr.10: 89007721-89007721 on Build GRCh38
5654165 c.222A>G p.T74T Chr.10: 89007725-89007725 on Build GRCh38
4898808 c.334G>A p.G112S Chr.10: 89007837-89007837 on Build GRCh38
6013980 c.324T>G p.D108E Chr.10: 89007827-89007827 on Build GRCh38
26167 c.308G>A p.R103K Chr.10: 89007811-89007811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763495508 Chr.10: 89007711-89007711 on Build GRCh38
rs574101415 Chr.10: 89007730-89007730 on Build GRCh38
rs755792620 Chr.10: 89008027-89008027 on Build GRCh38
rs145912526 Chr.10: 89008037-89008037 on Build GRCh38
rs3218618 Chr.10: 89007917-89007917 on Build GRCh38
rs778483787 Chr.10: 89007754-89007754 on Build GRCh38
rs781281918 Chr.10: 89007724-89007724 on Build GRCh38
rs771050216 Chr.10: 89007752-89007752 on Build GRCh38
rs751654768 Chr.10: 89007720-89007720 on Build GRCh38
rs1003918686 Chr.10: 89008047-89008047 on Build GRCh38
rs899557375 Chr.10: 89008012-89008012 on Build GRCh38
rs959465921 Chr.10: 89007885-89007885 on Build GRCh38
rs939925881 Chr.10: 89007991-89007991 on Build GRCh38
rs367693795 Chr.10: 89007848-89007848 on Build GRCh38
rs745375995 Chr.10: 89007773-89007773 on Build GRCh38
rs369400533 Chr.10: 89008014-89008014 on Build GRCh38
rs2296601 Chr.10: 89007883-89007883 on Build GRCh38
rs1027082379 Chr.10: 89007693-89007693 on Build GRCh38
rs193283551 Chr.10: 89008077-89008077 on Build GRCh38
rs606231362 Chr.10: 89007839-89007840 on Build GRCh38
rs606231361 Chr.10: 89007735-89007735 on Build GRCh38
rs567050515 Chr.10: 89007967-89007967 on Build GRCh38
rs1021116485 Chr.10: 89007935-89007935 on Build GRCh38
rs759213901 Chr.10: 89007654-89007654 on Build GRCh38
rs1036815834 Chr.10: 89007765-89007765 on Build GRCh38
rs776989861 Chr.10: 89007660-89007660 on Build GRCh38
rs775030741 Chr.10: 89007801-89007801 on Build GRCh38
rs199790261 Chr.10: 89007750-89007750 on Build GRCh38
rs1004851646 Chr.10: 89007729-89007729 on Build GRCh38
rs898277057 Chr.10: 89007799-89007799 on Build GRCh38
rs777751533 Chr.10: 89007749-89007749 on Build GRCh38
rs934024817 Chr.10: 89007664-89007664 on Build GRCh38
rs187793393 Chr.10: 89007689-89007689 on Build GRCh38
rs1009768537 Chr.10: 89007923-89007923 on Build GRCh38
rs774053789 Chr.10: 89007653-89007653 on Build GRCh38
rs764241259 Chr.10: 89007817-89007818 on Build GRCh38
rs759358476 Chr.10: 89007932-89007932 on Build GRCh38
rs549492411 Chr.10: 89008003-89008003 on Build GRCh38
rs756003974 Chr.10: 89007727-89007727 on Build GRCh38
rs111667735 Chr.10: 89007977-89007977 on Build GRCh38
rs552088345 Chr.10: 89007924-89007924 on Build GRCh38
rs200239064 Chr.10: 89007667-89007667 on Build GRCh38
rs559821285 Chr.10: 89007976-89007976 on Build GRCh38
rs1015885307 Chr.10: 89007743-89007743 on Build GRCh38
rs765687911 Chr.10: 89007682-89007682 on Build GRCh38
rs763387522 Chr.10: 89007857-89007857 on Build GRCh38
rs996587887 Chr.10: 89008019-89008019 on Build GRCh38
rs770100045 Chr.10: 89007821-89007821 on Build GRCh38
rs750788102 Chr.10: 89007924-89007925 on Build GRCh38
rs751648222 Chr.10: 89007863-89007863 on Build GRCh38
rs896732294 Chr.10: 89007909-89007909 on Build GRCh38
rs770534737 Chr.10: 89007650-89007650 on Build GRCh38
rs1035238918 Chr.10: 89007892-89007892 on Build GRCh38
rs912511928 Chr.10: 89007987-89007987 on Build GRCh38
rs2229521 Chr.10: 89007725-89007725 on Build GRCh38
rs762203075 Chr.10: 89007819-89007819 on Build GRCh38
rs914023711 Chr.10: 89007641-89007641 on Build GRCh38
rs947279537 Chr.10: 89007635-89007635 on Build GRCh38
rs931077331 Chr.10: 89008068-89008068 on Build GRCh38
rs915209760 Chr.10: 89007920-89007920 on Build GRCh38
rs559727135 Chr.10: 89008028-89008028 on Build GRCh38
rs377065670 Chr.10: 89007659-89007659 on Build GRCh38
rs752370494 Chr.10: 89008006-89008006 on Build GRCh38
rs767472244 Chr.10: 89007992-89007992 on Build GRCh38
rs372222615 Chr.10: 89007871-89007871 on Build GRCh38
rs1050992298 Chr.10: 89007670-89007670 on Build GRCh38
rs766429180 Chr.10: 89007719-89007719 on Build GRCh38
rs979513467 Chr.10: 89007963-89007963 on Build GRCh38
rs2031611 Chr.10: 89007638-89007638 on Build GRCh38
rs771699698 Chr.10: 89007782-89007782 on Build GRCh38
rs905498596 Chr.10: 89007880-89007880 on Build GRCh38
rs879321809 Chr.10: 89008080-89008080 on Build GRCh38
rs968145053 Chr.10: 89007960-89007960 on Build GRCh38
rs766940746 Chr.10: 89007861-89007861 on Build GRCh38

View Full Product Details