Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96981199-96981704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf12
Entrez Gene ID
26148
Gene Name
chromosome 10 open reading frame 12
Gene Aliases
-
UniGene
Hs.427927
Gene Chromosome Location
Chr.10: 96981284-96985828 on Build GRCh38
Transcripts
NM_015652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4410183 c.84T>C p.I28I Chr.10: 96981474-96981474 on Build GRCh38
1350135 c.203G>A p.R68H Chr.10: 96981593-96981593 on Build GRCh38
6067100 c.203G>T p.R68L Chr.10: 96981593-96981593 on Build GRCh38
1321967 c.210T>C p.D70D Chr.10: 96981600-96981600 on Build GRCh38
7210348 c.190A>T p.K64* Chr.10: 96981580-96981580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373132958 Chr.10: 96981354-96981354 on Build GRCh38
rs1033650920 Chr.10: 96981380-96981380 on Build GRCh38
rs141108014 Chr.10: 96981603-96981603 on Build GRCh38
rs201530736 Chr.10: 96981449-96981449 on Build GRCh38
rs758887432 Chr.10: 96981492-96981492 on Build GRCh38
rs912759878 Chr.10: 96981656-96981656 on Build GRCh38
rs374414530 Chr.10: 96981417-96981417 on Build GRCh38
rs200185687 Chr.10: 96981593-96981593 on Build GRCh38
rs746295089 Chr.10: 96981341-96981341 on Build GRCh38
rs375341794 Chr.10: 96981537-96981537 on Build GRCh38
rs573183510 Chr.10: 96981302-96981302 on Build GRCh38
rs183592849 Chr.10: 96981263-96981263 on Build GRCh38
rs767060322 Chr.10: 96981453-96981453 on Build GRCh38
rs201361952 Chr.10: 96981433-96981433 on Build GRCh38
rs544595807 Chr.10: 96981583-96981583 on Build GRCh38
rs755640559 Chr.10: 96981477-96981477 on Build GRCh38
rs779351732 Chr.10: 96981329-96981329 on Build GRCh38
rs940202054 Chr.10: 96981672-96981672 on Build GRCh38
rs768799814 Chr.10: 96981385-96981385 on Build GRCh38
rs771211882 Chr.10: 96981504-96981504 on Build GRCh38
rs777791004 Chr.10: 96981641-96981641 on Build GRCh38
rs1043631523 Chr.10: 96981466-96981466 on Build GRCh38
rs897299773 Chr.10: 96981597-96981597 on Build GRCh38
rs747183609 Chr.10: 96981462-96981462 on Build GRCh38
rs530129629 Chr.10: 96981661-96981661 on Build GRCh38
rs370241836 Chr.10: 96981467-96981467 on Build GRCh38
rs772565005 Chr.10: 96981346-96981346 on Build GRCh38
rs762829797 Chr.10: 96981540-96981540 on Build GRCh38
rs774067529 Chr.10: 96981566-96981566 on Build GRCh38
rs765616605 Chr.10: 96981591-96981591 on Build GRCh38
rs754585206 Chr.10: 96981450-96981450 on Build GRCh38
rs750118202 Chr.10: 96981232-96981232 on Build GRCh38
rs751912372 Chr.10: 96981617-96981617 on Build GRCh38
rs748232289 Chr.10: 96981533-96981533 on Build GRCh38
rs757640172 Chr.10: 96981304-96981304 on Build GRCh38
rs761811169 Chr.10: 96981393-96981393 on Build GRCh38
rs780518988 Chr.10: 96981282-96981282 on Build GRCh38
rs957948080 Chr.10: 96981485-96981485 on Build GRCh38
rs921688967 Chr.10: 96981231-96981231 on Build GRCh38
rs1038669957 Chr.10: 96981230-96981230 on Build GRCh38
rs945290597 Chr.10: 96981444-96981444 on Build GRCh38
rs1026931163 Chr.10: 96981322-96981322 on Build GRCh38
rs150728668 Chr.10: 96981361-96981361 on Build GRCh38
rs759200625 Chr.10: 96981573-96981573 on Build GRCh38
rs760148082 Chr.10: 96981592-96981592 on Build GRCh38
rs780514163 Chr.10: 96981494-96981494 on Build GRCh38
rs750985246 Chr.10: 96981595-96981595 on Build GRCh38
rs1037672968 Chr.10: 96981313-96981313 on Build GRCh38
rs758058212 Chr.10: 96981257-96981257 on Build GRCh38
rs368856693 Chr.10: 96981649-96981649 on Build GRCh38
rs762802894 Chr.10: 96981426-96981426 on Build GRCh38
rs779358670 Chr.10: 96981488-96981488 on Build GRCh38
rs150264369 Chr.10: 96981626-96981626 on Build GRCh38
rs772013606 Chr.10: 96981299-96981299 on Build GRCh38
rs965856029 Chr.10: 96981407-96981407 on Build GRCh38
rs746350416 Chr.10: 96981491-96981491 on Build GRCh38
rs768692125 Chr.10: 96981510-96981510 on Build GRCh38
rs780567495 Chr.10: 96981610-96981610 on Build GRCh38
rs575225989 Chr.10: 96981225-96981225 on Build GRCh38
rs754532101 Chr.10: 96981264-96981264 on Build GRCh38
rs749278456 Chr.10: 96981268-96981268 on Build GRCh38
rs1027151757 Chr.10: 96981680-96981680 on Build GRCh38
rs937731973 Chr.10: 96981534-96981534 on Build GRCh38
rs781333565 Chr.10: 96981627-96981627 on Build GRCh38
rs770743899 Chr.10: 96981555-96981555 on Build GRCh38
rs995740213 Chr.10: 96981303-96981303 on Build GRCh38
rs773186003 Chr.10: 96981422-96981422 on Build GRCh38
rs781342632 Chr.10: 96981527-96981527 on Build GRCh38
rs536489900 Chr.10: 96981262-96981262 on Build GRCh38
rs896265113 Chr.10: 96981544-96981544 on Build GRCh38
rs11188980 Chr.10: 96981536-96981536 on Build GRCh38
rs563045829 Chr.10: 96981639-96981639 on Build GRCh38
rs201068399 Chr.10: 96981392-96981392 on Build GRCh38
rs748370218 Chr.10: 96981628-96981628 on Build GRCh38

View Full Product Details