Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96983931-96984473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf12
Entrez Gene ID
26148
Gene Name
chromosome 10 open reading frame 12
Gene Aliases
-
UniGene
Hs.427927
Gene Chromosome Location
Chr.10: 96981284-96985828 on Build GRCh38
Transcripts
NM_015652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603916 c.2723A>C p.Q908P Chr.10: 96984113-96984113 on Build GRCh38
4806991 c.2731G>T p.A911S Chr.10: 96984121-96984121 on Build GRCh38
921963 c.2868T>C p.D956D Chr.10: 96984258-96984258 on Build GRCh38
3868210 c.2756C>T p.S919L Chr.10: 96984146-96984146 on Build GRCh38
4414811 c.3013A>C p.N1005H Chr.10: 96984403-96984403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199889747 Chr.10: 96984314-96984314 on Build GRCh38
rs775720838 Chr.10: 96984367-96984367 on Build GRCh38
rs763427126 Chr.10: 96984119-96984119 on Build GRCh38
rs761945854 Chr.10: 96984383-96984383 on Build GRCh38
rs758383614 Chr.10: 96984046-96984046 on Build GRCh38
rs778501806 Chr.10: 96984185-96984185 on Build GRCh38
rs137993125 Chr.10: 96984040-96984040 on Build GRCh38
rs750571955 Chr.10: 96984134-96984134 on Build GRCh38
rs752604704 Chr.10: 96983986-96983986 on Build GRCh38
rs559862086 Chr.10: 96984335-96984335 on Build GRCh38
rs765204328 Chr.10: 96984316-96984316 on Build GRCh38
rs755335544 Chr.10: 96984373-96984373 on Build GRCh38
rs750307576 Chr.10: 96984319-96984319 on Build GRCh38
rs185249748 Chr.10: 96984262-96984262 on Build GRCh38
rs372411239 Chr.10: 96984087-96984087 on Build GRCh38
rs367971002 Chr.10: 96984048-96984048 on Build GRCh38
rs778649685 Chr.10: 96984074-96984074 on Build GRCh38
rs534749612 Chr.10: 96984131-96984131 on Build GRCh38
rs201305477 Chr.10: 96984088-96984088 on Build GRCh38
rs377557529 Chr.10: 96984378-96984378 on Build GRCh38
rs772372887 Chr.10: 96984361-96984361 on Build GRCh38
rs780271847 Chr.10: 96984349-96984349 on Build GRCh38
rs764040208 Chr.10: 96984294-96984294 on Build GRCh38
rs745525574 Chr.10: 96984198-96984198 on Build GRCh38
rs759688914 Chr.10: 96984124-96984124 on Build GRCh38
rs771331717 Chr.10: 96984251-96984251 on Build GRCh38
rs753668414 Chr.10: 96984175-96984175 on Build GRCh38
rs750854444 Chr.10: 96984011-96984011 on Build GRCh38
rs752833926 Chr.10: 96984147-96984147 on Build GRCh38
rs375398676 Chr.10: 96984005-96984005 on Build GRCh38
rs767764456 Chr.10: 96984264-96984264 on Build GRCh38
rs751712472 Chr.10: 96984029-96984029 on Build GRCh38
rs779904619 Chr.10: 96984332-96984332 on Build GRCh38
rs757080916 Chr.10: 96984307-96984307 on Build GRCh38
rs751228929 Chr.10: 96984445-96984445 on Build GRCh38
rs760830884 Chr.10: 96984278-96984278 on Build GRCh38
rs142383450 Chr.10: 96984041-96984041 on Build GRCh38
rs138976357 Chr.10: 96984346-96984346 on Build GRCh38
rs746560727 Chr.10: 96984239-96984239 on Build GRCh38
rs768032236 Chr.10: 96984244-96984244 on Build GRCh38
rs757106790 Chr.10: 96984180-96984180 on Build GRCh38
rs190573704 Chr.10: 96984328-96984328 on Build GRCh38
rs765302956 Chr.10: 96984399-96984399 on Build GRCh38
rs767086232 Chr.10: 96983979-96983979 on Build GRCh38
rs371731816 Chr.10: 96983974-96983974 on Build GRCh38
rs1045414633 Chr.10: 96984270-96984270 on Build GRCh38
rs750457449 Chr.10: 96984132-96984132 on Build GRCh38
rs972045340 Chr.10: 96984042-96984042 on Build GRCh38
rs201325133 Chr.10: 96984255-96984255 on Build GRCh38
rs754551988 Chr.10: 96984336-96984336 on Build GRCh38
rs539559928 Chr.10: 96984417-96984417 on Build GRCh38
rs763329309 Chr.10: 96984249-96984249 on Build GRCh38
rs1023252122 Chr.10: 96983998-96983998 on Build GRCh38
rs779195157 Chr.10: 96984402-96984402 on Build GRCh38
rs779845490 Chr.10: 96984232-96984232 on Build GRCh38
rs771906215 Chr.10: 96984203-96984203 on Build GRCh38
rs1013501287 Chr.10: 96983990-96983990 on Build GRCh38
rs756153716 Chr.10: 96984162-96984162 on Build GRCh38
rs919121920 Chr.10: 96984171-96984171 on Build GRCh38
rs368991238 Chr.10: 96984446-96984446 on Build GRCh38
rs145759511 Chr.10: 96984295-96984295 on Build GRCh38
rs766667771 Chr.10: 96984013-96984013 on Build GRCh38
rs941243466 Chr.10: 96984222-96984222 on Build GRCh38
rs767574275 Chr.10: 96984102-96984102 on Build GRCh38
rs527405169 Chr.10: 96984093-96984093 on Build GRCh38
rs760742025 Chr.10: 96984369-96984369 on Build GRCh38
rs1039934705 Chr.10: 96984236-96984236 on Build GRCh38
rs143456541 Chr.10: 96984267-96984267 on Build GRCh38
rs376966258 Chr.10: 96984120-96984120 on Build GRCh38
rs749146230 Chr.10: 96984068-96984068 on Build GRCh38
rs776628659 Chr.10: 96984382-96984382 on Build GRCh38
rs77652574 Chr.10: 96984306-96984306 on Build GRCh38
rs756196923 Chr.10: 96984227-96984227 on Build GRCh38
rs981432655 Chr.10: 96984056-96984056 on Build GRCh38
rs984807042 Chr.10: 96984089-96984089 on Build GRCh38
rs200981330 Chr.10: 96984350-96984350 on Build GRCh38
rs752637557 Chr.10: 96984043-96984043 on Build GRCh38
rs777814463 Chr.10: 96984174-96984174 on Build GRCh38
rs193262451 Chr.10: 96984410-96984410 on Build GRCh38
rs369997809 Chr.10: 96984121-96984121 on Build GRCh38
rs773564203 Chr.10: 96984101-96984101 on Build GRCh38
rs762339616 Chr.10: 96983993-96983993 on Build GRCh38
rs951990101 Chr.10: 96984224-96984224 on Build GRCh38
rs770609460 Chr.10: 96984073-96984073 on Build GRCh38
rs767598808 Chr.10: 96984135-96984135 on Build GRCh38
rs201744063 Chr.10: 96984245-96984245 on Build GRCh38
rs146052398 Chr.10: 96984052-96984052 on Build GRCh38
rs141296765 Chr.10: 96984352-96984352 on Build GRCh38

View Full Product Details