Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97572417-97572921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
UniGene
Hs.73708
Gene Chromosome Location
Chr.10: 97572441-97583884 on Build GRCh38
Transcripts
NM_001129981.2 NM_001291218.1 NM_001291219.1 NM_020349.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442423 c.60C>T p.A20A Chr.10: 97572767-97572767 on Build GRCh38
1702870 c.59C>T p.A20V Chr.10: 97572766-97572766 on Build GRCh38
6857214 c.114G>C p.V38V Chr.10: 97572821-97572821 on Build GRCh38
4563994 c.9G>A p.K3K Chr.10: 97572716-97572716 on Build GRCh38
6423392 c.57G>T p.E19D Chr.10: 97572764-97572764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373923057 Chr.10: 97572812-97572812 on Build GRCh38
rs1023125733 Chr.10: 97572600-97572600 on Build GRCh38
rs201157692 Chr.10: 97572762-97572762 on Build GRCh38
rs762679689 Chr.10: 97572525-97572525 on Build GRCh38
rs1029444093 Chr.10: 97572500-97572500 on Build GRCh38
rs748954327 Chr.10: 97572831-97572831 on Build GRCh38
rs1012694247 Chr.10: 97572599-97572599 on Build GRCh38
rs537620334 Chr.10: 97572750-97572750 on Build GRCh38
rs541569245 Chr.10: 97572800-97572800 on Build GRCh38
rs41290450 Chr.10: 97572598-97572598 on Build GRCh38
rs750673921 Chr.10: 97572799-97572799 on Build GRCh38
rs776219303 Chr.10: 97572660-97572660 on Build GRCh38
rs568513038 Chr.10: 97572532-97572532 on Build GRCh38
rs141100599 Chr.10: 97572761-97572761 on Build GRCh38
rs971454862 Chr.10: 97572451-97572451 on Build GRCh38
rs976890472 Chr.10: 97572471-97572471 on Build GRCh38
rs553343852 Chr.10: 97572801-97572801 on Build GRCh38
rs765589738 Chr.10: 97572882-97572882 on Build GRCh38
rs745583105 Chr.10: 97572621-97572622 on Build GRCh38
rs530481073 Chr.10: 97572880-97572880 on Build GRCh38
rs909361465 Chr.10: 97572846-97572846 on Build GRCh38
rs754093162 Chr.10: 97572718-97572718 on Build GRCh38
rs950976621 Chr.10: 97572689-97572689 on Build GRCh38
rs374472766 Chr.10: 97572765-97572765 on Build GRCh38
rs1005154880 Chr.10: 97572677-97572677 on Build GRCh38
rs950925207 Chr.10: 97572483-97572483 on Build GRCh38
rs149677735 Chr.10: 97572693-97572693 on Build GRCh38
rs997896675 Chr.10: 97572742-97572742 on Build GRCh38
rs746966764 Chr.10: 97572741-97572741 on Build GRCh38
rs12219064 Chr.10: 97572466-97572466 on Build GRCh38
rs909355875 Chr.10: 97572516-97572516 on Build GRCh38
rs768190370 Chr.10: 97572685-97572685 on Build GRCh38
rs903102464 Chr.10: 97572575-97572575 on Build GRCh38
rs758493989 Chr.10: 97572734-97572734 on Build GRCh38
rs761259124 Chr.10: 97572785-97572785 on Build GRCh38
rs754268354 Chr.10: 97572790-97572790 on Build GRCh38
rs770433937 Chr.10: 97572835-97572835 on Build GRCh38
rs376730113 Chr.10: 97572673-97572673 on Build GRCh38
rs199730471 Chr.10: 97572818-97572818 on Build GRCh38
rs773969444 Chr.10: 97572842-97572842 on Build GRCh38
rs138627738 Chr.10: 97572759-97572759 on Build GRCh38
rs934529051 Chr.10: 97572584-97572584 on Build GRCh38
rs200310346 Chr.10: 97572774-97572774 on Build GRCh38
rs779956593 Chr.10: 97572738-97572738 on Build GRCh38
rs777145465 Chr.10: 97572878-97572878 on Build GRCh38
rs775406707 Chr.10: 97572522-97572522 on Build GRCh38
rs780964391 Chr.10: 97572813-97572813 on Build GRCh38
rs181283492 Chr.10: 97572822-97572822 on Build GRCh38
rs776543145 Chr.10: 97572752-97572752 on Build GRCh38
rs148057686 Chr.10: 97572849-97572849 on Build GRCh38
rs886087213 Chr.10: 97572836-97572836 on Build GRCh38
rs549723138 Chr.10: 97572513-97572513 on Build GRCh38
rs563936600 Chr.10: 97572446-97572446 on Build GRCh38
rs762525412 Chr.10: 97572683-97572683 on Build GRCh38
rs867172659 Chr.10: 97572547-97572547 on Build GRCh38
rs17107819 Chr.10: 97572622-97572624 on Build GRCh38
rs1049233988 Chr.10: 97572843-97572843 on Build GRCh38
rs769253923 Chr.10: 97572850-97572850 on Build GRCh38
rs978036000 Chr.10: 97572566-97572566 on Build GRCh38
rs765534840 Chr.10: 97572795-97572795 on Build GRCh38
rs747948721 Chr.10: 97572758-97572758 on Build GRCh38
rs764526377 Chr.10: 97572708-97572708 on Build GRCh38
rs981748164 Chr.10: 97572465-97572465 on Build GRCh38
rs984928295 Chr.10: 97572485-97572485 on Build GRCh38
rs761537111 Chr.10: 97572663-97572663 on Build GRCh38
rs373348579 Chr.10: 97572784-97572785 on Build GRCh38
rs762142474 Chr.10: 97572794-97572794 on Build GRCh38
rs147018759 Chr.10: 97572817-97572817 on Build GRCh38
rs535580900 Chr.10: 97572595-97572595 on Build GRCh38
rs145461170 Chr.10: 97572704-97572704 on Build GRCh38
rs185141703 Chr.10: 97572879-97572879 on Build GRCh38
rs751575047 Chr.10: 97572805-97572805 on Build GRCh38
rs977962383 Chr.10: 97572781-97572781 on Build GRCh38
rs943484062 Chr.10: 97572523-97572523 on Build GRCh38
rs547593264 Chr.10: 97572655-97572655 on Build GRCh38
rs12221474 Chr.10: 97572731-97572731 on Build GRCh38
rs370514584 Chr.10: 97572806-97572806 on Build GRCh38
rs533991801 Chr.10: 97572654-97572654 on Build GRCh38
rs757534824 Chr.10: 97572719-97572719 on Build GRCh38
rs768400636 Chr.10: 97572528-97572528 on Build GRCh38
rs547726490 Chr.10: 97572597-97572597 on Build GRCh38
rs773670423 Chr.10: 97572784-97572784 on Build GRCh38
rs906902618 Chr.10: 97572607-97572607 on Build GRCh38
rs953823404 Chr.10: 97572754-97572754 on Build GRCh38

View Full Product Details