Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97583485-97583984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
UniGene
Hs.73708
Gene Chromosome Location
Chr.10: 97572441-97583884 on Build GRCh38
Transcripts
NM_001129981.2 NM_001291218.1 NM_001291219.1 NM_020349.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978980 c.984G>C p.R328R Chr.10: 97583626-97583626 on Build GRCh38
3868222 c.973G>A p.A325T Chr.10: 97583615-97583615 on Build GRCh38
6600454 c.1008G>A p.P336P Chr.10: 97583650-97583650 on Build GRCh38
3442427 c.951G>A p.T317T Chr.10: 97583593-97583593 on Build GRCh38
3808132 c.1051C>T p.R351* Chr.10: 97583693-97583693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776515964 Chr.10: 97583560-97583560 on Build GRCh38
rs138862885 Chr.10: 97583711-97583711 on Build GRCh38
rs769717666 Chr.10: 97583570-97583570 on Build GRCh38
rs375635271 Chr.10: 97583629-97583629 on Build GRCh38
rs766820934 Chr.10: 97583698-97583698 on Build GRCh38
rs201529513 Chr.10: 97583665-97583665 on Build GRCh38
rs773029162 Chr.10: 97583575-97583575 on Build GRCh38
rs760333902 Chr.10: 97583774-97583774 on Build GRCh38
rs866083085 Chr.10: 97583714-97583714 on Build GRCh38
rs771808048 Chr.10: 97583770-97583770 on Build GRCh38
rs752822702 Chr.10: 97583729-97583729 on Build GRCh38
rs868355879 Chr.10: 97583795-97583795 on Build GRCh38
rs776264500 Chr.10: 97583776-97583776 on Build GRCh38
rs752139699 Chr.10: 97583593-97583593 on Build GRCh38
rs746233743 Chr.10: 97583549-97583549 on Build GRCh38
rs901045727 Chr.10: 97583798-97583798 on Build GRCh38
rs775418737 Chr.10: 97583605-97583605 on Build GRCh38
rs1011293931 Chr.10: 97583788-97583788 on Build GRCh38
rs915880099 Chr.10: 97583931-97583931 on Build GRCh38
rs1032357416 Chr.10: 97583875-97583875 on Build GRCh38
rs566640055 Chr.10: 97583625-97583625 on Build GRCh38
rs1041273451 Chr.10: 97583760-97583760 on Build GRCh38
rs931316227 Chr.10: 97583942-97583942 on Build GRCh38
rs779481163 Chr.10: 97583547-97583547 on Build GRCh38
rs191245945 Chr.10: 97583964-97583964 on Build GRCh38
rs199891248 Chr.10: 97583567-97583567 on Build GRCh38
rs755290843 Chr.10: 97583564-97583564 on Build GRCh38
rs781246969 Chr.10: 97583717-97583717 on Build GRCh38
rs775109690 Chr.10: 97583668-97583668 on Build GRCh38
rs780442859 Chr.10: 97583555-97583555 on Build GRCh38
rs553884274 Chr.10: 97583529-97583529 on Build GRCh38
rs967197980 Chr.10: 97583837-97583837 on Build GRCh38
rs370654638 Chr.10: 97583559-97583559 on Build GRCh38
rs770719162 Chr.10: 97583654-97583654 on Build GRCh38
rs1000838610 Chr.10: 97583872-97583872 on Build GRCh38
rs750967203 Chr.10: 97583693-97583693 on Build GRCh38
rs991825234 Chr.10: 97583865-97583865 on Build GRCh38
rs773960501 Chr.10: 97583655-97583655 on Build GRCh38
rs780214039 Chr.10: 97583624-97583624 on Build GRCh38
rs747544043 Chr.10: 97583755-97583755 on Build GRCh38
rs200702482 Chr.10: 97583543-97583543 on Build GRCh38
rs1040939137 Chr.10: 97583631-97583631 on Build GRCh38
rs879151949 Chr.10: 97583523-97583523 on Build GRCh38
rs749236620 Chr.10: 97583741-97583741 on Build GRCh38
rs766898912 Chr.10: 97583530-97583530 on Build GRCh38
rs745622512 Chr.10: 97583757-97583757 on Build GRCh38
rs532310933 Chr.10: 97583557-97583557 on Build GRCh38
rs763492039 Chr.10: 97583694-97583694 on Build GRCh38
rs945945093 Chr.10: 97583845-97583845 on Build GRCh38
rs367622586 Chr.10: 97583730-97583730 on Build GRCh38
rs977336023 Chr.10: 97583633-97583633 on Build GRCh38
rs143138428 Chr.10: 97583578-97583578 on Build GRCh38
rs959833562 Chr.10: 97583545-97583545 on Build GRCh38
rs767862109 Chr.10: 97583566-97583566 on Build GRCh38
rs767651764 Chr.10: 97583658-97583658 on Build GRCh38
rs201115762 Chr.10: 97583752-97583752 on Build GRCh38
rs765811149 Chr.10: 97583599-97583599 on Build GRCh38
rs575114826 Chr.10: 97583953-97583953 on Build GRCh38
rs895226691 Chr.10: 97583938-97583938 on Build GRCh38
rs752868828 Chr.10: 97583722-97583722 on Build GRCh38
rs773157878 Chr.10: 97583650-97583650 on Build GRCh38
rs148239796 Chr.10: 97583589-97583589 on Build GRCh38
rs1041279084 Chr.10: 97583797-97583797 on Build GRCh38
rs760242035 Chr.10: 97583674-97583674 on Build GRCh38
rs141723621 Chr.10: 97583613-97583613 on Build GRCh38
rs755142422 Chr.10: 97583626-97583626 on Build GRCh38
rs558102057 Chr.10: 97583862-97583862 on Build GRCh38
rs771507986 Chr.10: 97583851-97583851 on Build GRCh38
rs377348889 Chr.10: 97583554-97583554 on Build GRCh38
rs548309453 Chr.10: 97583590-97583590 on Build GRCh38
rs115015465 Chr.10: 97583775-97583775 on Build GRCh38
rs757785021 Chr.10: 97583544-97583544 on Build GRCh38
rs953171507 Chr.10: 97583781-97583781 on Build GRCh38
rs372700220 Chr.10: 97583649-97583649 on Build GRCh38
rs751864202 Chr.10: 97583709-97583709 on Build GRCh38
rs767072290 Chr.10: 97583592-97583592 on Build GRCh38
rs866978367 Chr.10: 97583856-97583856 on Build GRCh38
rs766115665 Chr.10: 97583586-97583586 on Build GRCh38
rs145073956 Chr.10: 97583692-97583692 on Build GRCh38
rs368630525 Chr.10: 97583630-97583630 on Build GRCh38
rs556715740 Chr.10: 97583920-97583920 on Build GRCh38
rs749185346 Chr.10: 97583651-97583651 on Build GRCh38
rs775034618 Chr.10: 97583771-97583771 on Build GRCh38

View Full Product Details