Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97864949-97865465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

SNP Detail

SNP ID Chromosome Location
rs551080516 Chr.10: 97865206-97865206 on Build GRCh38
rs965644962 Chr.10: 97865093-97865093 on Build GRCh38
rs114355950 Chr.10: 97865425-97865425 on Build GRCh38
rs567115022 Chr.10: 97865345-97865345 on Build GRCh38
rs538377935 Chr.10: 97865411-97865411 on Build GRCh38
rs904277733 Chr.10: 97864973-97864973 on Build GRCh38
rs935724810 Chr.10: 97864974-97864974 on Build GRCh38
rs937076582 Chr.10: 97865312-97865312 on Build GRCh38
rs1048527653 Chr.10: 97865394-97865394 on Build GRCh38
rs955547229 Chr.10: 97865112-97865112 on Build GRCh38
rs989067534 Chr.10: 97865147-97865147 on Build GRCh38
rs897151935 Chr.10: 97865040-97865040 on Build GRCh38
rs534146741 Chr.10: 97865350-97865350 on Build GRCh38
rs559077935 Chr.10: 97865373-97865373 on Build GRCh38
rs992648550 Chr.10: 97865315-97865315 on Build GRCh38
rs568763530 Chr.10: 97865333-97865333 on Build GRCh38
rs968380359 Chr.10: 97865240-97865240 on Build GRCh38
rs1037474505 Chr.10: 97865027-97865027 on Build GRCh38
rs376135474 Chr.10: 97865033-97865034 on Build GRCh38
rs879140852 Chr.10: 97865343-97865343 on Build GRCh38
rs537538189 Chr.10: 97865339-97865339 on Build GRCh38
rs905899121 Chr.10: 97865033-97865033 on Build GRCh38
rs927024310 Chr.10: 97865309-97865309 on Build GRCh38
rs755452510 Chr.10: 97865296-97865296 on Build GRCh38
rs1019775962 Chr.10: 97865083-97865083 on Build GRCh38
rs570687401 Chr.10: 97865385-97865385 on Build GRCh38
rs532321060 Chr.10: 97865055-97865055 on Build GRCh38
rs1026248293 Chr.10: 97865084-97865084 on Build GRCh38
rs1027036993 Chr.10: 97865067-97865067 on Build GRCh38
rs756133201 Chr.10: 97865048-97865048 on Build GRCh38
rs569315117 Chr.10: 97865245-97865245 on Build GRCh38
rs1007039504 Chr.10: 97865072-97865072 on Build GRCh38
rs11595577 Chr.10: 97865180-97865180 on Build GRCh38
rs571259411 Chr.10: 97865003-97865003 on Build GRCh38
rs913087145 Chr.10: 97865410-97865410 on Build GRCh38
rs561233090 Chr.10: 97864979-97864979 on Build GRCh38
rs889897106 Chr.10: 97865412-97865412 on Build GRCh38
rs750682715 Chr.10: 97865016-97865016 on Build GRCh38
rs964384873 Chr.10: 97865268-97865268 on Build GRCh38
rs116588961 Chr.10: 97865324-97865324 on Build GRCh38
rs532682664 Chr.10: 97865105-97865105 on Build GRCh38
rs940643647 Chr.10: 97865325-97865325 on Build GRCh38
rs142546456 Chr.10: 97865004-97865004 on Build GRCh38
rs1014611229 Chr.10: 97865026-97865026 on Build GRCh38
rs79132084 Chr.10: 97865071-97865071 on Build GRCh38

View Full Product Details