Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97865274-97865806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4017111 c.1952C>T p.S651L Chr.10: 97865582-97865582 on Build GRCh38
4017114 c.444C>T p.S148S Chr.10: 97865640-97865640 on Build GRCh38
1178537 c.1972G>A p.E658K Chr.10: 97865562-97865562 on Build GRCh38
4017112 c.394-8G>A p.? Chr.10: 97865582-97865582 on Build GRCh38
7324581 c.1881C>T p.A627A Chr.10: 97865653-97865653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368800333 Chr.10: 97865526-97865526 on Build GRCh38
rs768831216 Chr.10: 97865541-97865541 on Build GRCh38
rs1047629307 Chr.10: 97865750-97865750 on Build GRCh38
rs958485012 Chr.10: 97865534-97865534 on Build GRCh38
rs752404058 Chr.10: 97865591-97865591 on Build GRCh38
rs373874556 Chr.10: 97865687-97865687 on Build GRCh38
rs771051703 Chr.10: 97865749-97865749 on Build GRCh38
rs767364497 Chr.10: 97865674-97865674 on Build GRCh38
rs559077935 Chr.10: 97865373-97865373 on Build GRCh38
rs911228602 Chr.10: 97865782-97865782 on Build GRCh38
rs749346344 Chr.10: 97865758-97865758 on Build GRCh38
rs764952960 Chr.10: 97865638-97865639 on Build GRCh38
rs759330644 Chr.10: 97865671-97865671 on Build GRCh38
rs1010380728 Chr.10: 97865615-97865615 on Build GRCh38
rs747412494 Chr.10: 97865614-97865614 on Build GRCh38
rs138819524 Chr.10: 97865581-97865581 on Build GRCh38
rs958327725 Chr.10: 97865683-97865683 on Build GRCh38
rs1013968355 Chr.10: 97865692-97865692 on Build GRCh38
rs1018416660 Chr.10: 97865768-97865768 on Build GRCh38
rs751368422 Chr.10: 97865577-97865577 on Build GRCh38
rs111467483 Chr.10: 97865488-97865488 on Build GRCh38
rs116588961 Chr.10: 97865324-97865324 on Build GRCh38
rs774242787 Chr.10: 97865755-97865755 on Build GRCh38
rs753416888 Chr.10: 97865690-97865690 on Build GRCh38
rs767146682 Chr.10: 97865588-97865588 on Build GRCh38
rs376256449 Chr.10: 97865519-97865519 on Build GRCh38
rs567071631 Chr.10: 97865727-97865727 on Build GRCh38
rs755401919 Chr.10: 97865709-97865709 on Build GRCh38
rs770107830 Chr.10: 97865627-97865627 on Build GRCh38
rs937076582 Chr.10: 97865312-97865312 on Build GRCh38
rs552576574 Chr.10: 97865783-97865783 on Build GRCh38
rs747574968 Chr.10: 97865707-97865707 on Build GRCh38
rs56007204 Chr.10: 97865562-97865562 on Build GRCh38
rs992648550 Chr.10: 97865315-97865315 on Build GRCh38
rs777292975 Chr.10: 97865598-97865598 on Build GRCh38
rs568763530 Chr.10: 97865333-97865333 on Build GRCh38
rs775884488 Chr.10: 97865533-97865533 on Build GRCh38
rs371480295 Chr.10: 97865571-97865571 on Build GRCh38
rs752482571 Chr.10: 97865634-97865642 on Build GRCh38
rs149369679 Chr.10: 97865652-97865652 on Build GRCh38
rs574940451 Chr.10: 97865463-97865463 on Build GRCh38
rs746394085 Chr.10: 97865528-97865528 on Build GRCh38
rs759281769 Chr.10: 97865634-97865645 on Build GRCh38
rs761091655 Chr.10: 97865539-97865539 on Build GRCh38
rs905784361 Chr.10: 97865444-97865444 on Build GRCh38
rs1048452043 Chr.10: 97865582-97865582 on Build GRCh38
rs532338143 Chr.10: 97865545-97865545 on Build GRCh38
rs1041266564 Chr.10: 97865474-97865474 on Build GRCh38
rs775415171 Chr.10: 97865761-97865761 on Build GRCh38
rs777231847 Chr.10: 97865532-97865532 on Build GRCh38
rs766490550 Chr.10: 97865653-97865653 on Build GRCh38
rs1020507104 Chr.10: 97865515-97865515 on Build GRCh38
rs75406237 Chr.10: 97865499-97865499 on Build GRCh38
rs769079985 Chr.10: 97865616-97865616 on Build GRCh38
rs1048527653 Chr.10: 97865394-97865394 on Build GRCh38
rs898516803 Chr.10: 97865714-97865714 on Build GRCh38
rs770197602 Chr.10: 97865730-97865730 on Build GRCh38
rs775252392 Chr.10: 97865641-97865658 on Build GRCh38
rs548530776 Chr.10: 97865670-97865670 on Build GRCh38
rs565111213 Chr.10: 97865542-97865542 on Build GRCh38
rs537538189 Chr.10: 97865339-97865339 on Build GRCh38
rs570687401 Chr.10: 97865385-97865385 on Build GRCh38
rs530051798 Chr.10: 97865667-97865667 on Build GRCh38
rs972401835 Chr.10: 97865566-97865566 on Build GRCh38
rs375102264 Chr.10: 97865525-97865525 on Build GRCh38
rs913087145 Chr.10: 97865410-97865410 on Build GRCh38
rs777204918 Chr.10: 97865527-97865527 on Build GRCh38
rs763725933 Chr.10: 97865688-97865688 on Build GRCh38
rs763987216 Chr.10: 97865642-97865644 on Build GRCh38
rs748777328 Chr.10: 97865602-97865602 on Build GRCh38
rs375513503 Chr.10: 97865607-97865607 on Build GRCh38
rs780609461 Chr.10: 97865611-97865611 on Build GRCh38
rs200286666 Chr.10: 97865706-97865706 on Build GRCh38
rs940643647 Chr.10: 97865325-97865325 on Build GRCh38
rs114355950 Chr.10: 97865425-97865425 on Build GRCh38
rs1038271328 Chr.10: 97865704-97865704 on Build GRCh38
rs567115022 Chr.10: 97865345-97865345 on Build GRCh38
rs756768647 Chr.10: 97865696-97865696 on Build GRCh38
rs755711919 Chr.10: 97865596-97865596 on Build GRCh38
rs748435801 Chr.10: 97865625-97865625 on Build GRCh38
rs538377935 Chr.10: 97865411-97865411 on Build GRCh38
rs762942929 Chr.10: 97865641-97865642 on Build GRCh38
rs992536018 Chr.10: 97865543-97865543 on Build GRCh38
rs534146741 Chr.10: 97865350-97865350 on Build GRCh38
rs754634811 Chr.10: 97865523-97865523 on Build GRCh38
rs184784502 Chr.10: 97865734-97865734 on Build GRCh38
rs776864978 Chr.10: 97865619-97865633 on Build GRCh38
rs879140852 Chr.10: 97865343-97865343 on Build GRCh38
rs927024310 Chr.10: 97865309-97865309 on Build GRCh38
rs911217411 Chr.10: 97865586-97865586 on Build GRCh38
rs781652046 Chr.10: 97865725-97865725 on Build GRCh38
rs370716747 Chr.10: 97865524-97865524 on Build GRCh38
rs773387702 Chr.10: 97865563-97865563 on Build GRCh38
rs901352162 Chr.10: 97865608-97865608 on Build GRCh38
rs112433092 Chr.10: 97865518-97865518 on Build GRCh38
rs562926604 Chr.10: 97865584-97865584 on Build GRCh38
rs891213589 Chr.10: 97865754-97865754 on Build GRCh38
rs372046105 Chr.10: 97865756-97865756 on Build GRCh38
rs889897106 Chr.10: 97865412-97865412 on Build GRCh38
rs752391572 Chr.10: 97865676-97865676 on Build GRCh38
rs749454128 Chr.10: 97865735-97865735 on Build GRCh38
rs1042260529 Chr.10: 97865646-97865646 on Build GRCh38
rs776814365 Chr.10: 97865618-97865618 on Build GRCh38
rs773473661 Chr.10: 97865648-97865648 on Build GRCh38

View Full Product Details