Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97867089-97867592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

SNP Detail

SNP ID Chromosome Location
rs1030411625 Chr.10: 97867547-97867547 on Build GRCh38
rs760609545 Chr.10: 97867113-97867113 on Build GRCh38
rs933103901 Chr.10: 97867122-97867122 on Build GRCh38
rs922356906 Chr.10: 97867373-97867373 on Build GRCh38
rs895089282 Chr.10: 97867223-97867223 on Build GRCh38
rs966682467 Chr.10: 97867189-97867190 on Build GRCh38
rs964991041 Chr.10: 97867506-97867506 on Build GRCh38
rs3750606 Chr.10: 97867572-97867572 on Build GRCh38
rs936479816 Chr.10: 97867165-97867165 on Build GRCh38
rs959405321 Chr.10: 97867280-97867280 on Build GRCh38
rs769372523 Chr.10: 97867549-97867549 on Build GRCh38
rs1042294663 Chr.10: 97867491-97867491 on Build GRCh38
rs770972341 Chr.10: 97867205-97867205 on Build GRCh38
rs150194530 Chr.10: 97867472-97867472 on Build GRCh38
rs893721100 Chr.10: 97867141-97867141 on Build GRCh38
rs776751963 Chr.10: 97867206-97867206 on Build GRCh38
rs374285117 Chr.10: 97867518-97867518 on Build GRCh38
rs778193849 Chr.10: 97867270-97867270 on Build GRCh38
rs774108136 Chr.10: 97867573-97867573 on Build GRCh38
rs529462804 Chr.10: 97867170-97867170 on Build GRCh38
rs959457396 Chr.10: 97867562-97867570 on Build GRCh38
rs12252210 Chr.10: 97867565-97867565 on Build GRCh38
rs7096394 Chr.10: 97867488-97867488 on Build GRCh38
rs907163031 Chr.10: 97867356-97867356 on Build GRCh38
rs1050037997 Chr.10: 97867401-97867401 on Build GRCh38
rs977826673 Chr.10: 97867153-97867153 on Build GRCh38
rs562348428 Chr.10: 97867112-97867112 on Build GRCh38
rs566407293 Chr.10: 97867365-97867365 on Build GRCh38
rs932368196 Chr.10: 97867384-97867384 on Build GRCh38
rs902555730 Chr.10: 97867503-97867503 on Build GRCh38
rs556485742 Chr.10: 97867482-97867482 on Build GRCh38
rs182245593 Chr.10: 97867334-97867334 on Build GRCh38
rs570442493 Chr.10: 97867451-97867451 on Build GRCh38
rs923776440 Chr.10: 97867164-97867164 on Build GRCh38
rs141387994 Chr.10: 97867296-97867296 on Build GRCh38
rs1053648324 Chr.10: 97867179-97867179 on Build GRCh38
rs185189567 Chr.10: 97867411-97867411 on Build GRCh38
rs75917175 Chr.10: 97867366-97867366 on Build GRCh38
rs1044831456 Chr.10: 97867347-97867347 on Build GRCh38
rs954367829 Chr.10: 97867548-97867548 on Build GRCh38
rs758621467 Chr.10: 97867129-97867129 on Build GRCh38
rs561397440 Chr.10: 97867328-97867328 on Build GRCh38
rs996427383 Chr.10: 97867532-97867532 on Build GRCh38
rs982221993 Chr.10: 97867266-97867266 on Build GRCh38
rs1028116123 Chr.10: 97867127-97867128 on Build GRCh38

View Full Product Details