Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97867485-97867995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
UniGene
Hs.500736
Gene Chromosome Location
Chr.10: 97854711-98030828 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

Gene Symbol
GOLGA7B
Entrez Gene ID
401647
Gene Name
golgin A7 family member B
Gene Aliases
C10orf132, C10orf133, bA451M19.3, bA459F3.4
UniGene
Hs.567524
Gene Chromosome Location
Chr.10: 97850238-97871578 on Build GRCh38
Transcripts
NM_018058.6 XM_011539917.1 XM_017016367.1 NM_001010917.2

SNP Detail

SNP ID Chromosome Location
rs1037911521 Chr.10: 97867758-97867758 on Build GRCh38
rs576020133 Chr.10: 97867714-97867714 on Build GRCh38
rs868745285 Chr.10: 97867910-97867910 on Build GRCh38
rs1012776076 Chr.10: 97867955-97867955 on Build GRCh38
rs769372523 Chr.10: 97867549-97867549 on Build GRCh38
rs762512914 Chr.10: 97867618-97867618 on Build GRCh38
rs560100391 Chr.10: 97867773-97867773 on Build GRCh38
rs916424628 Chr.10: 97867834-97867834 on Build GRCh38
rs758382833 Chr.10: 97867575-97867576 on Build GRCh38
rs927928321 Chr.10: 97867878-97867878 on Build GRCh38
rs954544423 Chr.10: 97867780-97867780 on Build GRCh38
rs374285117 Chr.10: 97867518-97867518 on Build GRCh38
rs796952761 Chr.10: 97867702-97867702 on Build GRCh38
rs774108136 Chr.10: 97867573-97867573 on Build GRCh38
rs776859645 Chr.10: 97867795-97867795 on Build GRCh38
rs543752962 Chr.10: 97867744-97867744 on Build GRCh38
rs1006304377 Chr.10: 97867681-97867681 on Build GRCh38
rs12252210 Chr.10: 97867565-97867565 on Build GRCh38
rs765361515 Chr.10: 97867839-97867839 on Build GRCh38
rs989301285 Chr.10: 97867578-97867578 on Build GRCh38
rs908129048 Chr.10: 97867598-97867598 on Build GRCh38
rs532389847 Chr.10: 97867877-97867877 on Build GRCh38
rs1027733848 Chr.10: 97867970-97867970 on Build GRCh38
rs545813330 Chr.10: 97867661-97867661 on Build GRCh38
rs932420562 Chr.10: 97867650-97867650 on Build GRCh38
rs552236524 Chr.10: 97867953-97867953 on Build GRCh38
rs996427383 Chr.10: 97867532-97867532 on Build GRCh38
rs1030411625 Chr.10: 97867547-97867547 on Build GRCh38
rs915087343 Chr.10: 97867721-97867721 on Build GRCh38
rs149385875 Chr.10: 97867675-97867675 on Build GRCh38
rs1031070366 Chr.10: 97867796-97867796 on Build GRCh38
rs190156600 Chr.10: 97867772-97867772 on Build GRCh38
rs3750606 Chr.10: 97867572-97867572 on Build GRCh38
rs1048177292 Chr.10: 97867647-97867647 on Build GRCh38
rs928640643 Chr.10: 97867621-97867621 on Build GRCh38
rs947976329 Chr.10: 97867607-97867607 on Build GRCh38
rs138685506 Chr.10: 97867592-97867592 on Build GRCh38
rs989152192 Chr.10: 97867832-97867832 on Build GRCh38
rs529502273 Chr.10: 97867777-97867777 on Build GRCh38
rs969166279 Chr.10: 97867855-97867855 on Build GRCh38
rs375813796 Chr.10: 97867778-97867778 on Build GRCh38
rs959457396 Chr.10: 97867562-97867570 on Build GRCh38
rs572415315 Chr.10: 97867629-97867629 on Build GRCh38
rs1056498335 Chr.10: 97867935-97867935 on Build GRCh38
rs929871288 Chr.10: 97867895-97867895 on Build GRCh38
rs759475855 Chr.10: 97867808-97867808 on Build GRCh38
rs976393792 Chr.10: 97867630-97867630 on Build GRCh38
rs74153107 Chr.10: 97867716-97867716 on Build GRCh38
rs1046276957 Chr.10: 97867613-97867613 on Build GRCh38
rs954367829 Chr.10: 97867548-97867548 on Build GRCh38

View Full Product Details