Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101957459-101957958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
-
UniGene
Hs.16004
Gene Chromosome Location
Chr.10: 101845599-102056237 on Build GRCh38
Transcripts
NM_024541.2 XM_006717969.1 XM_011540151.2 XM_011540152.2 XM_011540153.2 XM_011540154.2 XM_011540156.2 XM_011540157.2 XM_017016635.1 XM_017016636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731615 c.1496-4A>T p.? Chr.10: 101957736-101957736 on Build GRCh38
4011169 c.1558A>T p.I520F Chr.10: 101957670-101957670 on Build GRCh38
7266073 c.1578+5G>A p.? Chr.10: 101957645-101957645 on Build GRCh38
6398071 c.1509G>T p.L503F Chr.10: 101957719-101957719 on Build GRCh38
6237686 c.1578+9C>A p.? Chr.10: 101957641-101957641 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs80213387 Chr.10: 101957916-101957916 on Build GRCh38
rs144960435 Chr.10: 101957483-101957483 on Build GRCh38
rs560646767 Chr.10: 101957820-101957820 on Build GRCh38
rs748600538 Chr.10: 101957724-101957724 on Build GRCh38
rs376923161 Chr.10: 101957751-101957752 on Build GRCh38
rs775461649 Chr.10: 101957600-101957600 on Build GRCh38
rs775124668 Chr.10: 101957621-101957621 on Build GRCh38
rs769652189 Chr.10: 101957616-101957616 on Build GRCh38
rs888428578 Chr.10: 101957572-101957572 on Build GRCh38
rs373770074 Chr.10: 101957932-101957932 on Build GRCh38
rs780511090 Chr.10: 101957695-101957695 on Build GRCh38
rs370503995 Chr.10: 101957736-101957737 on Build GRCh38
rs916974947 Chr.10: 101957508-101957508 on Build GRCh38
rs905502693 Chr.10: 101957869-101957869 on Build GRCh38
rs745802122 Chr.10: 101957748-101957749 on Build GRCh38
rs901235610 Chr.10: 101957643-101957643 on Build GRCh38
rs998679629 Chr.10: 101957670-101957670 on Build GRCh38
rs972967835 Chr.10: 101957501-101957501 on Build GRCh38
rs750684702 Chr.10: 101957682-101957682 on Build GRCh38
rs1032954290 Chr.10: 101957799-101957799 on Build GRCh38
rs770508494 Chr.10: 101957608-101957608 on Build GRCh38
rs762916173 Chr.10: 101957647-101957647 on Build GRCh38
rs768374023 Chr.10: 101957825-101957825 on Build GRCh38
rs191657538 Chr.10: 101957836-101957836 on Build GRCh38
rs949857048 Chr.10: 101957509-101957509 on Build GRCh38
rs982260302 Chr.10: 101957516-101957516 on Build GRCh38
rs754150261 Chr.10: 101957698-101957698 on Build GRCh38
rs186711754 Chr.10: 101957633-101957633 on Build GRCh38
rs769299131 Chr.10: 101957773-101957773 on Build GRCh38
rs555133769 Chr.10: 101957833-101957834 on Build GRCh38
rs917172561 Chr.10: 101957811-101957811 on Build GRCh38
rs868802350 Chr.10: 101957733-101957733 on Build GRCh38
rs979486590 Chr.10: 101957693-101957693 on Build GRCh38
rs930235077 Chr.10: 101957520-101957520 on Build GRCh38
rs796393862 Chr.10: 101957737-101957738 on Build GRCh38
rs762734001 Chr.10: 101957662-101957662 on Build GRCh38
rs942731652 Chr.10: 101957591-101957591 on Build GRCh38
rs745582121 Chr.10: 101957760-101957760 on Build GRCh38
rs557242117 Chr.10: 101957673-101957673 on Build GRCh38
rs371311945 Chr.10: 101957654-101957654 on Build GRCh38
rs375807542 Chr.10: 101957593-101957593 on Build GRCh38
rs918756770 Chr.10: 101957517-101957517 on Build GRCh38
rs925787265 Chr.10: 101957723-101957723 on Build GRCh38
rs375033413 Chr.10: 101957755-101957755 on Build GRCh38
rs758478944 Chr.10: 101957737-101957737 on Build GRCh38
rs1053025438 Chr.10: 101957787-101957787 on Build GRCh38
rs10128116 Chr.10: 101957856-101957856 on Build GRCh38
rs911327256 Chr.10: 101957568-101957570 on Build GRCh38
rs971135182 Chr.10: 101957908-101957908 on Build GRCh38
rs552929669 Chr.10: 101957610-101957610 on Build GRCh38
rs559727443 Chr.10: 101957878-101957878 on Build GRCh38
rs775419158 Chr.10: 101957782-101957782 on Build GRCh38
rs761890481 Chr.10: 101957671-101957671 on Build GRCh38
rs542220026 Chr.10: 101957736-101957736 on Build GRCh38
rs181690866 Chr.10: 101957613-101957613 on Build GRCh38
rs769594396 Chr.10: 101957485-101957485 on Build GRCh38
rs759070649 Chr.10: 101957615-101957615 on Build GRCh38
rs534805673 Chr.10: 101957556-101957556 on Build GRCh38
rs369020578 Chr.10: 101957594-101957594 on Build GRCh38
rs575095402 Chr.10: 101957721-101957721 on Build GRCh38
rs1020083125 Chr.10: 101957765-101957765 on Build GRCh38
rs567763042 Chr.10: 101957502-101957502 on Build GRCh38
rs1039693542 Chr.10: 101957620-101957620 on Build GRCh38
rs1029832414 Chr.10: 101957527-101957527 on Build GRCh38
rs386372278 Chr.10: 101957737-101957738 on Build GRCh38
rs1024391806 Chr.10: 101957889-101957889 on Build GRCh38
rs955600186 Chr.10: 101957548-101957548 on Build GRCh38
rs756270081 Chr.10: 101957691-101957691 on Build GRCh38
rs1048666531 Chr.10: 101957553-101957553 on Build GRCh38
rs765215747 Chr.10: 101957735-101957736 on Build GRCh38

View Full Product Details