Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117543042-117543581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMX2
Entrez Gene ID
2018
Gene Name
empty spiracles homeobox 2
Gene Aliases
-
UniGene
Hs.202095
Gene Chromosome Location
Chr.10: 117542445-117549546 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Gene Symbol
EMX2OS
Entrez Gene ID
196047
Gene Name
EMX2 opposite strand/antisense RNA
Gene Aliases
EMX2-AS1, NCRNA00045
UniGene
Hs.312592
Gene Chromosome Location
Chr.10: 117484293-117545068 on Build GRCh38
Transcripts
NM_001165924.1 NM_004098.3 NR_002791.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653933 c.243C>T p.P81P Chr.10: 117543510-117543510 on Build GRCh38
2055180 c.252A>G p.P84P Chr.10: 117543519-117543519 on Build GRCh38
4760622 c.143T>G p.L48R Chr.10: 117543410-117543410 on Build GRCh38
2055184 c.297A>G p.L99L Chr.10: 117543564-117543564 on Build GRCh38
2055185 c.309C>T p.H103H Chr.10: 117543576-117543576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8192643 Chr.10: 117543222-117543222 on Build GRCh38
rs113852997 Chr.10: 117543523-117543523 on Build GRCh38
rs747646645 Chr.10: 117543380-117543380 on Build GRCh38
rs750776907 Chr.10: 117543288-117543288 on Build GRCh38
rs761060914 Chr.10: 117543303-117543303 on Build GRCh38
rs377504325 Chr.10: 117543405-117543405 on Build GRCh38
rs762592184 Chr.10: 117543227-117543227 on Build GRCh38
rs768257816 Chr.10: 117543240-117543240 on Build GRCh38
rs749036031 Chr.10: 117543462-117543462 on Build GRCh38
rs758460881 Chr.10: 117543424-117543424 on Build GRCh38
rs751794084 Chr.10: 117543519-117543519 on Build GRCh38
rs372863199 Chr.10: 117543089-117543089 on Build GRCh38
rs755111431 Chr.10: 117543318-117543318 on Build GRCh38
rs865888874 Chr.10: 117543507-117543507 on Build GRCh38
rs757916173 Chr.10: 117543556-117543556 on Build GRCh38
rs921189469 Chr.10: 117543248-117543248 on Build GRCh38
rs766915981 Chr.10: 117543304-117543304 on Build GRCh38
rs780189136 Chr.10: 117543359-117543359 on Build GRCh38
rs762141298 Chr.10: 117543521-117543521 on Build GRCh38
rs1040675762 Chr.10: 117543472-117543472 on Build GRCh38
rs146160041 Chr.10: 117543360-117543360 on Build GRCh38
rs373030604 Chr.10: 117543372-117543372 on Build GRCh38
rs984575163 Chr.10: 117543241-117543241 on Build GRCh38
rs778870509 Chr.10: 117543468-117543468 on Build GRCh38
rs994408939 Chr.10: 117543177-117543177 on Build GRCh38
rs759842203 Chr.10: 117543509-117543509 on Build GRCh38
rs760998084 Chr.10: 117543384-117543384 on Build GRCh38
rs1006487322 Chr.10: 117543469-117543469 on Build GRCh38
rs951355872 Chr.10: 117543490-117543490 on Build GRCh38
rs1046119243 Chr.10: 117543361-117543361 on Build GRCh38
rs777999453 Chr.10: 117543342-117543342 on Build GRCh38
rs746663938 Chr.10: 117543501-117543501 on Build GRCh38
rs980174064 Chr.10: 117543137-117543137 on Build GRCh38
rs12784456 Chr.10: 117543103-117543103 on Build GRCh38
rs912653707 Chr.10: 117543066-117543066 on Build GRCh38
rs559537205 Chr.10: 117543142-117543142 on Build GRCh38
rs770506828 Chr.10: 117543505-117543505 on Build GRCh38
rs751783182 Chr.10: 117543437-117543437 on Build GRCh38
rs775528347 Chr.10: 117543225-117543225 on Build GRCh38
rs200918900 Chr.10: 117543526-117543526 on Build GRCh38
rs369213869 Chr.10: 117543181-117543181 on Build GRCh38
rs754788682 Chr.10: 117543527-117543527 on Build GRCh38
rs866810600 Chr.10: 117543356-117543356 on Build GRCh38
rs776990556 Chr.10: 117543508-117543508 on Build GRCh38
rs772894553 Chr.10: 117543381-117543381 on Build GRCh38
rs758560390 Chr.10: 117543341-117543341 on Build GRCh38
rs527345548 Chr.10: 117543250-117543250 on Build GRCh38
rs777192509 Chr.10: 117543414-117543414 on Build GRCh38
rs752738842 Chr.10: 117543423-117543423 on Build GRCh38
rs764263360 Chr.10: 117543431-117543431 on Build GRCh38
rs761378350 Chr.10: 117543260-117543260 on Build GRCh38
rs765299136 Chr.10: 117543325-117543325 on Build GRCh38
rs765761502 Chr.10: 117543511-117543511 on Build GRCh38
rs1019794614 Chr.10: 117543127-117543127 on Build GRCh38
rs371177025 Chr.10: 117543513-117543513 on Build GRCh38
rs200357168 Chr.10: 117543539-117543539 on Build GRCh38
rs747651960 Chr.10: 117543481-117543481 on Build GRCh38
rs958937476 Chr.10: 117543395-117543395 on Build GRCh38
rs777343626 Chr.10: 117543559-117543559 on Build GRCh38
rs771593269 Chr.10: 117543483-117543483 on Build GRCh38
rs866930422 Chr.10: 117543343-117543343 on Build GRCh38
rs529727475 Chr.10: 117543394-117543394 on Build GRCh38
rs926923674 Chr.10: 117543143-117543143 on Build GRCh38
rs762991629 Chr.10: 117543514-117543514 on Build GRCh38
rs149834474 Chr.10: 117543558-117543558 on Build GRCh38
rs765671833 Chr.10: 117543416-117543416 on Build GRCh38
rs760038502 Chr.10: 117543415-117543415 on Build GRCh38
rs149284992 Chr.10: 117543132-117543137 on Build GRCh38
rs549784838 Chr.10: 117543440-117543440 on Build GRCh38
rs745614991 Chr.10: 117543221-117543221 on Build GRCh38
rs140164453 Chr.10: 117543388-117543388 on Build GRCh38
rs779988495 Chr.10: 117543451-117543451 on Build GRCh38
rs906303624 Chr.10: 117543364-117543364 on Build GRCh38
rs755940979 Chr.10: 117543348-117543348 on Build GRCh38
rs752669966 Chr.10: 117543549-117543549 on Build GRCh38
rs540731987 Chr.10: 117543281-117543281 on Build GRCh38
rs965583845 Chr.10: 117543174-117543174 on Build GRCh38
rs764220035 Chr.10: 117543517-117543517 on Build GRCh38
rs886044442 Chr.10: 117543445-117543446 on Build GRCh38
rs752753584 Chr.10: 117543327-117543327 on Build GRCh38
rs369744669 Chr.10: 117543337-117543337 on Build GRCh38
rs766989996 Chr.10: 117543426-117543427 on Build GRCh38
rs745677645 Chr.10: 117543344-117543344 on Build GRCh38
rs768649851 Chr.10: 117543370-117543370 on Build GRCh38
rs962443860 Chr.10: 117543506-117543506 on Build GRCh38
rs756693906 Chr.10: 117543427-117543429 on Build GRCh38
rs1006564728 Chr.10: 117543067-117543067 on Build GRCh38
rs794726911 Chr.10: 117543440-117543445 on Build GRCh38
rs772801721 Chr.10: 117543494-117543494 on Build GRCh38
rs770590374 Chr.10: 117543563-117543563 on Build GRCh38
rs754989802 Chr.10: 117543464-117543464 on Build GRCh38
rs577004581 Chr.10: 117543082-117543082 on Build GRCh38
rs746455640 Chr.10: 117543561-117543561 on Build GRCh38
rs375691102 Chr.10: 117543309-117543309 on Build GRCh38
rs983219932 Chr.10: 117543496-117543496 on Build GRCh38
rs371549541 Chr.10: 117543280-117543280 on Build GRCh38
rs1039966586 Chr.10: 117543238-117543238 on Build GRCh38

View Full Product Details