Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124007574-124008071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST15
Entrez Gene ID
51363
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
BRAG, GALNAC4S-6ST
UniGene
Hs.287537
Gene Chromosome Location
Chr.10: 124006060-124093684 on Build GRCh38
Transcripts
NM_001270764.1 NM_015892.4 XM_005269891.3 XM_005269894.4 XM_006717891.3 XM_017016319.1

SNP Detail

SNP ID Chromosome Location
rs886146760 Chr.10: 124007668-124007668 on Build GRCh38
rs1039882535 Chr.10: 124007849-124007849 on Build GRCh38
rs76828416 Chr.10: 124007793-124007793 on Build GRCh38
rs954977669 Chr.10: 124007712-124007712 on Build GRCh38
rs979295847 Chr.10: 124007763-124007763 on Build GRCh38
rs118052074 Chr.10: 124007960-124007960 on Build GRCh38
rs904557867 Chr.10: 124007923-124007923 on Build GRCh38
rs544997926 Chr.10: 124007988-124007989 on Build GRCh38
rs76660677 Chr.10: 124007702-124007702 on Build GRCh38
rs975119328 Chr.10: 124008045-124008045 on Build GRCh38
rs961017486 Chr.10: 124007780-124007780 on Build GRCh38
rs779684553 Chr.10: 124007767-124007767 on Build GRCh38
rs569631937 Chr.10: 124007939-124007939 on Build GRCh38
rs1024151800 Chr.10: 124007970-124007970 on Build GRCh38
rs28521273 Chr.10: 124008006-124008006 on Build GRCh38
rs1000190664 Chr.10: 124007929-124007929 on Build GRCh38
rs932284513 Chr.10: 124007932-124007932 on Build GRCh38
rs532587405 Chr.10: 124007914-124007914 on Build GRCh38
rs982660258 Chr.10: 124007843-124007843 on Build GRCh38
rs60643581 Chr.10: 124007988-124007988 on Build GRCh38
rs879652199 Chr.10: 124007917-124007917 on Build GRCh38
rs1020250693 Chr.10: 124007742-124007742 on Build GRCh38
rs528516917 Chr.10: 124007833-124007833 on Build GRCh38
rs1031025291 Chr.10: 124007757-124007757 on Build GRCh38
rs996341087 Chr.10: 124007691-124007691 on Build GRCh38
rs200021931 Chr.10: 124007985-124007985 on Build GRCh38
rs368805189 Chr.10: 124007983-124007983 on Build GRCh38
rs1010479569 Chr.10: 124007915-124007915 on Build GRCh38
rs909581062 Chr.10: 124007846-124007846 on Build GRCh38
rs771330739 Chr.10: 124007919-124007919 on Build GRCh38
rs941032713 Chr.10: 124007848-124007848 on Build GRCh38
rs150485426 Chr.10: 124007936-124007936 on Build GRCh38
rs767533619 Chr.10: 124007701-124007701 on Build GRCh38
rs1014009881 Chr.10: 124008011-124008011 on Build GRCh38
rs563624854 Chr.10: 124007713-124007713 on Build GRCh38
rs929847652 Chr.10: 124007635-124007635 on Build GRCh38
rs41307068 Chr.10: 124007615-124007615 on Build GRCh38
rs561230422 Chr.10: 124007817-124007817 on Build GRCh38
rs200939986 Chr.10: 124007814-124007814 on Build GRCh38
rs577720955 Chr.10: 124007690-124007690 on Build GRCh38
rs904021681 Chr.10: 124007751-124007751 on Build GRCh38
rs28425468 Chr.10: 124007688-124007688 on Build GRCh38
rs941051943 Chr.10: 124008026-124008026 on Build GRCh38
rs868861186 Chr.10: 124007617-124007617 on Build GRCh38
rs762232086 Chr.10: 124007825-124007825 on Build GRCh38
rs556873495 Chr.10: 124007648-124007648 on Build GRCh38
rs769696125 Chr.10: 124008002-124008002 on Build GRCh38
rs565583734 Chr.10: 124007906-124007906 on Build GRCh38
rs754676725 Chr.10: 124007809-124007809 on Build GRCh38
rs28435845 Chr.10: 124007856-124007856 on Build GRCh38
rs71704905 Chr.10: 124007984-124007985 on Build GRCh38
rs751423875 Chr.10: 124007661-124007661 on Build GRCh38
rs538188851 Chr.10: 124007643-124007643 on Build GRCh38
rs755568421 Chr.10: 124007754-124007754 on Build GRCh38
rs778931735 Chr.10: 124007985-124007986 on Build GRCh38
rs747340081 Chr.10: 124007868-124007868 on Build GRCh38
rs79057288 Chr.10: 124008013-124008013 on Build GRCh38
rs577288718 Chr.10: 124007889-124007889 on Build GRCh38
rs543027461 Chr.10: 124007810-124007810 on Build GRCh38
rs781654098 Chr.10: 124007940-124007940 on Build GRCh38
rs1024663127 Chr.10: 124007795-124007801 on Build GRCh38
rs1046780270 Chr.10: 124007642-124007642 on Build GRCh38
rs999603253 Chr.10: 124007755-124007755 on Build GRCh38
rs930461153 Chr.10: 124007842-124007842 on Build GRCh38
rs573585662 Chr.10: 124007604-124007604 on Build GRCh38
rs200947102 Chr.10: 124007988-124007988 on Build GRCh38
rs764350461 Chr.10: 124007905-124007905 on Build GRCh38
rs183360613 Chr.10: 124007962-124007962 on Build GRCh38
rs746452998 Chr.10: 124007971-124007971 on Build GRCh38

View Full Product Details