Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124019458-124019948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST15
Entrez Gene ID
51363
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
BRAG, GALNAC4S-6ST
UniGene
Hs.287537
Gene Chromosome Location
Chr.10: 124006060-124093684 on Build GRCh38
Transcripts
NM_001270764.1 NM_001270765.1 NM_014863.3 NM_015892.4 XM_005269891.3 XM_005269892.4 XM_005269893.3 XM_005269894.4 XM_006717891.3 XM_017016319.1 XM_017016320.1 XM_017016321.1

SNP Detail

SNP ID Chromosome Location
rs575016770 Chr.10: 124019705-124019705 on Build GRCh38
rs761152031 Chr.10: 124019519-124019519 on Build GRCh38
rs76460523 Chr.10: 124019801-124019801 on Build GRCh38
rs144198836 Chr.10: 124019655-124019655 on Build GRCh38
rs1012892885 Chr.10: 124019777-124019777 on Build GRCh38
rs1054006129 Chr.10: 124019857-124019857 on Build GRCh38
rs1002583907 Chr.10: 124019702-124019702 on Build GRCh38
rs182956538 Chr.10: 124019662-124019662 on Build GRCh38
rs981382931 Chr.10: 124019485-124019485 on Build GRCh38
rs745873753 Chr.10: 124019888-124019888 on Build GRCh38
rs114722629 Chr.10: 124019602-124019602 on Build GRCh38
rs1055130914 Chr.10: 124019924-124019924 on Build GRCh38
rs1016061481 Chr.10: 124019734-124019734 on Build GRCh38
rs903470922 Chr.10: 124019737-124019737 on Build GRCh38
rs961298353 Chr.10: 124019827-124019827 on Build GRCh38
rs971100084 Chr.10: 124019478-124019478 on Build GRCh38
rs984667780 Chr.10: 124019732-124019732 on Build GRCh38
rs778091666 Chr.10: 124019749-124019749 on Build GRCh38
rs958656014 Chr.10: 124019713-124019713 on Build GRCh38
rs773488251 Chr.10: 124019489-124019489 on Build GRCh38
rs1033419256 Chr.10: 124019544-124019544 on Build GRCh38
rs781041121 Chr.10: 124019832-124019832 on Build GRCh38
rs76787698 Chr.10: 124019637-124019637 on Build GRCh38
rs577042279 Chr.10: 124019594-124019594 on Build GRCh38
rs573498018 Chr.10: 124019505-124019505 on Build GRCh38
rs147968573 Chr.10: 124019817-124019817 on Build GRCh38
rs948691826 Chr.10: 124019598-124019599 on Build GRCh38
rs985817086 Chr.10: 124019831-124019831 on Build GRCh38
rs889273492 Chr.10: 124019503-124019503 on Build GRCh38
rs999146711 Chr.10: 124019901-124019901 on Build GRCh38
rs892635367 Chr.10: 124019877-124019877 on Build GRCh38
rs368989788 Chr.10: 124019517-124019517 on Build GRCh38
rs565088789 Chr.10: 124019579-124019579 on Build GRCh38
rs903333901 Chr.10: 124019895-124019895 on Build GRCh38
rs765297607 Chr.10: 124019627-124019627 on Build GRCh38
rs957589553 Chr.10: 124019771-124019771 on Build GRCh38
rs936941165 Chr.10: 124019836-124019836 on Build GRCh38
rs919824571 Chr.10: 124019847-124019847 on Build GRCh38
rs1038076915 Chr.10: 124019626-124019626 on Build GRCh38
rs943761423 Chr.10: 124019887-124019887 on Build GRCh38
rs766943049 Chr.10: 124019586-124019586 on Build GRCh38
rs991965703 Chr.10: 124019573-124019573 on Build GRCh38
rs906635707 Chr.10: 124019880-124019880 on Build GRCh38
rs932326461 Chr.10: 124019872-124019872 on Build GRCh38
rs551032131 Chr.10: 124019891-124019891 on Build GRCh38
rs939930754 Chr.10: 124019609-124019609 on Build GRCh38
rs1033038371 Chr.10: 124019750-124019750 on Build GRCh38
rs11551352 Chr.10: 124019822-124019822 on Build GRCh38
rs532383416 Chr.10: 124019878-124019878 on Build GRCh38
rs190381216 Chr.10: 124019493-124019493 on Build GRCh38
rs373506484 Chr.10: 124019532-124019532 on Build GRCh38
rs544667781 Chr.10: 124019597-124019597 on Build GRCh38
rs754406135 Chr.10: 124019595-124019595 on Build GRCh38
rs912321655 Chr.10: 124019879-124019879 on Build GRCh38
rs1040530500 Chr.10: 124019736-124019736 on Build GRCh38
rs971417829 Chr.10: 124019815-124019815 on Build GRCh38
rs906551650 Chr.10: 124019675-124019675 on Build GRCh38
rs569299943 Chr.10: 124019896-124019896 on Build GRCh38
rs981250884 Chr.10: 124019816-124019816 on Build GRCh38
rs867541517 Chr.10: 124019640-124019640 on Build GRCh38
rs957518058 Chr.10: 124019559-124019559 on Build GRCh38

View Full Product Details