Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3667121-3667618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA10
Entrez Gene ID
57053
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
-
UniGene
Hs.732806
Gene Chromosome Location
Chr.11: 3665587-3673208 on Build GRCh38
Transcripts
NM_001303034.1 NM_001303035.1 NM_020402.3 XM_011520234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6431518 c.685A>G p.T229A Chr.11: 3667442-3667442 on Build GRCh38
926772 c.718G>A p.V240M Chr.11: 3667409-3667409 on Build GRCh38
6391760 c.809T>A p.L270Q Chr.11: 3667318-3667318 on Build GRCh38
3687282 c.857C>T p.A286V Chr.11: 3667270-3667270 on Build GRCh38
6412449 c.575G>A p.S192N Chr.11: 3667552-3667552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769469175 Chr.11: 3667201-3667201 on Build GRCh38
rs781223689 Chr.11: 3667332-3667332 on Build GRCh38
rs776814904 Chr.11: 3667482-3667482 on Build GRCh38
rs760737901 Chr.11: 3667416-3667416 on Build GRCh38
rs755960899 Chr.11: 3667346-3667346 on Build GRCh38
rs777354719 Chr.11: 3667426-3667426 on Build GRCh38
rs777343278 Chr.11: 3667394-3667394 on Build GRCh38
rs753813237 Chr.11: 3667388-3667388 on Build GRCh38
rs773474519 Chr.11: 3667415-3667415 on Build GRCh38
rs773659023 Chr.11: 3667182-3667183 on Build GRCh38
rs754915280 Chr.11: 3667420-3667420 on Build GRCh38
rs370718990 Chr.11: 3667308-3667308 on Build GRCh38
rs962478837 Chr.11: 3667524-3667524 on Build GRCh38
rs370949315 Chr.11: 3667418-3667418 on Build GRCh38
rs761146449 Chr.11: 3667244-3667245 on Build GRCh38
rs200192816 Chr.11: 3667485-3667485 on Build GRCh38
rs934086349 Chr.11: 3667189-3667189 on Build GRCh38
rs140742217 Chr.11: 3667331-3667331 on Build GRCh38
rs201163422 Chr.11: 3667479-3667479 on Build GRCh38
rs762896708 Chr.11: 3667536-3667536 on Build GRCh38
rs546755450 Chr.11: 3667596-3667596 on Build GRCh38
rs1049951697 Chr.11: 3667245-3667245 on Build GRCh38
rs756764123 Chr.11: 3667429-3667429 on Build GRCh38
rs758085196 Chr.11: 3667392-3667392 on Build GRCh38
rs1032692473 Chr.11: 3667582-3667582 on Build GRCh38
rs758459180 Chr.11: 3667486-3667512 on Build GRCh38
rs182159148 Chr.11: 3667573-3667573 on Build GRCh38
rs769677015 Chr.11: 3667268-3667268 on Build GRCh38
rs768352154 Chr.11: 3667274-3667274 on Build GRCh38
rs771119230 Chr.11: 3667518-3667518 on Build GRCh38
rs545699308 Chr.11: 3667178-3667178 on Build GRCh38
rs929777698 Chr.11: 3667565-3667565 on Build GRCh38
rs778006719 Chr.11: 3667566-3667566 on Build GRCh38
rs751774593 Chr.11: 3667321-3667321 on Build GRCh38
rs771706248 Chr.11: 3667284-3667284 on Build GRCh38
rs746641185 Chr.11: 3667427-3667427 on Build GRCh38
rs917849206 Chr.11: 3667152-3667152 on Build GRCh38
rs748982511 Chr.11: 3667353-3667353 on Build GRCh38
rs528669032 Chr.11: 3667589-3667589 on Build GRCh38
rs772904315 Chr.11: 3667592-3667592 on Build GRCh38
rs760932993 Chr.11: 3667467-3667467 on Build GRCh38
rs568176743 Chr.11: 3667421-3667421 on Build GRCh38
rs759872210 Chr.11: 3667422-3667424 on Build GRCh38
rs978017903 Chr.11: 3667410-3667410 on Build GRCh38
rs535473918 Chr.11: 3667424-3667424 on Build GRCh38
rs2231541 Chr.11: 3667569-3667569 on Build GRCh38
rs755440002 Chr.11: 3667236-3667236 on Build GRCh38
rs762488072 Chr.11: 3667209-3667209 on Build GRCh38
rs756891520 Chr.11: 3667501-3667501 on Build GRCh38
rs745759747 Chr.11: 3667266-3667266 on Build GRCh38
rs140976405 Chr.11: 3667366-3667366 on Build GRCh38
rs774014913 Chr.11: 3667282-3667282 on Build GRCh38
rs763825468 Chr.11: 3667538-3667538 on Build GRCh38
rs756087818 Chr.11: 3667386-3667386 on Build GRCh38
rs779521433 Chr.11: 3667547-3667547 on Build GRCh38
rs747827541 Chr.11: 3667391-3667391 on Build GRCh38
rs2231544 Chr.11: 3667214-3667214 on Build GRCh38
rs576316543 Chr.11: 3667431-3667431 on Build GRCh38
rs776973836 Chr.11: 3667309-3667328 on Build GRCh38
rs925139829 Chr.11: 3667222-3667222 on Build GRCh38
rs753015038 Chr.11: 3667243-3667243 on Build GRCh38
rs764357729 Chr.11: 3667320-3667320 on Build GRCh38
rs761272694 Chr.11: 3667217-3667217 on Build GRCh38
rs888647871 Chr.11: 3667364-3667364 on Build GRCh38
rs761971862 Chr.11: 3667382-3667382 on Build GRCh38
rs369074831 Chr.11: 3667441-3667441 on Build GRCh38
rs754864013 Chr.11: 3667389-3667389 on Build GRCh38
rs758227414 Chr.11: 3667570-3667570 on Build GRCh38
rs955026093 Chr.11: 3667432-3667432 on Build GRCh38
rs763924776 Chr.11: 3667496-3667496 on Build GRCh38
rs373527871 Chr.11: 3667406-3667406 on Build GRCh38
rs765438697 Chr.11: 3667443-3667445 on Build GRCh38
rs775899850 Chr.11: 3667401-3667401 on Build GRCh38
rs529715254 Chr.11: 3667218-3667218 on Build GRCh38
rs765657038 Chr.11: 3667562-3667562 on Build GRCh38
rs748626494 Chr.11: 3667510-3667510 on Build GRCh38
rs754387595 Chr.11: 3667233-3667233 on Build GRCh38
rs148870170 Chr.11: 3667422-3667422 on Build GRCh38
rs777237107 Chr.11: 3667290-3667290 on Build GRCh38
rs112052659 Chr.11: 3667204-3667204 on Build GRCh38
rs2231542 Chr.11: 3667385-3667385 on Build GRCh38
rs138838925 Chr.11: 3667387-3667387 on Build GRCh38
rs376404549 Chr.11: 3667412-3667412 on Build GRCh38
rs758501459 Chr.11: 3667250-3667250 on Build GRCh38
rs761731304 Chr.11: 3667590-3667590 on Build GRCh38
rs1051098155 Chr.11: 3667208-3667208 on Build GRCh38
rs763249970 Chr.11: 3667315-3667315 on Build GRCh38
rs765490656 Chr.11: 3667240-3667240 on Build GRCh38
rs759749490 Chr.11: 3667529-3667529 on Build GRCh38
rs750068006 Chr.11: 3667545-3667545 on Build GRCh38
rs373143429 Chr.11: 3667454-3667454 on Build GRCh38
rs765656579 Chr.11: 3667297-3667297 on Build GRCh38
rs780569791 Chr.11: 3667503-3667503 on Build GRCh38
rs557047608 Chr.11: 3667430-3667430 on Build GRCh38
rs375073911 Chr.11: 3667196-3667196 on Build GRCh38
rs772373582 Chr.11: 3667511-3667511 on Build GRCh38
rs117205648 Chr.11: 3667146-3667146 on Build GRCh38
rs201153030 Chr.11: 3667513-3667513 on Build GRCh38
rs747774482 Chr.11: 3667362-3667362 on Build GRCh38
rs988501910 Chr.11: 3667232-3667232 on Build GRCh38
rs776422383 Chr.11: 3667187-3667187 on Build GRCh38
rs762468164 Chr.11: 3667508-3667508 on Build GRCh38
rs779466362 Chr.11: 3667507-3667507 on Build GRCh38
rs1002127205 Chr.11: 3667526-3667526 on Build GRCh38
rs577106168 Chr.11: 3667498-3667498 on Build GRCh38
rs546692965 Chr.11: 3667419-3667419 on Build GRCh38
rs746558230 Chr.11: 3667396-3667396 on Build GRCh38
rs369141816 Chr.11: 3667265-3667265 on Build GRCh38
rs766938926 Chr.11: 3667221-3667221 on Build GRCh38
rs762696179 Chr.11: 3667491-3667491 on Build GRCh38
rs752898080 Chr.11: 3667450-3667451 on Build GRCh38
rs144794962 Chr.11: 3667574-3667574 on Build GRCh38
rs889505791 Chr.11: 3667248-3667248 on Build GRCh38
rs778830088 Chr.11: 3667390-3667390 on Build GRCh38
rs748997800 Chr.11: 3667271-3667271 on Build GRCh38
rs772484008 Chr.11: 3667457-3667457 on Build GRCh38
rs777862613 Chr.11: 3667251-3667251 on Build GRCh38
rs866721767 Chr.11: 3667505-3667505 on Build GRCh38
rs764409011 Chr.11: 3667377-3667377 on Build GRCh38
rs776864644 Chr.11: 3667519-3667519 on Build GRCh38
rs763375115 Chr.11: 3667375-3667375 on Build GRCh38
rs541831012 Chr.11: 3667546-3667546 on Build GRCh38
rs747228623 Chr.11: 3667514-3667514 on Build GRCh38
rs761382564 Chr.11: 3667539-3667539 on Build GRCh38
rs770470937 Chr.11: 3667397-3667397 on Build GRCh38
rs759550632 Chr.11: 3667483-3667483 on Build GRCh38
rs896866390 Chr.11: 3667458-3667458 on Build GRCh38
rs769081824 Chr.11: 3667448-3667448 on Build GRCh38
rs774655790 Chr.11: 3667409-3667409 on Build GRCh38
rs146548502 Chr.11: 3667359-3667359 on Build GRCh38
rs775618164 Chr.11: 3667535-3667535 on Build GRCh38
rs144182744 Chr.11: 3667348-3667348 on Build GRCh38
rs559435979 Chr.11: 3667202-3667202 on Build GRCh38
rs751827575 Chr.11: 3667380-3667380 on Build GRCh38
rs2231543 Chr.11: 3667363-3667363 on Build GRCh38
rs770133301 Chr.11: 3667374-3667374 on Build GRCh38
rs909765043 Chr.11: 3667388-3667411 on Build GRCh38
rs773954149 Chr.11: 3667216-3667216 on Build GRCh38
rs779662314 Chr.11: 3667269-3667269 on Build GRCh38
rs768577494 Chr.11: 3667587-3667587 on Build GRCh38
rs766908055 Chr.11: 3667255-3667256 on Build GRCh38
rs570947917 Chr.11: 3667411-3667411 on Build GRCh38
rs992747674 Chr.11: 3667147-3667147 on Build GRCh38
rs765909178 Chr.11: 3667595-3667595 on Build GRCh38
rs767210580 Chr.11: 3667544-3667544 on Build GRCh38
rs773351290 Chr.11: 3667466-3667466 on Build GRCh38
rs117521524 Chr.11: 3667584-3667584 on Build GRCh38
rs201720948 Chr.11: 3667314-3667314 on Build GRCh38
rs1048686864 Chr.11: 3667490-3667490 on Build GRCh38
rs759939729 Chr.11: 3667296-3667296 on Build GRCh38
rs775669605 Chr.11: 3667580-3667580 on Build GRCh38
rs574550219 Chr.11: 3667530-3667530 on Build GRCh38
rs774706922 Chr.11: 3667452-3667452 on Build GRCh38
rs146877572 Chr.11: 3667253-3667253 on Build GRCh38
rs760275688 Chr.11: 3667593-3667593 on Build GRCh38
rs750393817 Chr.11: 3667336-3667336 on Build GRCh38
rs750549709 Chr.11: 3667171-3667171 on Build GRCh38
rs747340658 Chr.11: 3667568-3667568 on Build GRCh38
rs201793421 Chr.11: 3667504-3667504 on Build GRCh38
rs767271907 Chr.11: 3667591-3667591 on Build GRCh38
rs74442809 Chr.11: 3667255-3667255 on Build GRCh38

View Full Product Details