Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3667521-3668016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA10
Entrez Gene ID
57053
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
Gene Aliases
-
UniGene
Hs.732806
Gene Chromosome Location
Chr.11: 3665587-3673208 on Build GRCh38
Transcripts
NM_001303034.1 NM_001303035.1 NM_020402.3 XM_011520234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672260 c.451C>T p.R151C Chr.11: 3667676-3667676 on Build GRCh38
6436907 c.572C>T p.A191V Chr.11: 3667555-3667555 on Build GRCh38
1201114 c.452G>A p.R151H Chr.11: 3667675-3667675 on Build GRCh38
1201115 c.428G>A p.R143H Chr.11: 3667699-3667699 on Build GRCh38
1201116 c.388G>A p.A130T Chr.11: 3667739-3667739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754115150 Chr.11: 3667813-3667813 on Build GRCh38
rs763250393 Chr.11: 3667689-3667689 on Build GRCh38
rs770709371 Chr.11: 3667764-3667764 on Build GRCh38
rs752128056 Chr.11: 3667707-3667707 on Build GRCh38
rs903349718 Chr.11: 3667954-3667954 on Build GRCh38
rs951079658 Chr.11: 3667831-3667831 on Build GRCh38
rs776387373 Chr.11: 3667770-3667770 on Build GRCh38
rs762423133 Chr.11: 3667790-3667790 on Build GRCh38
rs1017921550 Chr.11: 3667685-3667685 on Build GRCh38
rs976533704 Chr.11: 3667649-3667649 on Build GRCh38
rs936999101 Chr.11: 3667874-3667874 on Build GRCh38
rs771878441 Chr.11: 3667676-3667676 on Build GRCh38
rs1056084643 Chr.11: 3667884-3667884 on Build GRCh38
rs772955497 Chr.11: 3667687-3667687 on Build GRCh38
rs376274152 Chr.11: 3667662-3667662 on Build GRCh38
rs748015833 Chr.11: 3667669-3667669 on Build GRCh38
rs546755450 Chr.11: 3667596-3667596 on Build GRCh38
rs1032692473 Chr.11: 3667582-3667582 on Build GRCh38
rs182159148 Chr.11: 3667573-3667573 on Build GRCh38
rs759060304 Chr.11: 3667771-3667771 on Build GRCh38
rs929777698 Chr.11: 3667565-3667565 on Build GRCh38
rs778006719 Chr.11: 3667566-3667566 on Build GRCh38
rs569150940 Chr.11: 3667744-3667744 on Build GRCh38
rs528669032 Chr.11: 3667589-3667589 on Build GRCh38
rs772904315 Chr.11: 3667592-3667592 on Build GRCh38
rs768192564 Chr.11: 3667795-3667795 on Build GRCh38
rs1041791731 Chr.11: 3667758-3667758 on Build GRCh38
rs749437228 Chr.11: 3667731-3667731 on Build GRCh38
rs2231541 Chr.11: 3667569-3667569 on Build GRCh38
rs947979872 Chr.11: 3667739-3667739 on Build GRCh38
rs1006849025 Chr.11: 3667780-3667780 on Build GRCh38
rs761159846 Chr.11: 3667799-3667799 on Build GRCh38
rs374749795 Chr.11: 3667642-3667642 on Build GRCh38
rs779521433 Chr.11: 3667547-3667547 on Build GRCh38
rs896023477 Chr.11: 3667717-3667717 on Build GRCh38
rs755118049 Chr.11: 3667815-3667815 on Build GRCh38
rs773004798 Chr.11: 3667757-3667757 on Build GRCh38
rs780972264 Chr.11: 3667859-3667859 on Build GRCh38
rs1053064699 Chr.11: 3667608-3667608 on Build GRCh38
rs758227414 Chr.11: 3667570-3667570 on Build GRCh38
rs568235425 Chr.11: 3667661-3667661 on Build GRCh38
rs925301789 Chr.11: 3667821-3667821 on Build GRCh38
rs765657038 Chr.11: 3667562-3667562 on Build GRCh38
rs889342293 Chr.11: 3667663-3667663 on Build GRCh38
rs376864800 Chr.11: 3667916-3667916 on Build GRCh38
rs936631822 Chr.11: 3667827-3667827 on Build GRCh38
rs757799127 Chr.11: 3667713-3667713 on Build GRCh38
rs761731304 Chr.11: 3667590-3667590 on Build GRCh38
rs1033043447 Chr.11: 3667775-3667775 on Build GRCh38
rs778812493 Chr.11: 3667745-3667745 on Build GRCh38
rs750068006 Chr.11: 3667545-3667545 on Build GRCh38
rs769374649 Chr.11: 3667781-3667781 on Build GRCh38
rs766628071 Chr.11: 3667800-3667800 on Build GRCh38
rs918308408 Chr.11: 3667653-3667653 on Build GRCh38
rs756336483 Chr.11: 3667647-3667647 on Build GRCh38
rs200221704 Chr.11: 3667691-3667691 on Build GRCh38
rs984262722 Chr.11: 3667835-3667835 on Build GRCh38
rs368580345 Chr.11: 3667749-3667749 on Build GRCh38
rs781647147 Chr.11: 3667623-3667623 on Build GRCh38
rs370625946 Chr.11: 3667634-3667634 on Build GRCh38
rs774795655 Chr.11: 3667788-3667788 on Build GRCh38
rs755137618 Chr.11: 3667738-3667738 on Build GRCh38
rs757743997 Chr.11: 3667621-3667621 on Build GRCh38
rs750755223 Chr.11: 3667718-3667718 on Build GRCh38
rs774382849 Chr.11: 3667665-3667665 on Build GRCh38
rs910137057 Chr.11: 3667678-3667678 on Build GRCh38
rs144794962 Chr.11: 3667574-3667574 on Build GRCh38
rs374470427 Chr.11: 3667962-3667962 on Build GRCh38
rs917464781 Chr.11: 3667893-3667893 on Build GRCh38
rs965217281 Chr.11: 3667804-3667804 on Build GRCh38
rs756530346 Chr.11: 3667722-3667722 on Build GRCh38
rs780388423 Chr.11: 3667724-3667724 on Build GRCh38
rs541831012 Chr.11: 3667546-3667546 on Build GRCh38
rs147957618 Chr.11: 3667704-3667704 on Build GRCh38
rs2231540 Chr.11: 3667846-3667846 on Build GRCh38
rs746864387 Chr.11: 3667761-3667761 on Build GRCh38
rs530123890 Chr.11: 3667922-3667922 on Build GRCh38
rs767645462 Chr.11: 3667847-3667847 on Build GRCh38
rs758842352 Chr.11: 3667606-3667606 on Build GRCh38
rs771933661 Chr.11: 3667756-3667756 on Build GRCh38
rs768577494 Chr.11: 3667587-3667587 on Build GRCh38
rs898163584 Chr.11: 3667927-3667927 on Build GRCh38
rs1007806721 Chr.11: 3667892-3667892 on Build GRCh38
rs752007041 Chr.11: 3667611-3667611 on Build GRCh38
rs764721805 Chr.11: 3667610-3667610 on Build GRCh38
rs1051920081 Chr.11: 3667862-3667862 on Build GRCh38
rs765909178 Chr.11: 3667595-3667595 on Build GRCh38
rs767210580 Chr.11: 3667544-3667544 on Build GRCh38
rs746695483 Chr.11: 3667700-3667700 on Build GRCh38
rs117521524 Chr.11: 3667584-3667584 on Build GRCh38
rs750957071 Chr.11: 3667798-3667798 on Build GRCh38
rs775669605 Chr.11: 3667580-3667580 on Build GRCh38
rs953485710 Chr.11: 3667776-3667776 on Build GRCh38
rs760275688 Chr.11: 3667593-3667593 on Build GRCh38
rs768610689 Chr.11: 3667664-3667664 on Build GRCh38
rs747340658 Chr.11: 3667568-3667568 on Build GRCh38
rs1000262127 Chr.11: 3667972-3667972 on Build GRCh38
rs933488149 Chr.11: 3667852-3667852 on Build GRCh38
rs769170687 Chr.11: 3667982-3667982 on Build GRCh38
rs918355928 Chr.11: 3667826-3667826 on Build GRCh38
rs767271907 Chr.11: 3667591-3667591 on Build GRCh38

View Full Product Details