Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6217660-6218204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5677069 c.1497G>T p.V499V Chr.11: 6218131-6218131 on Build GRCh38
1355327 c.1566G>A p.Q522Q Chr.11: 6218062-6218062 on Build GRCh38
239777 c.1922C>G p.P641R Chr.11: 6217706-6217706 on Build GRCh38
5717558 c.1873G>A p.E625K Chr.11: 6217755-6217755 on Build GRCh38
6658718 c.1736G>A p.C579Y Chr.11: 6217892-6217892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750991220 Chr.11: 6218063-6218063 on Build GRCh38
rs35946899 Chr.11: 6217814-6217815 on Build GRCh38
rs780305032 Chr.11: 6217731-6217731 on Build GRCh38
rs200718157 Chr.11: 6218085-6218085 on Build GRCh38
rs752380583 Chr.11: 6217707-6217707 on Build GRCh38
rs767140179 Chr.11: 6217704-6217704 on Build GRCh38
rs780574233 Chr.11: 6218067-6218067 on Build GRCh38
rs756472160 Chr.11: 6217886-6217886 on Build GRCh38
rs758921301 Chr.11: 6218065-6218065 on Build GRCh38
rs149101080 Chr.11: 6217964-6217964 on Build GRCh38
rs781556644 Chr.11: 6217709-6217709 on Build GRCh38
rs142507687 Chr.11: 6218128-6218128 on Build GRCh38
rs761564270 Chr.11: 6217848-6217848 on Build GRCh38
rs747771810 Chr.11: 6217998-6217998 on Build GRCh38
rs776935007 Chr.11: 6217736-6217736 on Build GRCh38
rs980107549 Chr.11: 6217948-6217948 on Build GRCh38
rs200796083 Chr.11: 6218035-6218035 on Build GRCh38
rs898411548 Chr.11: 6217743-6217743 on Build GRCh38
rs769501783 Chr.11: 6217939-6217939 on Build GRCh38
rs768259794 Chr.11: 6217910-6217910 on Build GRCh38
rs112171346 Chr.11: 6218126-6218126 on Build GRCh38
rs373976769 Chr.11: 6217985-6217985 on Build GRCh38
rs769044717 Chr.11: 6217735-6217735 on Build GRCh38
rs777105237 Chr.11: 6218133-6218133 on Build GRCh38
rs747205123 Chr.11: 6217734-6217734 on Build GRCh38
rs752939614 Chr.11: 6217878-6217878 on Build GRCh38
rs912460755 Chr.11: 6218147-6218147 on Build GRCh38
rs562285223 Chr.11: 6217802-6217802 on Build GRCh38
rs767027507 Chr.11: 6217748-6217748 on Build GRCh38
rs977431748 Chr.11: 6217694-6217694 on Build GRCh38
rs573849642 Chr.11: 6218089-6218089 on Build GRCh38
rs573156575 Chr.11: 6218024-6218024 on Build GRCh38
rs760362636 Chr.11: 6217759-6217759 on Build GRCh38
rs867759355 Chr.11: 6217737-6217737 on Build GRCh38
rs747764873 Chr.11: 6218076-6218076 on Build GRCh38
rs3750944 Chr.11: 6218114-6218114 on Build GRCh38
rs773762775 Chr.11: 6217739-6217739 on Build GRCh38
rs146982586 Chr.11: 6217896-6217896 on Build GRCh38
rs748524729 Chr.11: 6217811-6217811 on Build GRCh38
rs561748424 Chr.11: 6217692-6217692 on Build GRCh38
rs773004260 Chr.11: 6217941-6217941 on Build GRCh38
rs372679753 Chr.11: 6217819-6217819 on Build GRCh38
rs1039100355 Chr.11: 6218051-6218051 on Build GRCh38
rs756018551 Chr.11: 6217708-6217708 on Build GRCh38
rs771512968 Chr.11: 6217825-6217825 on Build GRCh38
rs550965738 Chr.11: 6217845-6217845 on Build GRCh38
rs757737713 Chr.11: 6218052-6218052 on Build GRCh38
rs745818133 Chr.11: 6218091-6218091 on Build GRCh38
rs139700511 Chr.11: 6217771-6217771 on Build GRCh38
rs373805148 Chr.11: 6217768-6217768 on Build GRCh38
rs903499758 Chr.11: 6217817-6217817 on Build GRCh38
rs781545294 Chr.11: 6217804-6217804 on Build GRCh38
rs765052657 Chr.11: 6217852-6217852 on Build GRCh38
rs759658049 Chr.11: 6217973-6217973 on Build GRCh38
rs374161366 Chr.11: 6217721-6217721 on Build GRCh38
rs1007912924 Chr.11: 6217789-6217789 on Build GRCh38
rs374748276 Chr.11: 6218125-6218125 on Build GRCh38
rs1039332820 Chr.11: 6218176-6218176 on Build GRCh38
rs749302125 Chr.11: 6218169-6218169 on Build GRCh38
rs151025846 Chr.11: 6217684-6217684 on Build GRCh38
rs752890227 Chr.11: 6217976-6217976 on Build GRCh38
rs753986138 Chr.11: 6218048-6218048 on Build GRCh38
rs768588090 Chr.11: 6218104-6218104 on Build GRCh38
rs779612049 Chr.11: 6217994-6217994 on Build GRCh38
rs201971513 Chr.11: 6217738-6217738 on Build GRCh38
rs745664782 Chr.11: 6218018-6218018 on Build GRCh38
rs778681511 Chr.11: 6218178-6218178 on Build GRCh38
rs746348250 Chr.11: 6217995-6217995 on Build GRCh38
rs755042095 Chr.11: 6217876-6217876 on Build GRCh38
rs890037230 Chr.11: 6218026-6218026 on Build GRCh38
rs763409964 Chr.11: 6218118-6218118 on Build GRCh38
rs373679132 Chr.11: 6218113-6218113 on Build GRCh38
rs1011699412 Chr.11: 6217862-6217862 on Build GRCh38
rs1039384609 Chr.11: 6217999-6217999 on Build GRCh38
rs143191716 Chr.11: 6217975-6217975 on Build GRCh38
rs970008269 Chr.11: 6217945-6217945 on Build GRCh38
rs776188632 Chr.11: 6217919-6217919 on Build GRCh38
rs776410782 Chr.11: 6217847-6217847 on Build GRCh38
rs991893923 Chr.11: 6218094-6218094 on Build GRCh38
rs750361793 Chr.11: 6217861-6217861 on Build GRCh38
rs774310556 Chr.11: 6218021-6218021 on Build GRCh38
rs769425560 Chr.11: 6218001-6218001 on Build GRCh38
rs144693760 Chr.11: 6217779-6217779 on Build GRCh38
rs140222037 Chr.11: 6217996-6217996 on Build GRCh38
rs780121887 Chr.11: 6218167-6218167 on Build GRCh38
rs11040809 Chr.11: 6217909-6217909 on Build GRCh38
rs943993301 Chr.11: 6218170-6218170 on Build GRCh38
rs778158841 Chr.11: 6217890-6217890 on Build GRCh38
rs775580477 Chr.11: 6218027-6218027 on Build GRCh38
rs200454641 Chr.11: 6217877-6217877 on Build GRCh38
rs780071825 Chr.11: 6217798-6217798 on Build GRCh38
rs3750943 Chr.11: 6217730-6217730 on Build GRCh38
rs765148272 Chr.11: 6217776-6217776 on Build GRCh38
rs199932613 Chr.11: 6218084-6218084 on Build GRCh38
rs147897958 Chr.11: 6217873-6217873 on Build GRCh38
rs779056263 Chr.11: 6217723-6217723 on Build GRCh38
rs775321826 Chr.11: 6217749-6217749 on Build GRCh38
rs760417398 Chr.11: 6217835-6217835 on Build GRCh38
rs779714573 Chr.11: 6217908-6217908 on Build GRCh38
rs532824166 Chr.11: 6217809-6217809 on Build GRCh38
rs770178853 Chr.11: 6217814-6217814 on Build GRCh38
rs375709716 Chr.11: 6217968-6217968 on Build GRCh38
rs369543757 Chr.11: 6218103-6218103 on Build GRCh38
rs148185483 Chr.11: 6217977-6217977 on Build GRCh38
rs775090698 Chr.11: 6217831-6217831 on Build GRCh38
rs766971976 Chr.11: 6218123-6218123 on Build GRCh38
rs759021696 Chr.11: 6217744-6217744 on Build GRCh38
rs770686694 Chr.11: 6218015-6218015 on Build GRCh38
rs753857459 Chr.11: 6217720-6217720 on Build GRCh38
rs770329044 Chr.11: 6217997-6217997 on Build GRCh38
rs572973529 Chr.11: 6217879-6217879 on Build GRCh38
rs201174707 Chr.11: 6217864-6217864 on Build GRCh38
rs765659855 Chr.11: 6218053-6218053 on Build GRCh38
rs368836421 Chr.11: 6217971-6217971 on Build GRCh38
rs759228110 Chr.11: 6217696-6217696 on Build GRCh38
rs754164103 Chr.11: 6217979-6217979 on Build GRCh38
rs866984735 Chr.11: 6217851-6217851 on Build GRCh38
rs201235818 Chr.11: 6217829-6217829 on Build GRCh38
rs929758819 Chr.11: 6218136-6218136 on Build GRCh38
rs575632431 Chr.11: 6217815-6217815 on Build GRCh38
rs756844079 Chr.11: 6218146-6218146 on Build GRCh38
rs201955204 Chr.11: 6218012-6218012 on Build GRCh38
rs755570221 Chr.11: 6218079-6218079 on Build GRCh38
rs761488090 Chr.11: 6217920-6217920 on Build GRCh38
rs375469284 Chr.11: 6218102-6218102 on Build GRCh38
rs762272894 Chr.11: 6217691-6217691 on Build GRCh38
rs758448400 Chr.11: 6217795-6217795 on Build GRCh38
rs150866102 Chr.11: 6218142-6218142 on Build GRCh38
rs868028265 Chr.11: 6217740-6217740 on Build GRCh38
rs141687987 Chr.11: 6217895-6217895 on Build GRCh38
rs770041110 Chr.11: 6218160-6218161 on Build GRCh38
rs1056262109 Chr.11: 6217716-6217716 on Build GRCh38
rs984692768 Chr.11: 6217903-6217903 on Build GRCh38
rs938830089 Chr.11: 6217741-6217741 on Build GRCh38
rs755137939 Chr.11: 6217803-6217803 on Build GRCh38
rs760811561 Chr.11: 6218029-6218029 on Build GRCh38
rs372504079 Chr.11: 6218080-6218080 on Build GRCh38
rs772738564 Chr.11: 6218014-6218014 on Build GRCh38

View Full Product Details