Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6394842-6395342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB1
Entrez Gene ID
322
Gene Name
amyloid beta precursor protein binding family B member 1
Gene Aliases
FE65, MGC:9072, RIR
UniGene
Hs.372840
Gene Chromosome Location
Chr.11: 6395124-6419830 on Build GRCh38
Transcripts
NM_001164.4 NM_001257319.2 NM_001257320.2 NM_001257321.2 NM_001257323.2 NM_001257325.2 NM_001257326.2 NM_145689.2 NR_047512.2 XM_006718209.3 XM_006718210.3 XM_006718211.1 XM_006718212.3 XM_006718213.1 XM_006718214.3 XM_006718215.3 XM_011520037.2 XM_011520038.2 XM_011520040.2 XM_011520042.1 XM_017017639.1 XM_017017640.1 XM_017017641.1 XM_017017642.1 NM_000543.4 NM_001007593.2 NM_001318087.1 NM_001318088.1 NR_027400.2 NR_134502.1 XM_011520303.1

Gene Symbol
SMPD1
Entrez Gene ID
6609
Gene Name
sphingomyelin phosphodiesterase 1
Gene Aliases
ASM, ASMASE, NPD
UniGene
Hs.498173
Gene Chromosome Location
Chr.11: 6390301-6394998 on Build GRCh38
Transcripts
NM_001164.4 NM_001257319.2 NM_001257320.2 NM_001257321.2 NM_001257323.2 NM_001257325.2 NM_001257326.2 NM_145689.2 NR_047512.2 XM_006718209.3 XM_006718210.3 XM_006718211.1 XM_006718212.3 XM_006718213.1 XM_006718214.3 XM_006718215.3 XM_011520037.2 XM_011520038.2 XM_011520040.2 XM_011520042.1 XM_017017639.1 XM_017017640.1 XM_017017641.1 XM_017017642.1 NM_000543.4 NM_001007593.2 NM_001318087.1 NM_001318088.1 NR_027400.2 NR_134502.1 XM_011520303.1

SNP Detail

SNP ID Chromosome Location
rs763466464 Chr.11: 6395308-6395308 on Build GRCh38
rs939915609 Chr.11: 6395301-6395301 on Build GRCh38
rs1048221627 Chr.11: 6395282-6395282 on Build GRCh38
rs1041142773 Chr.11: 6395307-6395307 on Build GRCh38
rs551064846 Chr.11: 6395001-6395001 on Build GRCh38
rs1048358296 Chr.11: 6394955-6394955 on Build GRCh38
rs894066990 Chr.11: 6394979-6394979 on Build GRCh38
rs970582852 Chr.11: 6395185-6395185 on Build GRCh38
rs534098273 Chr.11: 6395204-6395204 on Build GRCh38
rs948794356 Chr.11: 6394900-6394900 on Build GRCh38
rs931145599 Chr.11: 6395246-6395246 on Build GRCh38
rs908459842 Chr.11: 6395296-6395296 on Build GRCh38
rs764479787 Chr.11: 6395197-6395197 on Build GRCh38
rs956257923 Chr.11: 6395138-6395138 on Build GRCh38
rs1025858354 Chr.11: 6395183-6395183 on Build GRCh38
rs763378997 Chr.11: 6395171-6395171 on Build GRCh38
rs757971873 Chr.11: 6395110-6395110 on Build GRCh38
rs1000298240 Chr.11: 6395130-6395130 on Build GRCh38
rs566809907 Chr.11: 6395181-6395181 on Build GRCh38
rs952570644 Chr.11: 6395187-6395187 on Build GRCh38
rs551779872 Chr.11: 6395090-6395090 on Build GRCh38
rs886993941 Chr.11: 6394982-6394982 on Build GRCh38
rs916490216 Chr.11: 6395226-6395226 on Build GRCh38
rs765829755 Chr.11: 6395067-6395067 on Build GRCh38
rs35636371 Chr.11: 6394888-6394889 on Build GRCh38
rs928017026 Chr.11: 6394892-6394892 on Build GRCh38
rs900956875 Chr.11: 6395047-6395047 on Build GRCh38
rs561607572 Chr.11: 6395104-6395104 on Build GRCh38
rs542620529 Chr.11: 6395040-6395040 on Build GRCh38
rs920650145 Chr.11: 6395233-6395233 on Build GRCh38
rs750067292 Chr.11: 6395213-6395213 on Build GRCh38
rs561965709 Chr.11: 6394910-6394910 on Build GRCh38
rs926470933 Chr.11: 6394933-6394933 on Build GRCh38
rs12278115 Chr.11: 6394901-6394901 on Build GRCh38
rs996601997 Chr.11: 6395055-6395055 on Build GRCh38
rs371122491 Chr.11: 6395064-6395064 on Build GRCh38
rs143357514 Chr.11: 6395082-6395082 on Build GRCh38
rs555651187 Chr.11: 6395220-6395220 on Build GRCh38
rs992793825 Chr.11: 6395156-6395156 on Build GRCh38
rs1031897642 Chr.11: 6395132-6395132 on Build GRCh38
rs373270465 Chr.11: 6394947-6394947 on Build GRCh38
rs530932169 Chr.11: 6395180-6395181 on Build GRCh38
rs1003887233 Chr.11: 6395034-6395034 on Build GRCh38
rs528743699 Chr.11: 6394879-6394879 on Build GRCh38

View Full Product Details