Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30330720-30331252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL14EP
Entrez Gene ID
120534
Gene Name
ADP ribosylation factor like GTPase 14 effector protein
Gene Aliases
ARF7EP, C11orf46, dJ299F11.1
UniGene
Hs.502208
Gene Chromosome Location
Chr.11: 30323074-30338458 on Build GRCh38
Transcripts
NM_152316.2 XM_005252792.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1703769 c.289G>A p.D97N Chr.11: 30331237-30331237 on Build GRCh38
1638814 c.267T>A p.N89K Chr.11: 30331215-30331215 on Build GRCh38
2113890 c.268T>G p.L90V Chr.11: 30331216-30331216 on Build GRCh38
6068266 c.283T>G p.L95V Chr.11: 30331231-30331231 on Build GRCh38
5693314 c.179A>G p.H60R Chr.11: 30331127-30331127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367725488 Chr.11: 30330946-30330946 on Build GRCh38
rs748205923 Chr.11: 30330948-30330948 on Build GRCh38
rs369708297 Chr.11: 30330982-30330982 on Build GRCh38
rs982764932 Chr.11: 30330773-30330773 on Build GRCh38
rs145619728 Chr.11: 30330927-30330928 on Build GRCh38
rs762400028 Chr.11: 30331215-30331215 on Build GRCh38
rs193033090 Chr.11: 30330841-30330841 on Build GRCh38
rs994629542 Chr.11: 30330975-30330975 on Build GRCh38
rs149778867 Chr.11: 30330925-30330926 on Build GRCh38
rs778717975 Chr.11: 30330817-30330817 on Build GRCh38
rs144537045 Chr.11: 30331011-30331011 on Build GRCh38
rs373951489 Chr.11: 30330752-30330752 on Build GRCh38
rs34392551 Chr.11: 30330928-30330929 on Build GRCh38
rs199851936 Chr.11: 30330915-30330915 on Build GRCh38
rs775434354 Chr.11: 30330979-30330979 on Build GRCh38
rs768218235 Chr.11: 30330987-30330987 on Build GRCh38
rs747967388 Chr.11: 30330900-30330900 on Build GRCh38
rs796327778 Chr.11: 30330782-30330785 on Build GRCh38
rs552646054 Chr.11: 30330779-30330779 on Build GRCh38
rs745854567 Chr.11: 30330962-30330962 on Build GRCh38
rs201186401 Chr.11: 30330924-30330924 on Build GRCh38
rs769433126 Chr.11: 30331170-30331170 on Build GRCh38
rs781424349 Chr.11: 30330947-30330947 on Build GRCh38
rs568629489 Chr.11: 30330999-30330999 on Build GRCh38
rs747441334 Chr.11: 30330997-30330997 on Build GRCh38
rs988856430 Chr.11: 30331224-30331224 on Build GRCh38
rs753010989 Chr.11: 30331078-30331078 on Build GRCh38
rs773468802 Chr.11: 30330984-30330986 on Build GRCh38
rs377036789 Chr.11: 30331148-30331148 on Build GRCh38
rs978526208 Chr.11: 30331221-30331221 on Build GRCh38
rs936115368 Chr.11: 30330804-30330804 on Build GRCh38
rs767739323 Chr.11: 30331072-30331072 on Build GRCh38
rs772696912 Chr.11: 30331012-30331012 on Build GRCh38
rs768536604 Chr.11: 30331141-30331141 on Build GRCh38
rs978599910 Chr.11: 30330822-30330822 on Build GRCh38
rs950261239 Chr.11: 30330908-30330908 on Build GRCh38
rs1008789673 Chr.11: 30331015-30331015 on Build GRCh38
rs747893357 Chr.11: 30331161-30331161 on Build GRCh38
rs952781125 Chr.11: 30330756-30330756 on Build GRCh38
rs1027066499 Chr.11: 30330754-30330754 on Build GRCh38
rs780454680 Chr.11: 30330927-30330927 on Build GRCh38
rs139402064 Chr.11: 30331010-30331010 on Build GRCh38
rs746813844 Chr.11: 30331131-30331131 on Build GRCh38
rs200464693 Chr.11: 30331119-30331119 on Build GRCh38
rs749318531 Chr.11: 30331102-30331102 on Build GRCh38
rs758427360 Chr.11: 30331107-30331107 on Build GRCh38
rs773429758 Chr.11: 30330968-30330968 on Build GRCh38
rs867256019 Chr.11: 30330958-30330958 on Build GRCh38
rs965085962 Chr.11: 30330765-30330765 on Build GRCh38
rs772236750 Chr.11: 30330967-30330967 on Build GRCh38
rs900119766 Chr.11: 30330955-30330955 on Build GRCh38
rs751883424 Chr.11: 30330929-30330929 on Build GRCh38
rs766901543 Chr.11: 30331218-30331218 on Build GRCh38
rs765765765 Chr.11: 30330905-30330905 on Build GRCh38
rs925671354 Chr.11: 30330884-30330884 on Build GRCh38
rs199566595 Chr.11: 30331019-30331019 on Build GRCh38
rs79806526 Chr.11: 30330926-30330926 on Build GRCh38
rs200948615 Chr.11: 30330924-30330925 on Build GRCh38
rs964681676 Chr.11: 30331081-30331081 on Build GRCh38
rs777974752 Chr.11: 30330959-30330959 on Build GRCh38
rs770030975 Chr.11: 30330951-30330951 on Build GRCh38
rs370388471 Chr.11: 30331204-30331204 on Build GRCh38
rs530987323 Chr.11: 30330764-30330764 on Build GRCh38
rs371036775 Chr.11: 30331095-30331095 on Build GRCh38
rs35824273 Chr.11: 30330926-30330927 on Build GRCh38
rs375725496 Chr.11: 30331018-30331018 on Build GRCh38
rs528815106 Chr.11: 30330857-30330857 on Build GRCh38
rs751890528 Chr.11: 30331064-30331064 on Build GRCh38
rs926010983 Chr.11: 30330784-30330784 on Build GRCh38

View Full Product Details