Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 30332699-30333215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL14EP
Entrez Gene ID
120534
Gene Name
ADP ribosylation factor like GTPase 14 effector protein
Gene Aliases
ARF7EP, C11orf46, dJ299F11.1
UniGene
Hs.502208
Gene Chromosome Location
Chr.11: 30323074-30338458 on Build GRCh38
Transcripts
NM_152316.2 XM_005252792.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838406 c.464C>T p.T155M Chr.11: 30332903-30332903 on Build GRCh38
4496227 c.470C>T p.P157L Chr.11: 30332909-30332909 on Build GRCh38
6579537 c.554G>C p.R185T Chr.11: 30332993-30332993 on Build GRCh38
5635740 c.490G>T p.A164S Chr.11: 30332929-30332929 on Build GRCh38
3383418 c.554+6C>T p.? Chr.11: 30332999-30332999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027114050 Chr.11: 30332919-30332919 on Build GRCh38
rs148960883 Chr.11: 30332980-30332980 on Build GRCh38
rs537805439 Chr.11: 30333169-30333169 on Build GRCh38
rs1029117682 Chr.11: 30333020-30333020 on Build GRCh38
rs753653286 Chr.11: 30333037-30333037 on Build GRCh38
rs762899273 Chr.11: 30332892-30332892 on Build GRCh38
rs901719415 Chr.11: 30332962-30332962 on Build GRCh38
rs538139502 Chr.11: 30333192-30333192 on Build GRCh38
rs765310058 Chr.11: 30332880-30332880 on Build GRCh38
rs777892259 Chr.11: 30333006-30333006 on Build GRCh38
rs760606587 Chr.11: 30333027-30333027 on Build GRCh38
rs377605857 Chr.11: 30332997-30332997 on Build GRCh38
rs561932163 Chr.11: 30332768-30332768 on Build GRCh38
rs913347549 Chr.11: 30332922-30332922 on Build GRCh38
rs977249200 Chr.11: 30333131-30333131 on Build GRCh38
rs560204891 Chr.11: 30332904-30332904 on Build GRCh38
rs778486161 Chr.11: 30332827-30332827 on Build GRCh38
rs945902970 Chr.11: 30333130-30333130 on Build GRCh38
rs372258835 Chr.11: 30332929-30332929 on Build GRCh38
rs1040404580 Chr.11: 30332934-30332934 on Build GRCh38
rs753101149 Chr.11: 30333074-30333074 on Build GRCh38
rs992269671 Chr.11: 30332808-30332808 on Build GRCh38
rs370929705 Chr.11: 30333001-30333001 on Build GRCh38
rs376948895 Chr.11: 30332861-30332861 on Build GRCh38
rs763958964 Chr.11: 30333033-30333033 on Build GRCh38
rs376788007 Chr.11: 30333021-30333021 on Build GRCh38
rs750537823 Chr.11: 30332885-30332885 on Build GRCh38
rs1034590504 Chr.11: 30333140-30333140 on Build GRCh38
rs971593055 Chr.11: 30333098-30333098 on Build GRCh38
rs752519318 Chr.11: 30332819-30332819 on Build GRCh38
rs376544225 Chr.11: 30332933-30332933 on Build GRCh38
rs914378422 Chr.11: 30333051-30333051 on Build GRCh38
rs747578502 Chr.11: 30332957-30332957 on Build GRCh38
rs961716639 Chr.11: 30332797-30332797 on Build GRCh38
rs562808490 Chr.11: 30332802-30332802 on Build GRCh38
rs544489694 Chr.11: 30332785-30332785 on Build GRCh38
rs529619941 Chr.11: 30332772-30332772 on Build GRCh38
rs766451036 Chr.11: 30332897-30332897 on Build GRCh38
rs1004438438 Chr.11: 30332961-30332961 on Build GRCh38
rs549012186 Chr.11: 30333151-30333151 on Build GRCh38
rs376320482 Chr.11: 30332729-30332729 on Build GRCh38
rs953095594 Chr.11: 30333034-30333034 on Build GRCh38
rs1879671 Chr.11: 30333160-30333160 on Build GRCh38
rs779692150 Chr.11: 30332942-30332942 on Build GRCh38
rs938631542 Chr.11: 30333185-30333185 on Build GRCh38
rs375200089 Chr.11: 30332821-30332821 on Build GRCh38
rs757889945 Chr.11: 30332938-30332938 on Build GRCh38
rs777607290 Chr.11: 30332903-30332903 on Build GRCh38
rs756944838 Chr.11: 30332921-30332921 on Build GRCh38
rs1041631655 Chr.11: 30332773-30332773 on Build GRCh38
rs374008062 Chr.11: 30333017-30333017 on Build GRCh38
rs186043737 Chr.11: 30333112-30333112 on Build GRCh38
rs145144020 Chr.11: 30332860-30332860 on Build GRCh38
rs777124285 Chr.11: 30332863-30332863 on Build GRCh38
rs541639979 Chr.11: 30332896-30332896 on Build GRCh38
rs894551530 Chr.11: 30332837-30332837 on Build GRCh38
rs144998106 Chr.11: 30332965-30332965 on Build GRCh38
rs756966084 Chr.11: 30333043-30333043 on Build GRCh38
rs369817567 Chr.11: 30333005-30333005 on Build GRCh38
rs772659806 Chr.11: 30332835-30332835 on Build GRCh38
rs75912598 Chr.11: 30332822-30332822 on Build GRCh38
rs752680347 Chr.11: 30332898-30332898 on Build GRCh38
rs368006380 Chr.11: 30332870-30332870 on Build GRCh38
rs776104834 Chr.11: 30332843-30332843 on Build GRCh38
rs373627092 Chr.11: 30332824-30332824 on Build GRCh38
rs552157674 Chr.11: 30332831-30332831 on Build GRCh38
rs756029099 Chr.11: 30332901-30332901 on Build GRCh38
rs368229438 Chr.11: 30333054-30333054 on Build GRCh38
rs945933916 Chr.11: 30332755-30332755 on Build GRCh38
rs753560560 Chr.11: 30332909-30332909 on Build GRCh38
rs966044626 Chr.11: 30332979-30332979 on Build GRCh38
rs1001730938 Chr.11: 30333104-30333104 on Build GRCh38
rs989867147 Chr.11: 30333050-30333050 on Build GRCh38
rs777142429 Chr.11: 30332973-30332973 on Build GRCh38
rs200109301 Chr.11: 30332966-30332966 on Build GRCh38
rs7940297 Chr.11: 30332977-30332977 on Build GRCh38

View Full Product Details