Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34152454-34152964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
UniGene
Hs.23361
Gene Chromosome Location
Chr.11: 34150987-34358008 on Build GRCh38
Transcripts
NM_145804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7255415 c.2985C>A p.R995R Chr.11: 34152480-34152480 on Build GRCh38
4809746 c.2890G>T p.A964S Chr.11: 34152575-34152575 on Build GRCh38
5960403 c.2905_2906insA p.L969fs*4 Chr.11: 34152559-34152560 on Build GRCh38
158834 c.2332G>T p.A778S Chr.11: 34152575-34152575 on Build GRCh38
7255416 c.2427C>A p.R809R Chr.11: 34152480-34152480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919069734 Chr.11: 34152599-34152599 on Build GRCh38
rs771979543 Chr.11: 34152905-34152905 on Build GRCh38
rs747587891 Chr.11: 34152597-34152597 on Build GRCh38
rs767404023 Chr.11: 34152534-34152534 on Build GRCh38
rs114282348 Chr.11: 34152577-34152577 on Build GRCh38
rs747581476 Chr.11: 34152497-34152497 on Build GRCh38
rs903843535 Chr.11: 34152912-34152912 on Build GRCh38
rs1042345116 Chr.11: 34152647-34152647 on Build GRCh38
rs762053097 Chr.11: 34152548-34152548 on Build GRCh38
rs111977288 Chr.11: 34152830-34152830 on Build GRCh38
rs764176017 Chr.11: 34152561-34152562 on Build GRCh38
rs200861449 Chr.11: 34152558-34152558 on Build GRCh38
rs775301186 Chr.11: 34152632-34152632 on Build GRCh38
rs919192984 Chr.11: 34152920-34152920 on Build GRCh38
rs141021753 Chr.11: 34152499-34152499 on Build GRCh38
rs867083138 Chr.11: 34152768-34152768 on Build GRCh38
rs904539978 Chr.11: 34152703-34152703 on Build GRCh38
rs972999979 Chr.11: 34152786-34152786 on Build GRCh38
rs772897792 Chr.11: 34152908-34152908 on Build GRCh38
rs370813934 Chr.11: 34152859-34152859 on Build GRCh38
rs1042313847 Chr.11: 34152897-34152897 on Build GRCh38
rs199813409 Chr.11: 34152500-34152500 on Build GRCh38
rs745976915 Chr.11: 34152675-34152675 on Build GRCh38
rs141885689 Chr.11: 34152527-34152527 on Build GRCh38
rs150248775 Chr.11: 34152509-34152509 on Build GRCh38
rs775427834 Chr.11: 34152559-34152560 on Build GRCh38
rs999087686 Chr.11: 34152712-34152712 on Build GRCh38
rs757220169 Chr.11: 34152563-34152569 on Build GRCh38
rs751453280 Chr.11: 34152562-34152563 on Build GRCh38
rs926433204 Chr.11: 34152842-34152842 on Build GRCh38
rs1001343116 Chr.11: 34152666-34152666 on Build GRCh38
rs889845485 Chr.11: 34152926-34152926 on Build GRCh38
rs769310495 Chr.11: 34152601-34152601 on Build GRCh38
rs527338832 Chr.11: 34152927-34152927 on Build GRCh38
rs375877700 Chr.11: 34152618-34152618 on Build GRCh38
rs777358324 Chr.11: 34152607-34152607 on Build GRCh38
rs371133755 Chr.11: 34152481-34152481 on Build GRCh38
rs374923576 Chr.11: 34152646-34152646 on Build GRCh38
rs980517771 Chr.11: 34152801-34152801 on Build GRCh38
rs527753386 Chr.11: 34152779-34152779 on Build GRCh38
rs200312319 Chr.11: 34152486-34152486 on Build GRCh38
rs1056261607 Chr.11: 34152869-34152869 on Build GRCh38
rs145850345 Chr.11: 34152525-34152525 on Build GRCh38
rs747667990 Chr.11: 34152506-34152506 on Build GRCh38
rs748418529 Chr.11: 34152560-34152560 on Build GRCh38
rs748973932 Chr.11: 34152609-34152609 on Build GRCh38
rs529082930 Chr.11: 34152890-34152890 on Build GRCh38
rs762680883 Chr.11: 34152522-34152522 on Build GRCh38
rs2473929 Chr.11: 34152749-34152749 on Build GRCh38
rs551709563 Chr.11: 34152692-34152692 on Build GRCh38
rs1050262061 Chr.11: 34152922-34152922 on Build GRCh38
rs147031456 Chr.11: 34152553-34152553 on Build GRCh38
rs772043041 Chr.11: 34152631-34152631 on Build GRCh38
rs1024716902 Chr.11: 34152765-34152765 on Build GRCh38
rs146303905 Chr.11: 34152549-34152549 on Build GRCh38
rs984930334 Chr.11: 34152833-34152833 on Build GRCh38
rs774174580 Chr.11: 34152619-34152619 on Build GRCh38
rs937766599 Chr.11: 34152858-34152858 on Build GRCh38
rs114735728 Chr.11: 34152663-34152663 on Build GRCh38
rs781613403 Chr.11: 34152591-34152591 on Build GRCh38
rs1032356962 Chr.11: 34152745-34152745 on Build GRCh38
rs969163952 Chr.11: 34152794-34152794 on Build GRCh38
rs796710234 Chr.11: 34152611-34152611 on Build GRCh38
rs933169602 Chr.11: 34152606-34152606 on Build GRCh38
rs759487474 Chr.11: 34152528-34152528 on Build GRCh38
rs917767341 Chr.11: 34152870-34152870 on Build GRCh38
rs757533385 Chr.11: 34152479-34152479 on Build GRCh38
rs746167793 Chr.11: 34152482-34152482 on Build GRCh38
rs963056353 Chr.11: 34152862-34152862 on Build GRCh38
rs373469123 Chr.11: 34152778-34152778 on Build GRCh38
rs114304923 Chr.11: 34152485-34152485 on Build GRCh38
rs772858597 Chr.11: 34152514-34152514 on Build GRCh38
rs751915029 Chr.11: 34152572-34152572 on Build GRCh38
rs1014864321 Chr.11: 34152758-34152758 on Build GRCh38
rs753941809 Chr.11: 34152537-34152537 on Build GRCh38
rs547897067 Chr.11: 34152828-34152828 on Build GRCh38
rs760608385 Chr.11: 34152630-34152630 on Build GRCh38
rs774395166 Chr.11: 34152759-34152759 on Build GRCh38

View Full Product Details