Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35176492-35176985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
UniGene
Hs.502328
Gene Chromosome Location
Chr.11: 35138870-35232402 on Build GRCh38
Transcripts
NM_000610.3 NM_001001389.1 NM_001001390.1 NM_001001391.1 NM_001001392.1 NM_001202555.1 NM_001202556.1 NM_001202557.1 XM_005253231.2 XM_005253232.2 XM_005253233.2 XM_005253234.2 XM_005253235.2 XM_005253238.2 XM_005253239.2 XM_005253240.2 XM_006718388.1 XM_006718390.3 XM_011520482.1 XM_011520483.1 XM_011520484.1 XM_011520485.1 XM_011520486.1 XM_011520487.2 XM_011520488.1 XM_011520489.2 XM_017018583.1 XM_017018584.1 XM_017018585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
299864 c.92C>T p.A31V Chr.11: 35176599-35176599 on Build GRCh38
3809156 c.199G>T p.E67* Chr.11: 35176706-35176706 on Build GRCh38
4468552 c.154C>T p.L52F Chr.11: 35176661-35176661 on Build GRCh38
6241822 c.212G>A p.S71N Chr.11: 35176719-35176719 on Build GRCh38
3967481 c.85C>T p.R29C Chr.11: 35176592-35176592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780409682 Chr.11: 35176730-35176730 on Build GRCh38
rs967250097 Chr.11: 35176693-35176693 on Build GRCh38
rs757127128 Chr.11: 35176628-35176628 on Build GRCh38
rs369473842 Chr.11: 35176644-35176644 on Build GRCh38
rs773706615 Chr.11: 35176533-35176533 on Build GRCh38
rs748214228 Chr.11: 35176756-35176756 on Build GRCh38
rs771915810 Chr.11: 35176633-35176633 on Build GRCh38
rs756890802 Chr.11: 35176723-35176723 on Build GRCh38
rs917922889 Chr.11: 35176881-35176881 on Build GRCh38
rs755467634 Chr.11: 35176548-35176548 on Build GRCh38
rs778704529 Chr.11: 35176743-35176743 on Build GRCh38
rs753441551 Chr.11: 35176708-35176708 on Build GRCh38
rs924557845 Chr.11: 35176955-35176956 on Build GRCh38
rs957377561 Chr.11: 35176567-35176567 on Build GRCh38
rs763103888 Chr.11: 35176648-35176648 on Build GRCh38
rs138821226 Chr.11: 35176697-35176697 on Build GRCh38
rs1055561529 Chr.11: 35176798-35176798 on Build GRCh38
rs763715662 Chr.11: 35176706-35176706 on Build GRCh38
rs369701383 Chr.11: 35176784-35176784 on Build GRCh38
rs777472273 Chr.11: 35176586-35176586 on Build GRCh38
rs543786312 Chr.11: 35176654-35176654 on Build GRCh38
rs1049204502 Chr.11: 35176924-35176924 on Build GRCh38
rs151338790 Chr.11: 35176947-35176947 on Build GRCh38
rs749053466 Chr.11: 35176612-35176612 on Build GRCh38
rs916654275 Chr.11: 35176818-35176818 on Build GRCh38
rs548473416 Chr.11: 35176755-35176755 on Build GRCh38
rs764483939 Chr.11: 35176904-35176904 on Build GRCh38
rs763412638 Chr.11: 35176535-35176535 on Build GRCh38
rs755752798 Chr.11: 35176568-35176568 on Build GRCh38
rs1033752531 Chr.11: 35176817-35176817 on Build GRCh38
rs749457039 Chr.11: 35176758-35176758 on Build GRCh38
rs928571731 Chr.11: 35176721-35176721 on Build GRCh38
rs1006831435 Chr.11: 35176807-35176807 on Build GRCh38
rs768073670 Chr.11: 35176696-35176696 on Build GRCh38
rs374806320 Chr.11: 35176757-35176757 on Build GRCh38
rs762067831 Chr.11: 35176523-35176523 on Build GRCh38
rs755758051 Chr.11: 35176550-35176551 on Build GRCh38
rs745778696 Chr.11: 35176631-35176631 on Build GRCh38
rs760222232 Chr.11: 35176698-35176698 on Build GRCh38
rs779685882 Chr.11: 35176556-35176556 on Build GRCh38
rs766771461 Chr.11: 35176538-35176538 on Build GRCh38
rs4756196 Chr.11: 35176561-35176561 on Build GRCh38
rs765612909 Chr.11: 35176524-35176524 on Build GRCh38
rs568705475 Chr.11: 35176857-35176857 on Build GRCh38
rs77499795 Chr.11: 35176891-35176891 on Build GRCh38
rs375417895 Chr.11: 35176777-35176777 on Build GRCh38
rs755323887 Chr.11: 35176858-35176858 on Build GRCh38
rs989971374 Chr.11: 35176613-35176613 on Build GRCh38
rs965689724 Chr.11: 35176530-35176530 on Build GRCh38
rs150209357 Chr.11: 35176667-35176667 on Build GRCh38
rs539408862 Chr.11: 35176964-35176964 on Build GRCh38
rs74594564 Chr.11: 35176792-35176792 on Build GRCh38
rs557534635 Chr.11: 35176908-35176908 on Build GRCh38
rs121909545 Chr.11: 35176643-35176643 on Build GRCh38
rs758352011 Chr.11: 35176745-35176745 on Build GRCh38
rs773614663 Chr.11: 35176646-35176646 on Build GRCh38
rs766988805 Chr.11: 35176556-35176556 on Build GRCh38
rs750242872 Chr.11: 35176744-35176744 on Build GRCh38
rs752052394 Chr.11: 35176546-35176546 on Build GRCh38
rs774579206 Chr.11: 35176663-35176663 on Build GRCh38

View Full Product Details