Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35198032-35198534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
UniGene
Hs.502328
Gene Chromosome Location
Chr.11: 35138870-35232402 on Build GRCh38
Transcripts
NM_000610.3 NM_001001389.1 NM_001001390.1 NM_001001391.1 NM_001001392.1 NM_001202555.1 NM_001202556.1 NM_001202557.1 XM_005253231.2 XM_005253232.2 XM_005253233.2 XM_005253234.2 XM_005253235.2 XM_005253238.2 XM_005253239.2 XM_005253240.2 XM_006718388.1 XM_006718390.3 XM_011520482.1 XM_011520483.1 XM_011520484.1 XM_011520485.1 XM_011520486.1 XM_011520487.2 XM_011520488.1 XM_011520489.2 XM_017018583.1 XM_017018584.1 XM_017018585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623783 c.800C>T p.T267M Chr.11: 35198124-35198124 on Build GRCh38
370396 c.802T>A p.S268T Chr.11: 35198126-35198126 on Build GRCh38
4191223 c.883T>C p.S295P Chr.11: 35198207-35198207 on Build GRCh38
6068497 c.898G>T p.D300Y Chr.11: 35198222-35198222 on Build GRCh38
7288009 c.801G>A p.T267T Chr.11: 35198125-35198125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925670416 Chr.11: 35198437-35198437 on Build GRCh38
rs761529122 Chr.11: 35198294-35198294 on Build GRCh38
rs569594035 Chr.11: 35198468-35198468 on Build GRCh38
rs771444004 Chr.11: 35198122-35198122 on Build GRCh38
rs773617820 Chr.11: 35198112-35198112 on Build GRCh38
rs149929466 Chr.11: 35198367-35198367 on Build GRCh38
rs370440642 Chr.11: 35198238-35198238 on Build GRCh38
rs746320462 Chr.11: 35198245-35198245 on Build GRCh38
rs1010080122 Chr.11: 35198356-35198356 on Build GRCh38
rs948237099 Chr.11: 35198219-35198219 on Build GRCh38
rs753639479 Chr.11: 35198153-35198153 on Build GRCh38
rs748322693 Chr.11: 35198087-35198087 on Build GRCh38
rs200983620 Chr.11: 35198285-35198285 on Build GRCh38
rs1043385450 Chr.11: 35198381-35198381 on Build GRCh38
rs370026708 Chr.11: 35198155-35198155 on Build GRCh38
rs191756046 Chr.11: 35198083-35198083 on Build GRCh38
rs987647910 Chr.11: 35198377-35198377 on Build GRCh38
rs765105797 Chr.11: 35198169-35198169 on Build GRCh38
rs747065666 Chr.11: 35198081-35198081 on Build GRCh38
rs377112972 Chr.11: 35198124-35198124 on Build GRCh38
rs58757235 Chr.11: 35198433-35198433 on Build GRCh38
rs763780713 Chr.11: 35198281-35198281 on Build GRCh38
rs200817484 Chr.11: 35198084-35198084 on Build GRCh38
rs772629451 Chr.11: 35198250-35198250 on Build GRCh38
rs920743223 Chr.11: 35198214-35198214 on Build GRCh38
rs777902530 Chr.11: 35198228-35198228 on Build GRCh38
rs367954296 Chr.11: 35198191-35198191 on Build GRCh38
rs537829693 Chr.11: 35198505-35198505 on Build GRCh38
rs760366186 Chr.11: 35198145-35198145 on Build GRCh38
rs778757462 Chr.11: 35198071-35198071 on Build GRCh38
rs754440585 Chr.11: 35198317-35198317 on Build GRCh38
rs949867832 Chr.11: 35198496-35198496 on Build GRCh38
rs1052911356 Chr.11: 35198475-35198475 on Build GRCh38
rs368245075 Chr.11: 35198474-35198474 on Build GRCh38
rs769967363 Chr.11: 35198102-35198102 on Build GRCh38
rs766950689 Chr.11: 35198126-35198126 on Build GRCh38
rs75536042 Chr.11: 35198395-35198395 on Build GRCh38
rs375741863 Chr.11: 35198202-35198202 on Build GRCh38
rs993621962 Chr.11: 35198315-35198315 on Build GRCh38
rs184343364 Chr.11: 35198278-35198278 on Build GRCh38
rs200273155 Chr.11: 35198125-35198125 on Build GRCh38
rs781407455 Chr.11: 35198206-35198206 on Build GRCh38
rs996419076 Chr.11: 35198387-35198387 on Build GRCh38
rs574476713 Chr.11: 35198454-35198454 on Build GRCh38
rs553283536 Chr.11: 35198422-35198424 on Build GRCh38
rs748304500 Chr.11: 35198222-35198222 on Build GRCh38
rs756324513 Chr.11: 35198226-35198226 on Build GRCh38
rs763720891 Chr.11: 35198148-35198148 on Build GRCh38
rs774998866 Chr.11: 35198144-35198144 on Build GRCh38
rs2273690 Chr.11: 35198394-35198394 on Build GRCh38
rs376481612 Chr.11: 35198085-35198085 on Build GRCh38
rs954326731 Chr.11: 35198104-35198104 on Build GRCh38
rs928034042 Chr.11: 35198259-35198259 on Build GRCh38
rs774976608 Chr.11: 35198271-35198271 on Build GRCh38
rs1026897655 Chr.11: 35198324-35198324 on Build GRCh38
rs760393280 Chr.11: 35198215-35198217 on Build GRCh38
rs904317322 Chr.11: 35198385-35198385 on Build GRCh38
rs917077961 Chr.11: 35198492-35198492 on Build GRCh38
rs202081334 Chr.11: 35198093-35198093 on Build GRCh38
rs954911373 Chr.11: 35198333-35198333 on Build GRCh38
rs774704864 Chr.11: 35198242-35198242 on Build GRCh38
rs758233452 Chr.11: 35198483-35198483 on Build GRCh38
rs375829626 Chr.11: 35198237-35198237 on Build GRCh38
rs762054400 Chr.11: 35198234-35198234 on Build GRCh38
rs763263092 Chr.11: 35198114-35198114 on Build GRCh38

View Full Product Details