Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44053064-44053561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
NM_001031854.2 XM_017017714.1 XM_017017715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926936 c.956A>G p.K319R Chr.11: 44053413-44053413 on Build GRCh38
926937 c.964G>T p.G322C Chr.11: 44053421-44053421 on Build GRCh38
7353918 c.948G>C p.E316D Chr.11: 44053068-44053068 on Build GRCh38
6591902 c.961C>T p.R321* Chr.11: 44053418-44053418 on Build GRCh38
6898035 c.962G>A p.R321Q Chr.11: 44053419-44053419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368009999 Chr.11: 44053423-44053423 on Build GRCh38
rs552945468 Chr.11: 44053224-44053224 on Build GRCh38
rs754435890 Chr.11: 44053380-44053380 on Build GRCh38
rs1034529955 Chr.11: 44053123-44053123 on Build GRCh38
rs565746758 Chr.11: 44053170-44053170 on Build GRCh38
rs202078482 Chr.11: 44053413-44053413 on Build GRCh38
rs978052628 Chr.11: 44053223-44053223 on Build GRCh38
rs1029771924 Chr.11: 44053215-44053215 on Build GRCh38
rs929927559 Chr.11: 44053430-44053430 on Build GRCh38
rs751288310 Chr.11: 44053522-44053522 on Build GRCh38
rs957565167 Chr.11: 44053330-44053330 on Build GRCh38
rs745576480 Chr.11: 44053090-44053090 on Build GRCh38
rs751153787 Chr.11: 44053419-44053419 on Build GRCh38
rs769123181 Chr.11: 44053482-44053482 on Build GRCh38
rs950097090 Chr.11: 44053404-44053404 on Build GRCh38
rs780356695 Chr.11: 44053456-44053456 on Build GRCh38
rs576747766 Chr.11: 44053476-44053476 on Build GRCh38
rs370615315 Chr.11: 44053393-44053393 on Build GRCh38
rs998331447 Chr.11: 44053186-44053186 on Build GRCh38
rs374762382 Chr.11: 44053379-44053379 on Build GRCh38
rs142195468 Chr.11: 44053118-44053118 on Build GRCh38
rs765951797 Chr.11: 44053519-44053519 on Build GRCh38
rs376871071 Chr.11: 44053107-44053107 on Build GRCh38
rs368547223 Chr.11: 44053500-44053500 on Build GRCh38
rs779808371 Chr.11: 44053418-44053418 on Build GRCh38
rs375226611 Chr.11: 44053486-44053486 on Build GRCh38
rs188401793 Chr.11: 44053422-44053422 on Build GRCh38
rs745906456 Chr.11: 44053438-44053438 on Build GRCh38
rs762417134 Chr.11: 44053488-44053488 on Build GRCh38
rs776676379 Chr.11: 44053115-44053115 on Build GRCh38
rs557042944 Chr.11: 44053473-44053473 on Build GRCh38
rs181373487 Chr.11: 44053160-44053160 on Build GRCh38
rs544011780 Chr.11: 44053529-44053529 on Build GRCh38
rs746411599 Chr.11: 44053405-44053405 on Build GRCh38
rs758759345 Chr.11: 44053427-44053427 on Build GRCh38
rs775830025 Chr.11: 44053460-44053460 on Build GRCh38
rs556760042 Chr.11: 44053375-44053375 on Build GRCh38
rs757809481 Chr.11: 44053220-44053220 on Build GRCh38
rs770417311 Chr.11: 44053356-44053356 on Build GRCh38
rs762214359 Chr.11: 44053388-44053388 on Build GRCh38
rs894125330 Chr.11: 44053129-44053129 on Build GRCh38
rs781746956 Chr.11: 44053382-44053383 on Build GRCh38
rs925975833 Chr.11: 44053290-44053290 on Build GRCh38
rs371871831 Chr.11: 44053432-44053432 on Build GRCh38
rs1018012118 Chr.11: 44053142-44053142 on Build GRCh38
rs772213674 Chr.11: 44053371-44053371 on Build GRCh38
rs768504187 Chr.11: 44053108-44053108 on Build GRCh38
rs1021851909 Chr.11: 44053205-44053205 on Build GRCh38
rs34476974 Chr.11: 44053219-44053220 on Build GRCh38
rs762604782 Chr.11: 44053511-44053511 on Build GRCh38
rs764855175 Chr.11: 44053391-44053391 on Build GRCh38
rs758159007 Chr.11: 44053415-44053415 on Build GRCh38
rs1039324751 Chr.11: 44053530-44053530 on Build GRCh38
rs7106872 Chr.11: 44053355-44053355 on Build GRCh38
rs184403112 Chr.11: 44053367-44053367 on Build GRCh38
rs373854043 Chr.11: 44053159-44053159 on Build GRCh38
rs374124555 Chr.11: 44053414-44053414 on Build GRCh38
rs780841726 Chr.11: 44053426-44053426 on Build GRCh38
rs758069020 Chr.11: 44053094-44053094 on Build GRCh38
rs769659142 Chr.11: 44053428-44053428 on Build GRCh38
rs919834017 Chr.11: 44053517-44053517 on Build GRCh38
rs749978552 Chr.11: 44053504-44053504 on Build GRCh38
rs377450567 Chr.11: 44053390-44053390 on Build GRCh38
rs774398564 Chr.11: 44053365-44053365 on Build GRCh38
rs779185580 Chr.11: 44053242-44053242 on Build GRCh38
rs756522209 Chr.11: 44053432-44053433 on Build GRCh38
rs929933113 Chr.11: 44053535-44053535 on Build GRCh38
rs941069416 Chr.11: 44053521-44053521 on Build GRCh38

View Full Product Details