Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45924467-45924980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARGE2
Entrez Gene ID
120071
Gene Name
LARGE xylosyl- and glucuronyltransferase 2
Gene Aliases
GYLTL1B, PP5656
UniGene
Hs.86543
Gene Chromosome Location
Chr.11: 45921621-45929096 on Build GRCh38
Transcripts
NM_001300721.1 NM_001300722.1 NM_152312.4 XM_005252787.2 XM_006718140.1 XM_006718141.3 XM_011519886.1 XM_011519888.1 XM_011519889.1 XM_011519890.1 XM_011519891.1 XM_011519892.2 XM_011519893.1 XM_017017173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927096 c.586C>A p.R196S Chr.11: 45924599-45924599 on Build GRCh38
4977164 c.702C>T p.D234D Chr.11: 45924822-45924822 on Build GRCh38
927097 c.632C>T p.S211L Chr.11: 45924645-45924645 on Build GRCh38
1353895 c.701A>G p.D234G Chr.11: 45924821-45924821 on Build GRCh38
2151555 c.543A>C p.L181L Chr.11: 45924556-45924556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923088326 Chr.11: 45924801-45924801 on Build GRCh38
rs776687854 Chr.11: 45924849-45924849 on Build GRCh38
rs775582149 Chr.11: 45924788-45924788 on Build GRCh38
rs772123378 Chr.11: 45924627-45924627 on Build GRCh38
rs763585348 Chr.11: 45924894-45924894 on Build GRCh38
rs752491288 Chr.11: 45924544-45924544 on Build GRCh38
rs370021898 Chr.11: 45924547-45924547 on Build GRCh38
rs776510436 Chr.11: 45924516-45924516 on Build GRCh38
rs769469019 Chr.11: 45924726-45924726 on Build GRCh38
rs747847544 Chr.11: 45924605-45924605 on Build GRCh38
rs200282189 Chr.11: 45924520-45924520 on Build GRCh38
rs370221047 Chr.11: 45924757-45924757 on Build GRCh38
rs377634656 Chr.11: 45924542-45924542 on Build GRCh38
rs746673024 Chr.11: 45924600-45924600 on Build GRCh38
rs200854034 Chr.11: 45924545-45924545 on Build GRCh38
rs201967383 Chr.11: 45924602-45924602 on Build GRCh38
rs779714492 Chr.11: 45924670-45924670 on Build GRCh38
rs759724120 Chr.11: 45924624-45924624 on Build GRCh38
rs573113951 Chr.11: 45924718-45924718 on Build GRCh38
rs762063159 Chr.11: 45924646-45924646 on Build GRCh38
rs368096853 Chr.11: 45924918-45924918 on Build GRCh38
rs144027171 Chr.11: 45924528-45924528 on Build GRCh38
rs553199792 Chr.11: 45924512-45924512 on Build GRCh38
rs57937599 Chr.11: 45924882-45924882 on Build GRCh38
rs972951261 Chr.11: 45924774-45924774 on Build GRCh38
rs1050394623 Chr.11: 45924579-45924579 on Build GRCh38
rs754698392 Chr.11: 45924685-45924685 on Build GRCh38
rs189140907 Chr.11: 45924917-45924917 on Build GRCh38
rs764263826 Chr.11: 45924644-45924644 on Build GRCh38
rs931507165 Chr.11: 45924496-45924496 on Build GRCh38
rs532814183 Chr.11: 45924843-45924843 on Build GRCh38
rs376397657 Chr.11: 45924745-45924745 on Build GRCh38
rs957257937 Chr.11: 45924742-45924742 on Build GRCh38
rs138459534 Chr.11: 45924523-45924523 on Build GRCh38
rs765815732 Chr.11: 45924651-45924651 on Build GRCh38
rs575030437 Chr.11: 45924652-45924652 on Build GRCh38
rs887768124 Chr.11: 45924593-45924593 on Build GRCh38
rs909135231 Chr.11: 45924875-45924875 on Build GRCh38
rs765214518 Chr.11: 45924502-45924527 on Build GRCh38
rs540619048 Chr.11: 45924737-45924737 on Build GRCh38
rs760698896 Chr.11: 45924796-45924796 on Build GRCh38
rs931869979 Chr.11: 45924828-45924828 on Build GRCh38
rs751072656 Chr.11: 45924681-45924681 on Build GRCh38
rs925745022 Chr.11: 45924922-45924922 on Build GRCh38
rs12576236 Chr.11: 45924821-45924821 on Build GRCh38
rs552600788 Chr.11: 45924881-45924881 on Build GRCh38
rs748012529 Chr.11: 45924721-45924721 on Build GRCh38
rs961715128 Chr.11: 45924739-45924739 on Build GRCh38
rs575347152 Chr.11: 45924541-45924541 on Build GRCh38
rs747695475 Chr.11: 45924795-45924795 on Build GRCh38
rs768356234 Chr.11: 45924601-45924601 on Build GRCh38
rs779334648 Chr.11: 45924921-45924921 on Build GRCh38
rs769816841 Chr.11: 45924618-45924618 on Build GRCh38
rs746840891 Chr.11: 45924511-45924511 on Build GRCh38
rs866210861 Chr.11: 45924807-45924807 on Build GRCh38
rs912951142 Chr.11: 45924754-45924754 on Build GRCh38
rs750731745 Chr.11: 45924892-45924892 on Build GRCh38
rs969623944 Chr.11: 45924489-45924489 on Build GRCh38
rs561356468 Chr.11: 45924632-45924632 on Build GRCh38
rs757952553 Chr.11: 45924594-45924594 on Build GRCh38
rs975853295 Chr.11: 45924800-45924800 on Build GRCh38
rs941993848 Chr.11: 45924876-45924876 on Build GRCh38
rs377007291 Chr.11: 45924645-45924645 on Build GRCh38
rs931873733 Chr.11: 45924578-45924578 on Build GRCh38
rs767764254 Chr.11: 45924532-45924532 on Build GRCh38
rs548421543 Chr.11: 45924941-45924941 on Build GRCh38
rs758188398 Chr.11: 45924729-45924730 on Build GRCh38
rs758040500 Chr.11: 45924663-45924663 on Build GRCh38
rs989834067 Chr.11: 45924750-45924750 on Build GRCh38
rs201899150 Chr.11: 45924599-45924599 on Build GRCh38
rs1002854210 Chr.11: 45924731-45924731 on Build GRCh38
rs752907441 Chr.11: 45924538-45924538 on Build GRCh38
rs775261700 Chr.11: 45924506-45924506 on Build GRCh38
rs189478803 Chr.11: 45924919-45924919 on Build GRCh38
rs112067243 Chr.11: 45924700-45924700 on Build GRCh38
rs942761616 Chr.11: 45924905-45924905 on Build GRCh38
rs369644747 Chr.11: 45924499-45924499 on Build GRCh38
rs374032175 Chr.11: 45924529-45924529 on Build GRCh38
rs772989369 Chr.11: 45924619-45924619 on Build GRCh38
rs1023905791 Chr.11: 45924724-45924724 on Build GRCh38
rs373542212 Chr.11: 45924715-45924715 on Build GRCh38
rs760896718 Chr.11: 45924633-45924633 on Build GRCh38
rs560303405 Chr.11: 45924738-45924738 on Build GRCh38
rs1013680782 Chr.11: 45924631-45924631 on Build GRCh38
rs757650680 Chr.11: 45924490-45924490 on Build GRCh38
rs762307296 Chr.11: 45924852-45924852 on Build GRCh38
rs778492429 Chr.11: 45924495-45924495 on Build GRCh38
rs146900807 Chr.11: 45924855-45924855 on Build GRCh38

View Full Product Details