Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46410187-46410686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
UniGene
Hs.654644
Gene Chromosome Location
Chr.11: 46396412-46594069 on Build GRCh38
Transcripts
NM_001267782.1 NM_001267783.1 NM_001300731.1 NM_017749.3 XM_005253009.3 XM_005253011.3 XM_005253014.3 XM_006718259.2 XM_006718260.2 XM_017018007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1980489 c.3202C>T p.R1068W Chr.11: 46410283-46410283 on Build GRCh38
6268396 c.3119C>A p.A1040D Chr.11: 46410366-46410366 on Build GRCh38
5673065 c.3174T>C p.T1058T Chr.11: 46410311-46410311 on Build GRCh38
1188218 c.3146G>T p.W1049L Chr.11: 46410339-46410339 on Build GRCh38
6268395 c.2849C>A p.A950D Chr.11: 46410366-46410366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024401866 Chr.11: 46410650-46410651 on Build GRCh38
rs553082131 Chr.11: 46410634-46410634 on Build GRCh38
rs911838017 Chr.11: 46410603-46410603 on Build GRCh38
rs199528432 Chr.11: 46410260-46410260 on Build GRCh38
rs1033056526 Chr.11: 46410548-46410548 on Build GRCh38
rs773491987 Chr.11: 46410387-46410387 on Build GRCh38
rs767740370 Chr.11: 46410523-46410523 on Build GRCh38
rs529008297 Chr.11: 46410335-46410335 on Build GRCh38
rs528117239 Chr.11: 46410453-46410453 on Build GRCh38
rs372708312 Chr.11: 46410403-46410403 on Build GRCh38
rs776724460 Chr.11: 46410357-46410357 on Build GRCh38
rs901119081 Chr.11: 46410571-46410571 on Build GRCh38
rs767039388 Chr.11: 46410400-46410400 on Build GRCh38
rs765709475 Chr.11: 46410233-46410233 on Build GRCh38
rs969773905 Chr.11: 46410488-46410488 on Build GRCh38
rs758798163 Chr.11: 46410392-46410394 on Build GRCh38
rs749222350 Chr.11: 46410320-46410320 on Build GRCh38
rs1013065455 Chr.11: 46410637-46410637 on Build GRCh38
rs1039540489 Chr.11: 46410558-46410558 on Build GRCh38
rs752010183 Chr.11: 46410283-46410283 on Build GRCh38
rs774112609 Chr.11: 46410342-46410342 on Build GRCh38
rs896637050 Chr.11: 46410384-46410384 on Build GRCh38
rs755580265 Chr.11: 46410311-46410311 on Build GRCh38
rs765394689 Chr.11: 46410385-46410385 on Build GRCh38
rs909626568 Chr.11: 46410491-46410491 on Build GRCh38
rs900564623 Chr.11: 46410289-46410289 on Build GRCh38
rs761894991 Chr.11: 46410360-46410360 on Build GRCh38
rs777325811 Chr.11: 46410312-46410312 on Build GRCh38
rs1049493515 Chr.11: 46410298-46410298 on Build GRCh38
rs549140079 Chr.11: 46410402-46410402 on Build GRCh38
rs370242228 Chr.11: 46410567-46410567 on Build GRCh38
rs750980890 Chr.11: 46410254-46410254 on Build GRCh38
rs150768979 Chr.11: 46410536-46410536 on Build GRCh38
rs1023823738 Chr.11: 46410424-46410424 on Build GRCh38
rs538274090 Chr.11: 46410240-46410240 on Build GRCh38
rs989133219 Chr.11: 46410602-46410602 on Build GRCh38
rs1041721420 Chr.11: 46410609-46410609 on Build GRCh38
rs745673960 Chr.11: 46410354-46410354 on Build GRCh38
rs930694057 Chr.11: 46410654-46410654 on Build GRCh38
rs746864991 Chr.11: 46410282-46410282 on Build GRCh38
rs1005534886 Chr.11: 46410344-46410344 on Build GRCh38
rs573243913 Chr.11: 46410663-46410663 on Build GRCh38
rs944582351 Chr.11: 46410223-46410223 on Build GRCh38
rs561421424 Chr.11: 46410213-46410213 on Build GRCh38
rs780727462 Chr.11: 46410297-46410297 on Build GRCh38
rs747650808 Chr.11: 46410310-46410310 on Build GRCh38
rs551073733 Chr.11: 46410472-46410472 on Build GRCh38
rs955767147 Chr.11: 46410582-46410582 on Build GRCh38
rs763522288 Chr.11: 46410395-46410395 on Build GRCh38
rs573148689 Chr.11: 46410232-46410232 on Build GRCh38
rs141278389 Chr.11: 46410512-46410512 on Build GRCh38
rs567078349 Chr.11: 46410622-46410622 on Build GRCh38
rs764549693 Chr.11: 46410249-46410249 on Build GRCh38
rs1000192247 Chr.11: 46410623-46410623 on Build GRCh38
rs200038782 Chr.11: 46410281-46410281 on Build GRCh38
rs180716764 Chr.11: 46410414-46410414 on Build GRCh38
rs950287353 Chr.11: 46410322-46410322 on Build GRCh38
rs79928554 Chr.11: 46410627-46410627 on Build GRCh38
rs1048586895 Chr.11: 46410458-46410458 on Build GRCh38
rs149732166 Chr.11: 46410286-46410286 on Build GRCh38

View Full Product Details