Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46493525-46494037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBRA1
Entrez Gene ID
55626
Gene Name
autophagy and beclin 1 regulator 1
Gene Aliases
DCAF3, WDR94
UniGene
Hs.654644
Gene Chromosome Location
Chr.11: 46396412-46594069 on Build GRCh38
Transcripts
NM_001267782.1 NM_001267783.1 NM_001300731.1 NM_017749.3 XM_005253009.3 XM_005253011.3 XM_005253014.3 XM_006718259.2 XM_006718260.2 XM_017018007.1 XM_017018008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700074 c.2245C>T p.L749L Chr.11: 46493614-46493614 on Build GRCh38
6239213 c.2457G>T p.G819G Chr.11: 46493672-46493672 on Build GRCh38
5500705 c.2251+2T>C p.? Chr.11: 46493606-46493606 on Build GRCh38
6239212 c.2187G>T p.G729G Chr.11: 46493672-46493672 on Build GRCh38
3700075 c.2515C>T p.L839L Chr.11: 46493614-46493614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988916805 Chr.11: 46493780-46493780 on Build GRCh38
rs998444175 Chr.11: 46493804-46493804 on Build GRCh38
rs1017886336 Chr.11: 46493862-46493862 on Build GRCh38
rs991067655 Chr.11: 46493982-46493982 on Build GRCh38
rs73467919 Chr.11: 46493854-46493854 on Build GRCh38
rs879844299 Chr.11: 46493976-46493976 on Build GRCh38
rs760677728 Chr.11: 46493571-46493571 on Build GRCh38
rs916754294 Chr.11: 46494005-46494005 on Build GRCh38
rs758077019 Chr.11: 46493646-46493646 on Build GRCh38
rs748070184 Chr.11: 46493715-46493715 on Build GRCh38
rs185441245 Chr.11: 46493977-46493977 on Build GRCh38
rs757907400 Chr.11: 46493615-46493615 on Build GRCh38
rs1050221342 Chr.11: 46494011-46494011 on Build GRCh38
rs201268663 Chr.11: 46493717-46493717 on Build GRCh38
rs746460100 Chr.11: 46493663-46493663 on Build GRCh38
rs937792887 Chr.11: 46493881-46493881 on Build GRCh38
rs890413859 Chr.11: 46493845-46493845 on Build GRCh38
rs750869328 Chr.11: 46493633-46493633 on Build GRCh38
rs375784286 Chr.11: 46493610-46493610 on Build GRCh38
rs375435986 Chr.11: 46493730-46493730 on Build GRCh38
rs973908641 Chr.11: 46493892-46493892 on Build GRCh38
rs956191105 Chr.11: 46493754-46493754 on Build GRCh38
rs576492478 Chr.11: 46493966-46493966 on Build GRCh38
rs917634438 Chr.11: 46493949-46493949 on Build GRCh38
rs370409176 Chr.11: 46493753-46493753 on Build GRCh38
rs564271931 Chr.11: 46493830-46493830 on Build GRCh38
rs926307663 Chr.11: 46493856-46493856 on Build GRCh38
rs904210848 Chr.11: 46493798-46493798 on Build GRCh38
rs571743691 Chr.11: 46493651-46493651 on Build GRCh38
rs1031552536 Chr.11: 46493824-46493824 on Build GRCh38
rs767068601 Chr.11: 46493567-46493567 on Build GRCh38
rs765666115 Chr.11: 46493620-46493620 on Build GRCh38
rs1042790440 Chr.11: 46493772-46493772 on Build GRCh38
rs767772554 Chr.11: 46493562-46493562 on Build GRCh38
rs376203986 Chr.11: 46493612-46493612 on Build GRCh38
rs371129735 Chr.11: 46493712-46493712 on Build GRCh38
rs1036968651 Chr.11: 46493554-46493554 on Build GRCh38
rs950696660 Chr.11: 46493945-46493945 on Build GRCh38
rs1027704303 Chr.11: 46493898-46493898 on Build GRCh38
rs764608295 Chr.11: 46493597-46493597 on Build GRCh38
rs769758770 Chr.11: 46493727-46493727 on Build GRCh38
rs979070707 Chr.11: 46493855-46493855 on Build GRCh38
rs995418292 Chr.11: 46494009-46494009 on Build GRCh38
rs541000946 Chr.11: 46493776-46493776 on Build GRCh38
rs772967520 Chr.11: 46493728-46493728 on Build GRCh38
rs772827490 Chr.11: 46493726-46493727 on Build GRCh38
rs199820988 Chr.11: 46493864-46493864 on Build GRCh38
rs181239163 Chr.11: 46493570-46493570 on Build GRCh38

View Full Product Details