Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47723909-47724457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP4
Entrez Gene ID
23360
Gene Name
formin binding protein 4
Gene Aliases
FBP30
UniGene
Hs.6834
Gene Chromosome Location
Chr.11: 47716510-47767478 on Build GRCh38
Transcripts
NM_001318339.1 NM_015308.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7145656 c.2340C>T p.I780I Chr.11: 47724152-47724152 on Build GRCh38
4927217 c.2336C>T p.T779I Chr.11: 47724156-47724156 on Build GRCh38
1981954 c.2352_2354delTTC p.S785delS Chr.11: 47724138-47724140 on Build GRCh38
5038781 c.2322_2324delTTC p.S774_S775>R Chr.11: 47724168-47724170 on Build GRCh38
6665845 c.2453C>T p.A818V Chr.11: 47724039-47724039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910668300 Chr.11: 47724390-47724390 on Build GRCh38
rs577349308 Chr.11: 47724203-47724203 on Build GRCh38
rs977999986 Chr.11: 47724363-47724363 on Build GRCh38
rs907559912 Chr.11: 47724343-47724343 on Build GRCh38
rs773910814 Chr.11: 47723978-47723978 on Build GRCh38
rs550950890 Chr.11: 47724298-47724298 on Build GRCh38
rs763323682 Chr.11: 47724010-47724010 on Build GRCh38
rs753991243 Chr.11: 47724220-47724221 on Build GRCh38
rs187611974 Chr.11: 47724096-47724096 on Build GRCh38
rs899379289 Chr.11: 47724409-47724409 on Build GRCh38
rs947212616 Chr.11: 47724272-47724272 on Build GRCh38
rs749642462 Chr.11: 47724051-47724051 on Build GRCh38
rs769883159 Chr.11: 47724154-47724154 on Build GRCh38
rs773887179 Chr.11: 47723988-47723988 on Build GRCh38
rs933346244 Chr.11: 47724372-47724372 on Build GRCh38
rs772007250 Chr.11: 47723996-47723996 on Build GRCh38
rs954849128 Chr.11: 47724197-47724197 on Build GRCh38
rs778476742 Chr.11: 47724055-47724055 on Build GRCh38
rs1052295090 Chr.11: 47724075-47724075 on Build GRCh38
rs899449124 Chr.11: 47724122-47724122 on Build GRCh38
rs539771522 Chr.11: 47724301-47724301 on Build GRCh38
rs763136740 Chr.11: 47724226-47724226 on Build GRCh38
rs761636132 Chr.11: 47724140-47724140 on Build GRCh38
rs779644385 Chr.11: 47724017-47724017 on Build GRCh38
rs745401670 Chr.11: 47724062-47724062 on Build GRCh38
rs776553642 Chr.11: 47724133-47724133 on Build GRCh38
rs138653244 Chr.11: 47724144-47724144 on Build GRCh38
rs914473623 Chr.11: 47724266-47724266 on Build GRCh38
rs374015807 Chr.11: 47724011-47724011 on Build GRCh38
rs768642417 Chr.11: 47724433-47724433 on Build GRCh38
rs773762101 Chr.11: 47724227-47724227 on Build GRCh38
rs746494841 Chr.11: 47724418-47724418 on Build GRCh38
rs192814501 Chr.11: 47724187-47724187 on Build GRCh38
rs996318487 Chr.11: 47724169-47724169 on Build GRCh38
rs779672201 Chr.11: 47724216-47724216 on Build GRCh38
rs541830758 Chr.11: 47724376-47724376 on Build GRCh38
rs761108896 Chr.11: 47724265-47724265 on Build GRCh38
rs757965098 Chr.11: 47724208-47724208 on Build GRCh38
rs761304377 Chr.11: 47724179-47724179 on Build GRCh38
rs552486053 Chr.11: 47723944-47723944 on Build GRCh38
rs750112750 Chr.11: 47724206-47724206 on Build GRCh38
rs766723434 Chr.11: 47724142-47724142 on Build GRCh38
rs752701873 Chr.11: 47724160-47724160 on Build GRCh38
rs768463293 Chr.11: 47724120-47724120 on Build GRCh38
rs368064726 Chr.11: 47724007-47724007 on Build GRCh38
rs987437292 Chr.11: 47723951-47723951 on Build GRCh38
rs755052992 Chr.11: 47724014-47724014 on Build GRCh38
rs943541318 Chr.11: 47724402-47724402 on Build GRCh38
rs766563479 Chr.11: 47724138-47724140 on Build GRCh38
rs35040940 Chr.11: 47724112-47724112 on Build GRCh38
rs34242224 Chr.11: 47724037-47724037 on Build GRCh38
rs759720318 Chr.11: 47724147-47724147 on Build GRCh38
rs767679455 Chr.11: 47724159-47724159 on Build GRCh38
rs923932565 Chr.11: 47723959-47723959 on Build GRCh38
rs1037839005 Chr.11: 47724421-47724421 on Build GRCh38
rs751696301 Chr.11: 47724012-47724012 on Build GRCh38
rs1025946011 Chr.11: 47724166-47724166 on Build GRCh38
rs771566617 Chr.11: 47724080-47724080 on Build GRCh38
rs756551702 Chr.11: 47724042-47724042 on Build GRCh38
rs764506853 Chr.11: 47724182-47724182 on Build GRCh38
rs924586267 Chr.11: 47724370-47724370 on Build GRCh38
rs889278751 Chr.11: 47724084-47724084 on Build GRCh38
rs756905405 Chr.11: 47724053-47724053 on Build GRCh38
rs373208800 Chr.11: 47724262-47724262 on Build GRCh38
rs771174354 Chr.11: 47724344-47724344 on Build GRCh38
rs778252868 Chr.11: 47724050-47724050 on Build GRCh38
rs970815884 Chr.11: 47724181-47724181 on Build GRCh38
rs755173529 Chr.11: 47724426-47724427 on Build GRCh38
rs182652767 Chr.11: 47724015-47724015 on Build GRCh38
rs745307581 Chr.11: 47723980-47723980 on Build GRCh38
rs959216027 Chr.11: 47724245-47724245 on Build GRCh38
rs556435907 Chr.11: 47724327-47724327 on Build GRCh38
rs757565867 Chr.11: 47724196-47724196 on Build GRCh38
rs568611961 Chr.11: 47724239-47724239 on Build GRCh38
rs370575997 Chr.11: 47724009-47724009 on Build GRCh38
rs979927484 Chr.11: 47724284-47724284 on Build GRCh38
rs576069234 Chr.11: 47724346-47724346 on Build GRCh38
rs940335821 Chr.11: 47724369-47724369 on Build GRCh38
rs927112323 Chr.11: 47724286-47724286 on Build GRCh38
rs745648799 Chr.11: 47723997-47723997 on Build GRCh38

View Full Product Details