Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57744285-57744794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5753727 c.1652C>A p.P551H Chr.11: 57744621-57744621 on Build GRCh38
467005 c.1572G>A p.T524T Chr.11: 57744701-57744701 on Build GRCh38
4755976 c.1565G>A p.C522Y Chr.11: 57744708-57744708 on Build GRCh38
1585825 c.1755T>C p.L585L Chr.11: 57744518-57744518 on Build GRCh38
1185033 c.1871G>A p.G624E Chr.11: 57744402-57744402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953548753 Chr.11: 57744634-57744634 on Build GRCh38
rs755818808 Chr.11: 57744447-57744447 on Build GRCh38
rs930604935 Chr.11: 57744702-57744702 on Build GRCh38
rs751493867 Chr.11: 57744368-57744368 on Build GRCh38
rs970341531 Chr.11: 57744524-57744524 on Build GRCh38
rs996565572 Chr.11: 57744586-57744586 on Build GRCh38
rs920611068 Chr.11: 57744701-57744701 on Build GRCh38
rs754842711 Chr.11: 57744404-57744404 on Build GRCh38
rs548447040 Chr.11: 57744356-57744356 on Build GRCh38
rs750275119 Chr.11: 57744346-57744346 on Build GRCh38
rs755552092 Chr.11: 57744715-57744715 on Build GRCh38
rs751295702 Chr.11: 57744663-57744663 on Build GRCh38
rs962349130 Chr.11: 57744695-57744695 on Build GRCh38
rs556095073 Chr.11: 57744573-57744573 on Build GRCh38
rs757462127 Chr.11: 57744595-57744595 on Build GRCh38
rs755508233 Chr.11: 57744516-57744516 on Build GRCh38
rs561617168 Chr.11: 57744753-57744753 on Build GRCh38
rs538231381 Chr.11: 57744544-57744544 on Build GRCh38
rs375829022 Chr.11: 57744675-57744675 on Build GRCh38
rs772307955 Chr.11: 57744464-57744464 on Build GRCh38
rs1023254314 Chr.11: 57744498-57744498 on Build GRCh38
rs922688328 Chr.11: 57744655-57744655 on Build GRCh38
rs939416519 Chr.11: 57744738-57744738 on Build GRCh38
rs760620984 Chr.11: 57744744-57744745 on Build GRCh38
rs148470181 Chr.11: 57744741-57744741 on Build GRCh38
rs773268461 Chr.11: 57744736-57744742 on Build GRCh38
rs956392943 Chr.11: 57744583-57744583 on Build GRCh38
rs899145760 Chr.11: 57744411-57744411 on Build GRCh38
rs536181991 Chr.11: 57744332-57744332 on Build GRCh38
rs374727411 Chr.11: 57744434-57744434 on Build GRCh38
rs972704078 Chr.11: 57744700-57744700 on Build GRCh38
rs780691637 Chr.11: 57744423-57744423 on Build GRCh38
rs759490656 Chr.11: 57744678-57744678 on Build GRCh38
rs751365973 Chr.11: 57744504-57744504 on Build GRCh38
rs1049673631 Chr.11: 57744331-57744331 on Build GRCh38
rs955409581 Chr.11: 57744658-57744658 on Build GRCh38
rs746289388 Chr.11: 57744459-57744459 on Build GRCh38
rs892282536 Chr.11: 57744759-57744759 on Build GRCh38
rs183196273 Chr.11: 57744745-57744745 on Build GRCh38
rs769240391 Chr.11: 57744506-57744506 on Build GRCh38
rs767400209 Chr.11: 57744686-57744686 on Build GRCh38
rs926655906 Chr.11: 57744719-57744719 on Build GRCh38
rs773294188 Chr.11: 57744581-57744581 on Build GRCh38
rs776883145 Chr.11: 57744542-57744542 on Build GRCh38
rs558312760 Chr.11: 57744347-57744347 on Build GRCh38
rs781457115 Chr.11: 57744645-57744645 on Build GRCh38
rs367567782 Chr.11: 57744365-57744365 on Build GRCh38
rs879263915 Chr.11: 57744417-57744417 on Build GRCh38
rs941241719 Chr.11: 57744363-57744363 on Build GRCh38
rs987696038 Chr.11: 57744660-57744660 on Build GRCh38
rs1027994578 Chr.11: 57744616-57744616 on Build GRCh38
rs988936645 Chr.11: 57744633-57744633 on Build GRCh38
rs780503922 Chr.11: 57744475-57744475 on Build GRCh38
rs190995804 Chr.11: 57744480-57744480 on Build GRCh38
rs940935933 Chr.11: 57744393-57744393 on Build GRCh38
rs538714959 Chr.11: 57744661-57744661 on Build GRCh38
rs770103270 Chr.11: 57744578-57744578 on Build GRCh38
rs761770489 Chr.11: 57744567-57744567 on Build GRCh38
rs777258436 Chr.11: 57744455-57744455 on Build GRCh38
rs752241282 Chr.11: 57744712-57744712 on Build GRCh38
rs912107237 Chr.11: 57744391-57744391 on Build GRCh38
rs905043010 Chr.11: 57744440-57744440 on Build GRCh38
rs577727393 Chr.11: 57744604-57744604 on Build GRCh38
rs777603099 Chr.11: 57744426-57744426 on Build GRCh38

View Full Product Details