Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57745091-57745596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1354816 c.1127C>A p.P376H Chr.11: 57745146-57745146 on Build GRCh38
4638682 c.1013G>A p.R338Q Chr.11: 57745260-57745260 on Build GRCh38
4977847 c.721G>T p.D241Y Chr.11: 57745552-57745552 on Build GRCh38
5003323 c.677G>T p.S226I Chr.11: 57745596-57745596 on Build GRCh38
1354818 c.912T>C p.N304N Chr.11: 57745361-57745361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368415279 Chr.11: 57745389-57745389 on Build GRCh38
rs12787776 Chr.11: 57745515-57745515 on Build GRCh38
rs952029779 Chr.11: 57745117-57745117 on Build GRCh38
rs370489662 Chr.11: 57745179-57745179 on Build GRCh38
rs755434207 Chr.11: 57745502-57745502 on Build GRCh38
rs745718258 Chr.11: 57745401-57745401 on Build GRCh38
rs549782162 Chr.11: 57745452-57745452 on Build GRCh38
rs753344337 Chr.11: 57745231-57745231 on Build GRCh38
rs12794987 Chr.11: 57745572-57745572 on Build GRCh38
rs536677168 Chr.11: 57745361-57745361 on Build GRCh38
rs979267290 Chr.11: 57745559-57745559 on Build GRCh38
rs201912857 Chr.11: 57745218-57745218 on Build GRCh38
rs759077785 Chr.11: 57745315-57745315 on Build GRCh38
rs926603423 Chr.11: 57745132-57745132 on Build GRCh38
rs945169599 Chr.11: 57745229-57745229 on Build GRCh38
rs569933884 Chr.11: 57745423-57745423 on Build GRCh38
rs12794985 Chr.11: 57745566-57745566 on Build GRCh38
rs776048962 Chr.11: 57745527-57745527 on Build GRCh38
rs992270250 Chr.11: 57745487-57745487 on Build GRCh38
rs529887501 Chr.11: 57745360-57745360 on Build GRCh38
rs770537834 Chr.11: 57745249-57745249 on Build GRCh38
rs1006957962 Chr.11: 57745387-57745387 on Build GRCh38
rs775300523 Chr.11: 57745446-57745446 on Build GRCh38
rs532266525 Chr.11: 57745130-57745130 on Build GRCh38
rs1010405978 Chr.11: 57745357-57745357 on Build GRCh38
rs764670484 Chr.11: 57745333-57745333 on Build GRCh38
rs1015728374 Chr.11: 57745345-57745345 on Build GRCh38
rs898003403 Chr.11: 57745346-57745346 on Build GRCh38
rs781781795 Chr.11: 57745339-57745339 on Build GRCh38
rs755384035 Chr.11: 57745336-57745336 on Build GRCh38
rs756584253 Chr.11: 57745366-57745366 on Build GRCh38
rs771179268 Chr.11: 57745390-57745390 on Build GRCh38
rs913936202 Chr.11: 57745541-57745541 on Build GRCh38
rs1007993258 Chr.11: 57745344-57745344 on Build GRCh38
rs78162678 Chr.11: 57745368-57745368 on Build GRCh38
rs754369746 Chr.11: 57745297-57745297 on Build GRCh38
rs760296393 Chr.11: 57745140-57745140 on Build GRCh38
rs749346935 Chr.11: 57745386-57745386 on Build GRCh38
rs774530773 Chr.11: 57745398-57745398 on Build GRCh38
rs765062929 Chr.11: 57745216-57745218 on Build GRCh38
rs547492085 Chr.11: 57745135-57745135 on Build GRCh38
rs994035961 Chr.11: 57745350-57745350 on Build GRCh38
rs764057238 Chr.11: 57745491-57745491 on Build GRCh38
rs1056933437 Chr.11: 57745149-57745149 on Build GRCh38
rs375878108 Chr.11: 57745305-57745305 on Build GRCh38
rs945477818 Chr.11: 57745542-57745542 on Build GRCh38
rs140102739 Chr.11: 57745448-57745448 on Build GRCh38
rs903544657 Chr.11: 57745246-57745246 on Build GRCh38
rs903818754 Chr.11: 57745260-57745260 on Build GRCh38
rs752561051 Chr.11: 57745123-57745123 on Build GRCh38

View Full Product Details