Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57751352-57751860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

SNP Detail

SNP ID Chromosome Location
rs567390921 Chr.11: 57751446-57751446 on Build GRCh38
rs191155072 Chr.11: 57751504-57751504 on Build GRCh38
rs538901546 Chr.11: 57751553-57751553 on Build GRCh38
rs866899860 Chr.11: 57751646-57751646 on Build GRCh38
rs941067505 Chr.11: 57751471-57751471 on Build GRCh38
rs78109339 Chr.11: 57751379-57751379 on Build GRCh38
rs1043373620 Chr.11: 57751816-57751816 on Build GRCh38
rs987697292 Chr.11: 57751450-57751450 on Build GRCh38
rs956021017 Chr.11: 57751638-57751644 on Build GRCh38
rs776263553 Chr.11: 57751526-57751528 on Build GRCh38
rs573170620 Chr.11: 57751627-57751627 on Build GRCh38
rs920715065 Chr.11: 57751572-57751572 on Build GRCh38
rs557891124 Chr.11: 57751624-57751624 on Build GRCh38
rs773307437 Chr.11: 57751420-57751420 on Build GRCh38
rs1038109564 Chr.11: 57751777-57751777 on Build GRCh38
rs952156370 Chr.11: 57751586-57751586 on Build GRCh38
rs911032253 Chr.11: 57751733-57751733 on Build GRCh38
rs549957574 Chr.11: 57751510-57751510 on Build GRCh38
rs896057250 Chr.11: 57751811-57751811 on Build GRCh38
rs1000803936 Chr.11: 57751429-57751429 on Build GRCh38
rs551162410 Chr.11: 57751411-57751411 on Build GRCh38
rs1000208250 Chr.11: 57751589-57751589 on Build GRCh38
rs913580887 Chr.11: 57751723-57751723 on Build GRCh38
rs1038057181 Chr.11: 57751487-57751487 on Build GRCh38
rs941121158 Chr.11: 57751744-57751744 on Build GRCh38
rs1021967951 Chr.11: 57751380-57751380 on Build GRCh38
rs945006028 Chr.11: 57751746-57751746 on Build GRCh38
rs1056098790 Chr.11: 57751807-57751807 on Build GRCh38
rs892109821 Chr.11: 57751377-57751377 on Build GRCh38
rs929618760 Chr.11: 57751800-57751800 on Build GRCh38
rs903627240 Chr.11: 57751817-57751817 on Build GRCh38
rs955279376 Chr.11: 57751720-57751720 on Build GRCh38
rs1033001969 Chr.11: 57751625-57751625 on Build GRCh38
rs534072262 Chr.11: 57751641-57751641 on Build GRCh38
rs1023091897 Chr.11: 57751560-57751560 on Build GRCh38
rs996589722 Chr.11: 57751836-57751836 on Build GRCh38
rs184780764 Chr.11: 57751695-57751695 on Build GRCh38
rs972144041 Chr.11: 57751542-57751542 on Build GRCh38
rs1026979272 Chr.11: 57751525-57751525 on Build GRCh38
rs960683859 Chr.11: 57751473-57751473 on Build GRCh38
rs981042723 Chr.11: 57751612-57751612 on Build GRCh38
rs766626062 Chr.11: 57751552-57751553 on Build GRCh38
rs1029148290 Chr.11: 57751439-57751439 on Build GRCh38
rs549049051 Chr.11: 57751402-57751402 on Build GRCh38
rs189145958 Chr.11: 57751837-57751837 on Build GRCh38
rs962004509 Chr.11: 57751523-57751523 on Build GRCh38
rs904694198 Chr.11: 57751410-57751410 on Build GRCh38
rs918297119 Chr.11: 57751784-57751784 on Build GRCh38
rs777005829 Chr.11: 57751700-57751700 on Build GRCh38
rs571575050 Chr.11: 57751519-57751519 on Build GRCh38

View Full Product Details