Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59841196-59841706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIF
Entrez Gene ID
2694
Gene Name
gastric intrinsic factor
Gene Aliases
IF, IFMH, INF, TCN3
UniGene
Hs.110014
Gene Chromosome Location
Chr.11: 59829268-59845529 on Build GRCh38
Transcripts
NM_005142.2 XM_011544939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929123 c.555_557delCAA p.N185delN Chr.11: 59841279-59841281 on Build GRCh38
5361119 c.589T>C p.S197P Chr.11: 59841247-59841247 on Build GRCh38
1704072 c.595T>G p.F199V Chr.11: 59841241-59841241 on Build GRCh38
6037440 c.513C>T p.D171D Chr.11: 59841323-59841323 on Build GRCh38
1746363 c.519A>C p.G173G Chr.11: 59841317-59841317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762598409 Chr.11: 59841291-59841292 on Build GRCh38
rs889933603 Chr.11: 59841521-59841521 on Build GRCh38
rs754526451 Chr.11: 59841601-59841601 on Build GRCh38
rs746791682 Chr.11: 59841515-59841515 on Build GRCh38
rs148960211 Chr.11: 59841341-59841341 on Build GRCh38
rs777870219 Chr.11: 59841493-59841493 on Build GRCh38
rs377009224 Chr.11: 59841339-59841339 on Build GRCh38
rs984041418 Chr.11: 59841508-59841508 on Build GRCh38
rs888449656 Chr.11: 59841599-59841599 on Build GRCh38
rs529329032 Chr.11: 59841581-59841581 on Build GRCh38
rs761501191 Chr.11: 59841279-59841281 on Build GRCh38
rs1002877562 Chr.11: 59841299-59841299 on Build GRCh38
rs543770620 Chr.11: 59841447-59841447 on Build GRCh38
rs541810303 Chr.11: 59841484-59841484 on Build GRCh38
rs563535498 Chr.11: 59841479-59841479 on Build GRCh38
rs907299949 Chr.11: 59841568-59841568 on Build GRCh38
rs886048404 Chr.11: 59841312-59841312 on Build GRCh38
rs936902620 Chr.11: 59841480-59841480 on Build GRCh38
rs1050451079 Chr.11: 59841490-59841490 on Build GRCh38
rs965048683 Chr.11: 59841462-59841462 on Build GRCh38
rs867085394 Chr.11: 59841531-59841531 on Build GRCh38
rs747647693 Chr.11: 59841364-59841364 on Build GRCh38
rs761773560 Chr.11: 59841322-59841322 on Build GRCh38
rs775795949 Chr.11: 59841268-59841268 on Build GRCh38
rs757773932 Chr.11: 59841368-59841368 on Build GRCh38
rs780662774 Chr.11: 59841363-59841363 on Build GRCh38
rs916410310 Chr.11: 59841522-59841522 on Build GRCh38
rs985191245 Chr.11: 59841435-59841435 on Build GRCh38
rs375980909 Chr.11: 59841256-59841256 on Build GRCh38
rs540701871 Chr.11: 59841553-59841553 on Build GRCh38
rs751175352 Chr.11: 59841349-59841349 on Build GRCh38
rs143729995 Chr.11: 59841527-59841527 on Build GRCh38
rs760980454 Chr.11: 59841287-59841287 on Build GRCh38
rs78681553 Chr.11: 59841592-59841592 on Build GRCh38
rs764229455 Chr.11: 59841296-59841296 on Build GRCh38
rs375486811 Chr.11: 59841489-59841489 on Build GRCh38
rs555776939 Chr.11: 59841340-59841340 on Build GRCh38
rs938637433 Chr.11: 59841569-59841569 on Build GRCh38
rs977766435 Chr.11: 59841467-59841467 on Build GRCh38
rs1034025130 Chr.11: 59841677-59841677 on Build GRCh38
rs755614414 Chr.11: 59841535-59841535 on Build GRCh38
rs774742217 Chr.11: 59841221-59841221 on Build GRCh38
rs897285615 Chr.11: 59841554-59841554 on Build GRCh38
rs1021778584 Chr.11: 59841393-59841393 on Build GRCh38
rs370957853 Chr.11: 59841353-59841353 on Build GRCh38
rs994163287 Chr.11: 59841590-59841590 on Build GRCh38
rs954389483 Chr.11: 59841398-59841398 on Build GRCh38
rs1006111793 Chr.11: 59841653-59841653 on Build GRCh38
rs746282268 Chr.11: 59841228-59841228 on Build GRCh38
rs776872892 Chr.11: 59841313-59841313 on Build GRCh38

View Full Product Details