Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64232338-64232833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC4
Entrez Gene ID
3338
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Gene Aliases
DANJC4, HSPF2, MCG18
UniGene
Hs.172847
Gene Chromosome Location
Chr.11: 64230281-64234281 on Build GRCh38
Transcripts
NM_001307980.1 NM_001307981.1 NM_005528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165594 c.333_336delAAAC p.Q114fs*>21 Chr.11: 64232794-64232797 on Build GRCh38
7400822 c.310C>T p.P104S Chr.11: 64232559-64232559 on Build GRCh38
4146047 c.281G>C p.S94T Chr.11: 64232530-64232530 on Build GRCh38
689746 c.91C>A p.L31M Chr.11: 64232463-64232463 on Build GRCh38
7321282 c.241G>A p.E81K Chr.11: 64232490-64232490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048048095 Chr.11: 64232572-64232572 on Build GRCh38
rs764043069 Chr.11: 64232488-64232488 on Build GRCh38
rs979210242 Chr.11: 64232450-64232450 on Build GRCh38
rs1000579773 Chr.11: 64232714-64232714 on Build GRCh38
rs747615523 Chr.11: 64232750-64232750 on Build GRCh38
rs750527086 Chr.11: 64232546-64232546 on Build GRCh38
rs773308185 Chr.11: 64232669-64232669 on Build GRCh38
rs752821502 Chr.11: 64232728-64232728 on Build GRCh38
rs763012788 Chr.11: 64232427-64232427 on Build GRCh38
rs748902639 Chr.11: 64232625-64232625 on Build GRCh38
rs117464526 Chr.11: 64232547-64232547 on Build GRCh38
rs764733432 Chr.11: 64232414-64232414 on Build GRCh38
rs754624376 Chr.11: 64232747-64232747 on Build GRCh38
rs769249584 Chr.11: 64232465-64232465 on Build GRCh38
rs370287965 Chr.11: 64232530-64232530 on Build GRCh38
rs768195270 Chr.11: 64232629-64232629 on Build GRCh38
rs745534264 Chr.11: 64232473-64232473 on Build GRCh38
rs766341609 Chr.11: 64232505-64232505 on Build GRCh38
rs764543996 Chr.11: 64232730-64232730 on Build GRCh38
rs747323857 Chr.11: 64232583-64232583 on Build GRCh38
rs950719391 Chr.11: 64232416-64232416 on Build GRCh38
rs781464158 Chr.11: 64232741-64232741 on Build GRCh38
rs528015044 Chr.11: 64232806-64232806 on Build GRCh38
rs748467006 Chr.11: 64232467-64232467 on Build GRCh38
rs375366918 Chr.11: 64232760-64232760 on Build GRCh38
rs113915915 Chr.11: 64232615-64232615 on Build GRCh38
rs759735167 Chr.11: 64232714-64232714 on Build GRCh38
rs776200378 Chr.11: 64232472-64232472 on Build GRCh38
rs767413232 Chr.11: 64232543-64232543 on Build GRCh38
rs541349768 Chr.11: 64232479-64232479 on Build GRCh38
rs201258154 Chr.11: 64232489-64232489 on Build GRCh38
rs776855176 Chr.11: 64232713-64232713 on Build GRCh38
rs200012234 Chr.11: 64232738-64232738 on Build GRCh38
rs879131289 Chr.11: 64232419-64232419 on Build GRCh38
rs899358431 Chr.11: 64232553-64232553 on Build GRCh38
rs760616442 Chr.11: 64232787-64232790 on Build GRCh38
rs377031581 Chr.11: 64232379-64232379 on Build GRCh38
rs766577923 Chr.11: 64232555-64232555 on Build GRCh38
rs986889001 Chr.11: 64232387-64232387 on Build GRCh38
rs569307400 Chr.11: 64232773-64232773 on Build GRCh38
rs777790627 Chr.11: 64232758-64232758 on Build GRCh38
rs770623397 Chr.11: 64232395-64232395 on Build GRCh38
rs753521926 Chr.11: 64232746-64232746 on Build GRCh38
rs753719279 Chr.11: 64232804-64232805 on Build GRCh38
rs771742960 Chr.11: 64232752-64232752 on Build GRCh38
rs755019048 Chr.11: 64232574-64232574 on Build GRCh38
rs753541691 Chr.11: 64232794-64232797 on Build GRCh38
rs374099725 Chr.11: 64232523-64232523 on Build GRCh38
rs779129165 Chr.11: 64232466-64232466 on Build GRCh38
rs375824405 Chr.11: 64232513-64232513 on Build GRCh38
rs754043668 Chr.11: 64232571-64232571 on Build GRCh38
rs753191459 Chr.11: 64232579-64232579 on Build GRCh38
rs763763877 Chr.11: 64232437-64232437 on Build GRCh38
rs150439143 Chr.11: 64232642-64232642 on Build GRCh38
rs996675849 Chr.11: 64232646-64232646 on Build GRCh38
rs774284402 Chr.11: 64232533-64232533 on Build GRCh38
rs751818674 Chr.11: 64232757-64232757 on Build GRCh38
rs770636275 Chr.11: 64232703-64232703 on Build GRCh38
rs571900284 Chr.11: 64232458-64232458 on Build GRCh38
rs980693338 Chr.11: 64232793-64232793 on Build GRCh38
rs749756355 Chr.11: 64232460-64232460 on Build GRCh38
rs977974004 Chr.11: 64232698-64232698 on Build GRCh38
rs776359033 Chr.11: 64232770-64232770 on Build GRCh38
rs756469089 Chr.11: 64232503-64232503 on Build GRCh38
rs372029552 Chr.11: 64232749-64232749 on Build GRCh38
rs369456737 Chr.11: 64232448-64232448 on Build GRCh38
rs200699204 Chr.11: 64232510-64232510 on Build GRCh38
rs770747108 Chr.11: 64232762-64232762 on Build GRCh38
rs199745306 Chr.11: 64232457-64232457 on Build GRCh38
rs775103818 Chr.11: 64232475-64232475 on Build GRCh38
rs760217798 Chr.11: 64232385-64232385 on Build GRCh38
rs761896083 Chr.11: 64232535-64232535 on Build GRCh38
rs757140841 Chr.11: 64232443-64232443 on Build GRCh38
rs758973410 Chr.11: 64232581-64232581 on Build GRCh38
rs879710727 Chr.11: 64232373-64232373 on Build GRCh38
rs752093263 Chr.11: 64232807-64232807 on Build GRCh38
rs769533900 Chr.11: 64232524-64232524 on Build GRCh38
rs375673802 Chr.11: 64232731-64232731 on Build GRCh38
rs765246616 Chr.11: 64232575-64232575 on Build GRCh38
rs759008176 Chr.11: 64232408-64232408 on Build GRCh38
rs775719182 Chr.11: 64232782-64232782 on Build GRCh38
rs781520025 Chr.11: 64232447-64232447 on Build GRCh38
rs778170358 Chr.11: 64232517-64232517 on Build GRCh38
rs372252507 Chr.11: 64232722-64232722 on Build GRCh38
rs200183856 Chr.11: 64232526-64232526 on Build GRCh38
rs750657148 Chr.11: 64232500-64232500 on Build GRCh38
rs752521946 Chr.11: 64232424-64232424 on Build GRCh38
rs201697121 Chr.11: 64232527-64232527 on Build GRCh38
rs776288235 Chr.11: 64232400-64232400 on Build GRCh38
rs751418913 Chr.11: 64232442-64232442 on Build GRCh38
rs528878768 Chr.11: 64232490-64232490 on Build GRCh38
rs201362496 Chr.11: 64232529-64232529 on Build GRCh38
rs370434882 Chr.11: 64232515-64232515 on Build GRCh38
rs1008866641 Chr.11: 64232598-64232598 on Build GRCh38
rs193059245 Chr.11: 64232499-64232499 on Build GRCh38
rs762953184 Chr.11: 64232796-64232796 on Build GRCh38
rs967876596 Chr.11: 64232680-64232680 on Build GRCh38
rs143213764 Chr.11: 64232461-64232462 on Build GRCh38
rs760482568 Chr.11: 64232678-64232678 on Build GRCh38
rs562381353 Chr.11: 64232512-64232512 on Build GRCh38
rs760961158 Chr.11: 64232548-64232548 on Build GRCh38
rs767684425 Chr.11: 64232414-64232414 on Build GRCh38
rs769952533 Chr.11: 64232779-64232779 on Build GRCh38
rs763041151 Chr.11: 64232729-64232729 on Build GRCh38
rs995070830 Chr.11: 64232570-64232570 on Build GRCh38
rs772458068 Chr.11: 64232468-64232468 on Build GRCh38
rs778629748 Chr.11: 64232635-64232635 on Build GRCh38
rs764172045 Chr.11: 64232802-64232802 on Build GRCh38
rs919262581 Chr.11: 64232423-64232423 on Build GRCh38
rs369570434 Chr.11: 64232390-64232390 on Build GRCh38
rs772188350 Chr.11: 64232656-64232656 on Build GRCh38
rs529657121 Chr.11: 64232606-64232606 on Build GRCh38
rs768979824 Chr.11: 64232582-64232582 on Build GRCh38
rs755040167 Chr.11: 64232511-64232511 on Build GRCh38
rs1032103487 Chr.11: 64232719-64232723 on Build GRCh38
rs766471862 Chr.11: 64232368-64232368 on Build GRCh38

View Full Product Details