Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78214145-78214684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387453 c.972G>A p.P324P Chr.17: 78214420-78214420 on Build GRCh38
7357525 c.25G>T p.A9S Chr.17: 78214341-78214341 on Build GRCh38
1716121 c.58T>A p.S20T Chr.17: 78214374-78214374 on Build GRCh38
5101109 c.1000G>A p.A334T Chr.17: 78214392-78214392 on Build GRCh38
1386406 c.76C>T p.P26S Chr.17: 78214392-78214392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370361101 Chr.17: 78214400-78214400 on Build GRCh38
rs921253761 Chr.17: 78214573-78214573 on Build GRCh38
rs912702673 Chr.17: 78214621-78214621 on Build GRCh38
rs960768768 Chr.17: 78214253-78214253 on Build GRCh38
rs765374415 Chr.17: 78214209-78214209 on Build GRCh38
rs1035190115 Chr.17: 78214488-78214488 on Build GRCh38
rs770168115 Chr.17: 78214252-78214252 on Build GRCh38
rs749745235 Chr.17: 78214310-78214310 on Build GRCh38
rs556970411 Chr.17: 78214449-78214450 on Build GRCh38
rs757193907 Chr.17: 78214292-78214292 on Build GRCh38
rs536546621 Chr.17: 78214489-78214489 on Build GRCh38
rs754589913 Chr.17: 78214417-78214417 on Build GRCh38
rs543367853 Chr.17: 78214339-78214339 on Build GRCh38
rs767364276 Chr.17: 78214205-78214205 on Build GRCh38
rs753843703 Chr.17: 78214291-78214291 on Build GRCh38
rs1022682908 Chr.17: 78214236-78214236 on Build GRCh38
rs750450342 Chr.17: 78214304-78214305 on Build GRCh38
rs765133832 Chr.17: 78214635-78214635 on Build GRCh38
rs202101234 Chr.17: 78214293-78214293 on Build GRCh38
rs988814205 Chr.17: 78214490-78214490 on Build GRCh38
rs762363555 Chr.17: 78214647-78214647 on Build GRCh38
rs572142200 Chr.17: 78214591-78214591 on Build GRCh38
rs974461485 Chr.17: 78214511-78214511 on Build GRCh38
rs1037373394 Chr.17: 78214651-78214651 on Build GRCh38
rs748729106 Chr.17: 78214211-78214211 on Build GRCh38
rs771722954 Chr.17: 78214237-78214237 on Build GRCh38
rs538731180 Chr.17: 78214534-78214534 on Build GRCh38
rs979488642 Chr.17: 78214558-78214558 on Build GRCh38
rs571941548 Chr.17: 78214212-78214212 on Build GRCh38
rs969829480 Chr.17: 78214453-78214453 on Build GRCh38
rs755988912 Chr.17: 78214289-78214289 on Build GRCh38
rs776366364 Chr.17: 78214628-78214628 on Build GRCh38
rs567991073 Chr.17: 78214475-78214475 on Build GRCh38
rs914136347 Chr.17: 78214524-78214524 on Build GRCh38
rs17882312 Chr.17: 78214176-78214176 on Build GRCh38
rs372753451 Chr.17: 78214375-78214375 on Build GRCh38
rs899470863 Chr.17: 78214439-78214439 on Build GRCh38
rs928110233 Chr.17: 78214263-78214263 on Build GRCh38
rs9904341 Chr.17: 78214286-78214286 on Build GRCh38
rs939624266 Chr.17: 78214278-78214278 on Build GRCh38
rs373880047 Chr.17: 78214307-78214307 on Build GRCh38
rs770250176 Chr.17: 78214306-78214306 on Build GRCh38
rs896576002 Chr.17: 78214479-78214479 on Build GRCh38
rs143160639 Chr.17: 78214341-78214341 on Build GRCh38
rs766631590 Chr.17: 78214269-78214269 on Build GRCh38
rs1043785405 Chr.17: 78214202-78214202 on Build GRCh38
rs751886129 Chr.17: 78214199-78214199 on Build GRCh38
rs145526644 Chr.17: 78214406-78214406 on Build GRCh38
rs774532555 Chr.17: 78214320-78214320 on Build GRCh38
rs962365441 Chr.17: 78214225-78214225 on Build GRCh38
rs1043005222 Chr.17: 78214442-78214442 on Build GRCh38
rs762028911 Chr.17: 78214208-78214209 on Build GRCh38
rs370205360 Chr.17: 78214182-78214182 on Build GRCh38
rs752405432 Chr.17: 78214287-78214287 on Build GRCh38
rs777808233 Chr.17: 78214210-78214210 on Build GRCh38
rs751292269 Chr.17: 78214414-78214414 on Build GRCh38
rs763927641 Chr.17: 78214290-78214290 on Build GRCh38
rs1015713511 Chr.17: 78214220-78214220 on Build GRCh38
rs773816169 Chr.17: 78214260-78214260 on Build GRCh38
rs759731481 Chr.17: 78214330-78214330 on Build GRCh38
rs527915990 Chr.17: 78214440-78214440 on Build GRCh38
rs779792860 Chr.17: 78214296-78214296 on Build GRCh38
rs199661984 Chr.17: 78214460-78214460 on Build GRCh38
rs771587762 Chr.17: 78214314-78214314 on Build GRCh38
rs998676813 Chr.17: 78214451-78214451 on Build GRCh38
rs763111647 Chr.17: 78214264-78214264 on Build GRCh38
rs982941178 Chr.17: 78214352-78214352 on Build GRCh38
rs1051088655 Chr.17: 78214600-78214600 on Build GRCh38
rs532219684 Chr.17: 78214358-78214358 on Build GRCh38
rs1004123291 Chr.17: 78214215-78214215 on Build GRCh38
rs778535940 Chr.17: 78214214-78214214 on Build GRCh38
rs765188612 Chr.17: 78214393-78214393 on Build GRCh38
rs771536267 Chr.17: 78214430-78214430 on Build GRCh38
rs745932468 Chr.17: 78214459-78214459 on Build GRCh38
rs779557679 Chr.17: 78214410-78214410 on Build GRCh38
rs112948461 Chr.17: 78214567-78214567 on Build GRCh38
rs762112729 Chr.17: 78214285-78214285 on Build GRCh38
rs1022618408 Chr.17: 78214445-78214445 on Build GRCh38
rs776808244 Chr.17: 78214249-78214249 on Build GRCh38
rs569729981 Chr.17: 78214505-78214505 on Build GRCh38
rs1024069279 Chr.17: 78214495-78214495 on Build GRCh38
rs267605071 Chr.17: 78214392-78214392 on Build GRCh38
rs761634652 Chr.17: 78214630-78214630 on Build GRCh38
rs768778329 Chr.17: 78214476-78214476 on Build GRCh38
rs144198399 Chr.17: 78214574-78214574 on Build GRCh38
rs534855597 Chr.17: 78214625-78214625 on Build GRCh38
rs760898261 Chr.17: 78214474-78214474 on Build GRCh38
rs995816357 Chr.17: 78214662-78214662 on Build GRCh38
rs892464657 Chr.17: 78214189-78214189 on Build GRCh38
rs748574527 Chr.17: 78214250-78214250 on Build GRCh38
rs148424824 Chr.17: 78214421-78214421 on Build GRCh38
rs745454097 Chr.17: 78214229-78214229 on Build GRCh38
rs748438531 Chr.17: 78214297-78214297 on Build GRCh38
rs775441897 Chr.17: 78214470-78214470 on Build GRCh38
rs753296580 Chr.17: 78214641-78214641 on Build GRCh38
rs368593302 Chr.17: 78214648-78214648 on Build GRCh38
rs755966559 Chr.17: 78214206-78214206 on Build GRCh38
rs549736720 Chr.17: 78214496-78214496 on Build GRCh38
rs765848428 Chr.17: 78214656-78214656 on Build GRCh38
rs968679445 Chr.17: 78214172-78214172 on Build GRCh38
rs558295554 Chr.17: 78214203-78214203 on Build GRCh38
rs920106823 Chr.17: 78214338-78214338 on Build GRCh38
rs554483754 Chr.17: 78214501-78214501 on Build GRCh38

View Full Product Details