Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 42262811-42263377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML4
Entrez Gene ID
27436
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
UniGene
Hs.713173
Gene Chromosome Location
Chr.2: 42169338-42332548 on Build GRCh38
Transcripts
NM_001145076.2 NM_019063.4 XM_005264267.1 XM_005264268.3 XM_006711991.1 XM_006711992.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426460 c.622G>T p.V208F Chr.2: 42263287-42263287 on Build GRCh38
1327081 c.545A>G p.H182R Chr.2: 42263210-42263210 on Build GRCh38
3771091 c.592A>G p.I198V Chr.2: 42263257-42263257 on Build GRCh38
1020948 c.619A>C p.K207Q Chr.2: 42263284-42263284 on Build GRCh38
1660114 c.537A>G p.E179E Chr.2: 42263202-42263202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886251298 Chr.2: 42263061-42263061 on Build GRCh38
rs868664334 Chr.2: 42263177-42263177 on Build GRCh38
rs375356882 Chr.2: 42262840-42262840 on Build GRCh38
rs376838322 Chr.2: 42263239-42263239 on Build GRCh38
rs766113787 Chr.2: 42263152-42263152 on Build GRCh38
rs1055046399 Chr.2: 42263022-42263022 on Build GRCh38
rs777087884 Chr.2: 42263252-42263252 on Build GRCh38
rs928060027 Chr.2: 42262994-42262994 on Build GRCh38
rs761095538 Chr.2: 42263321-42263323 on Build GRCh38
rs988989496 Chr.2: 42263097-42263097 on Build GRCh38
rs17029465 Chr.2: 42263079-42263079 on Build GRCh38
rs1056374138 Chr.2: 42262990-42262992 on Build GRCh38
rs767741679 Chr.2: 42263147-42263147 on Build GRCh38
rs950638621 Chr.2: 42262861-42262861 on Build GRCh38
rs75801954 Chr.2: 42263029-42263029 on Build GRCh38
rs1012205299 Chr.2: 42263268-42263268 on Build GRCh38
rs964348363 Chr.2: 42262890-42262890 on Build GRCh38
rs373665253 Chr.2: 42263139-42263139 on Build GRCh38
rs370198065 Chr.2: 42263344-42263344 on Build GRCh38
rs190116750 Chr.2: 42262884-42262884 on Build GRCh38
rs369456066 Chr.2: 42263134-42263134 on Build GRCh38
rs181458054 Chr.2: 42263108-42263108 on Build GRCh38
rs572102744 Chr.2: 42262950-42262950 on Build GRCh38
rs80152963 Chr.2: 42262956-42262956 on Build GRCh38
rs965736126 Chr.2: 42263118-42263118 on Build GRCh38
rs779948023 Chr.2: 42263227-42263227 on Build GRCh38
rs771258840 Chr.2: 42263249-42263249 on Build GRCh38
rs765784924 Chr.2: 42263258-42263258 on Build GRCh38
rs190935330 Chr.2: 42263208-42263208 on Build GRCh38
rs201200456 Chr.2: 42263165-42263165 on Build GRCh38
rs73933519 Chr.2: 42263316-42263316 on Build GRCh38
rs922087485 Chr.2: 42262920-42262920 on Build GRCh38
rs750394165 Chr.2: 42262977-42262977 on Build GRCh38
rs181738370 Chr.2: 42263332-42263332 on Build GRCh38
rs745578815 Chr.2: 42263221-42263221 on Build GRCh38
rs542620564 Chr.2: 42263140-42263140 on Build GRCh38
rs755869215 Chr.2: 42263292-42263292 on Build GRCh38
rs773409961 Chr.2: 42263244-42263244 on Build GRCh38
rs974954695 Chr.2: 42263123-42263123 on Build GRCh38
rs763474548 Chr.2: 42263186-42263186 on Build GRCh38
rs749147061 Chr.2: 42263235-42263235 on Build GRCh38
rs369089327 Chr.2: 42262893-42262893 on Build GRCh38
rs930565395 Chr.2: 42263045-42263045 on Build GRCh38
rs772121409 Chr.2: 42263336-42263336 on Build GRCh38
rs755162574 Chr.2: 42263283-42263283 on Build GRCh38
rs750078431 Chr.2: 42263285-42263285 on Build GRCh38
rs185738356 Chr.2: 42263128-42263128 on Build GRCh38
rs753681722 Chr.2: 42263313-42263313 on Build GRCh38
rs751135716 Chr.2: 42263260-42263260 on Build GRCh38
rs767194253 Chr.2: 42263277-42263277 on Build GRCh38
rs1048132233 Chr.2: 42263059-42263059 on Build GRCh38
rs988509307 Chr.2: 42262856-42262856 on Build GRCh38
rs896531772 Chr.2: 42262993-42262993 on Build GRCh38
rs955802583 Chr.2: 42263092-42263092 on Build GRCh38
rs766791365 Chr.2: 42263175-42263175 on Build GRCh38
rs930833596 Chr.2: 42262934-42262935 on Build GRCh38
rs1039341883 Chr.2: 42262909-42262909 on Build GRCh38
rs1008634323 Chr.2: 42262880-42262880 on Build GRCh38
rs34398507 Chr.2: 42262923-42262924 on Build GRCh38
rs148374489 Chr.2: 42263012-42263012 on Build GRCh38
rs369448526 Chr.2: 42263240-42263240 on Build GRCh38
rs900257401 Chr.2: 42263122-42263122 on Build GRCh38
rs561087552 Chr.2: 42263144-42263144 on Build GRCh38
rs146405197 Chr.2: 42263253-42263253 on Build GRCh38
rs200459540 Chr.2: 42263238-42263238 on Build GRCh38
rs908060698 Chr.2: 42263234-42263234 on Build GRCh38
rs990167201 Chr.2: 42262953-42262953 on Build GRCh38
rs768669032 Chr.2: 42263237-42263237 on Build GRCh38
rs200273713 Chr.2: 42263272-42263272 on Build GRCh38
rs937974682 Chr.2: 42263011-42263011 on Build GRCh38
rs757950489 Chr.2: 42263207-42263207 on Build GRCh38
rs768594279 Chr.2: 42263142-42263142 on Build GRCh38
rs764531580 Chr.2: 42263183-42263183 on Build GRCh38
rs1026140820 Chr.2: 42262855-42262855 on Build GRCh38
rs775677349 Chr.2: 42263067-42263067 on Build GRCh38
rs945566044 Chr.2: 42262959-42262959 on Build GRCh38
rs557018967 Chr.2: 42262943-42262943 on Build GRCh38
rs780992466 Chr.2: 42263216-42263216 on Build GRCh38
rs754282899 Chr.2: 42263179-42263179 on Build GRCh38
rs571318886 Chr.2: 42263327-42263327 on Build GRCh38
rs537031178 Chr.2: 42262935-42262935 on Build GRCh38
rs141433646 Chr.2: 42263190-42263190 on Build GRCh38
rs1022846948 Chr.2: 42263027-42263027 on Build GRCh38
rs761399901 Chr.2: 42263274-42263274 on Build GRCh38
rs755551451 Chr.2: 42263185-42263185 on Build GRCh38
rs755959177 Chr.2: 42263224-42263224 on Build GRCh38

View Full Product Details