Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25446370-25446826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
UniGene
Hs.658464
Gene Chromosome Location
Chr.20: 25407470-25448568 on Build GRCh38
Transcripts
NM_021067.4 NR_134574.1 XM_017028162.1

SNP Detail

SNP ID Chromosome Location
rs984542869 Chr.20: 25446511-25446511 on Build GRCh38
rs548520143 Chr.20: 25446707-25446707 on Build GRCh38
rs972305901 Chr.20: 25446616-25446616 on Build GRCh38
rs1026632802 Chr.20: 25446593-25446593 on Build GRCh38
rs1018194919 Chr.20: 25446538-25446538 on Build GRCh38
rs182163699 Chr.20: 25446568-25446568 on Build GRCh38
rs765476387 Chr.20: 25446669-25446669 on Build GRCh38
rs892508337 Chr.20: 25446407-25446407 on Build GRCh38
rs568094212 Chr.20: 25446724-25446724 on Build GRCh38
rs371295950 Chr.20: 25446530-25446530 on Build GRCh38
rs937307357 Chr.20: 25446714-25446714 on Build GRCh38
rs890719795 Chr.20: 25446459-25446459 on Build GRCh38
rs963781164 Chr.20: 25446557-25446557 on Build GRCh38
rs868586646 Chr.20: 25446791-25446791 on Build GRCh38
rs953003396 Chr.20: 25446476-25446476 on Build GRCh38
rs878889239 Chr.20: 25446712-25446712 on Build GRCh38
rs879108676 Chr.20: 25446708-25446708 on Build GRCh38
rs541306415 Chr.20: 25446639-25446639 on Build GRCh38
rs531890448 Chr.20: 25446655-25446655 on Build GRCh38
rs1044964441 Chr.20: 25446704-25446704 on Build GRCh38
rs1009758364 Chr.20: 25446435-25446436 on Build GRCh38
rs764939841 Chr.20: 25446754-25446754 on Build GRCh38
rs2387887 Chr.20: 25446445-25446445 on Build GRCh38
rs1030917736 Chr.20: 25446696-25446696 on Build GRCh38

View Full Product Details