Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117361695-117362194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP164
Entrez Gene ID
22897
Gene Name
centrosomal protein 164
Gene Aliases
NPHP15
UniGene
Hs.504009
Gene Chromosome Location
Chr.11: 117316346-117413266 on Build GRCh38
Transcripts
NM_001271933.1 NM_014956.4 XM_005271453.1 XM_005271456.1 XM_005271457.1 XM_006718788.1 XM_006718794.1 XM_011542674.1 XM_011542690.1 XM_017017364.1 XM_017017365.1 XM_017017366.1 XM_017017367.1 XM_017017368.1 XM_017017369.1 XM_017017370.1 XM_017017371.1 XM_017017372.1 XM_017017373.1 XM_017017374.1 XM_017017375.1 XM_017017376.1 XM_017017377.1 XM_017017378.1 XM_017017379.1 XM_017017380.1 XM_017017381.1 XM_017017382.1 XM_017017383.1 XM_017017384.1 XM_017017385.1 XM_017017386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4018154 c.437G>A p.G146D Chr.11: 117361878-117361878 on Build GRCh38
304698 c.467C>A p.T156N Chr.11: 117361908-117361908 on Build GRCh38
6626883 c.529C>G p.R177G Chr.11: 117361970-117361970 on Build GRCh38
7079811 c.498C>T p.S166S Chr.11: 117361939-117361939 on Build GRCh38
39944 c.427C>T p.P143S Chr.11: 117361868-117361868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778187174 Chr.11: 117362032-117362032 on Build GRCh38
rs754626177 Chr.11: 117362007-117362007 on Build GRCh38
rs200336962 Chr.11: 117361840-117361840 on Build GRCh38
rs376357203 Chr.11: 117362039-117362039 on Build GRCh38
rs938469001 Chr.11: 117362063-117362063 on Build GRCh38
rs200045306 Chr.11: 117361863-117361863 on Build GRCh38
rs764197141 Chr.11: 117361850-117361850 on Build GRCh38
rs774763557 Chr.11: 117361970-117361970 on Build GRCh38
rs756111498 Chr.11: 117361849-117361849 on Build GRCh38
rs114317885 Chr.11: 117362087-117362087 on Build GRCh38
rs533975246 Chr.11: 117362137-117362137 on Build GRCh38
rs771250417 Chr.11: 117361969-117361969 on Build GRCh38
rs564746056 Chr.11: 117361893-117361893 on Build GRCh38
rs781697417 Chr.11: 117361891-117361891 on Build GRCh38
rs749377313 Chr.11: 117362034-117362034 on Build GRCh38
rs375118998 Chr.11: 117361898-117361898 on Build GRCh38
rs183408857 Chr.11: 117362141-117362141 on Build GRCh38
rs61746874 Chr.11: 117361836-117361836 on Build GRCh38
rs755283139 Chr.11: 117361933-117361933 on Build GRCh38
rs367677064 Chr.11: 117361819-117361819 on Build GRCh38
rs139763142 Chr.11: 117361926-117361926 on Build GRCh38
rs771340026 Chr.11: 117361909-117361909 on Build GRCh38
rs753257758 Chr.11: 117361856-117361856 on Build GRCh38
rs144206271 Chr.11: 117361989-117361989 on Build GRCh38
rs779722076 Chr.11: 117361931-117361931 on Build GRCh38
rs1004213152 Chr.11: 117362112-117362112 on Build GRCh38
rs903602917 Chr.11: 117362154-117362154 on Build GRCh38
rs201696624 Chr.11: 117362023-117362023 on Build GRCh38
rs538170911 Chr.11: 117362043-117362043 on Build GRCh38
rs764102727 Chr.11: 117361917-117361917 on Build GRCh38
rs952724198 Chr.11: 117362068-117362068 on Build GRCh38
rs777623758 Chr.11: 117361813-117361814 on Build GRCh38
rs998296274 Chr.11: 117362146-117362146 on Build GRCh38
rs908786819 Chr.11: 117361853-117361853 on Build GRCh38
rs770213078 Chr.11: 117361942-117361942 on Build GRCh38
rs769208186 Chr.11: 117361784-117361784 on Build GRCh38
rs947103111 Chr.11: 117361717-117361717 on Build GRCh38
rs761992889 Chr.11: 117361793-117361793 on Build GRCh38
rs749917447 Chr.11: 117361988-117361988 on Build GRCh38
rs753063818 Chr.11: 117362009-117362009 on Build GRCh38
rs773059080 Chr.11: 117362010-117362010 on Build GRCh38
rs977536274 Chr.11: 117362051-117362051 on Build GRCh38
rs756840685 Chr.11: 117361857-117361857 on Build GRCh38
rs888403362 Chr.11: 117362095-117362095 on Build GRCh38
rs201463212 Chr.11: 117361817-117361817 on Build GRCh38
rs561298323 Chr.11: 117361739-117361739 on Build GRCh38
rs761756138 Chr.11: 117361922-117361922 on Build GRCh38
rs547394550 Chr.11: 117361915-117361915 on Build GRCh38
rs769120227 Chr.11: 117361876-117361876 on Build GRCh38
rs143378905 Chr.11: 117362089-117362089 on Build GRCh38
rs768683135 Chr.11: 117361795-117361795 on Build GRCh38
rs199701600 Chr.11: 117361971-117361971 on Build GRCh38
rs779165404 Chr.11: 117362040-117362040 on Build GRCh38
rs749360330 Chr.11: 117362035-117362035 on Build GRCh38
rs763180176 Chr.11: 117361908-117361908 on Build GRCh38
rs775425959 Chr.11: 117361914-117361914 on Build GRCh38
rs778310001 Chr.11: 117361859-117361859 on Build GRCh38
rs149195472 Chr.11: 117361892-117361892 on Build GRCh38
rs748619597 Chr.11: 117361940-117361940 on Build GRCh38
rs2305825 Chr.11: 117361776-117361776 on Build GRCh38
rs764739972 Chr.11: 117361925-117361925 on Build GRCh38
rs931735196 Chr.11: 117362094-117362094 on Build GRCh38
rs145771994 Chr.11: 117361951-117361951 on Build GRCh38
rs918032175 Chr.11: 117361767-117361767 on Build GRCh38
rs772199591 Chr.11: 117362044-117362044 on Build GRCh38
rs761794892 Chr.11: 117361980-117361980 on Build GRCh38
rs982800833 Chr.11: 117362072-117362072 on Build GRCh38
rs965491187 Chr.11: 117361946-117361946 on Build GRCh38
rs546289402 Chr.11: 117362056-117362056 on Build GRCh38
rs761689140 Chr.11: 117361924-117361924 on Build GRCh38
rs781437579 Chr.11: 117361939-117361939 on Build GRCh38
rs908812602 Chr.11: 117362084-117362084 on Build GRCh38
rs780381078 Chr.11: 117361865-117361865 on Build GRCh38
rs573791479 Chr.11: 117361768-117361768 on Build GRCh38
rs568657159 Chr.11: 117362025-117362025 on Build GRCh38
rs145551039 Chr.11: 117361837-117361837 on Build GRCh38
rs564463892 Chr.11: 117362111-117362111 on Build GRCh38
rs773431572 Chr.11: 117361805-117361805 on Build GRCh38
rs1048646571 Chr.11: 117362093-117362093 on Build GRCh38
rs148498531 Chr.11: 117361975-117361975 on Build GRCh38
rs765152748 Chr.11: 117361985-117361985 on Build GRCh38
rs777899286 Chr.11: 117361948-117361948 on Build GRCh38
rs368573245 Chr.11: 117362001-117362001 on Build GRCh38
rs188292128 Chr.11: 117362162-117362162 on Build GRCh38
rs149806421 Chr.11: 117361932-117361932 on Build GRCh38
rs753434921 Chr.11: 117362092-117362092 on Build GRCh38
rs868692268 Chr.11: 117361862-117361862 on Build GRCh38
rs991996087 Chr.11: 117361718-117361718 on Build GRCh38
rs756643754 Chr.11: 117362012-117362012 on Build GRCh38
rs371658661 Chr.11: 117361860-117361860 on Build GRCh38

View Full Product Details