Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117363347-117363761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP164
Entrez Gene ID
22897
Gene Name
centrosomal protein 164
Gene Aliases
NPHP15
UniGene
Hs.504009
Gene Chromosome Location
Chr.11: 117316346-117413266 on Build GRCh38
Transcripts
NM_001271933.1 NM_014956.4 XM_005271453.1 XM_005271456.1 XM_005271457.1 XM_006718788.1 XM_006718794.1 XM_011542674.1 XM_011542690.1 XM_017017364.1 XM_017017365.1 XM_017017366.1 XM_017017367.1 XM_017017368.1 XM_017017369.1 XM_017017370.1 XM_017017371.1 XM_017017372.1 XM_017017373.1 XM_017017374.1 XM_017017375.1 XM_017017376.1 XM_017017377.1 XM_017017378.1 XM_017017379.1 XM_017017380.1 XM_017017381.1 XM_017017382.1 XM_017017383.1 XM_017017384.1 XM_017017385.1 XM_017017386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7337476 c.748G>A p.G250S Chr.11: 117363489-117363489 on Build GRCh38
1351521 c.735_736insG p.A247fs*5 Chr.11: 117363476-117363477 on Build GRCh38
6576668 c.693C>T p.V231V Chr.11: 117363434-117363434 on Build GRCh38
6131284 c.757G>C p.E253Q Chr.11: 117363498-117363498 on Build GRCh38
4671606 c.744delG p.G250fs*9 Chr.11: 117363485-117363485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201189268 Chr.11: 117363437-117363437 on Build GRCh38
rs896896553 Chr.11: 117363672-117363672 on Build GRCh38
rs993943625 Chr.11: 117363705-117363705 on Build GRCh38
rs181192172 Chr.11: 117363668-117363668 on Build GRCh38
rs949970866 Chr.11: 117363572-117363572 on Build GRCh38
rs901284620 Chr.11: 117363390-117363390 on Build GRCh38
rs766308731 Chr.11: 117363503-117363503 on Build GRCh38
rs539939395 Chr.11: 117363422-117363422 on Build GRCh38
rs141163158 Chr.11: 117363502-117363502 on Build GRCh38
rs768039587 Chr.11: 117363695-117363695 on Build GRCh38
rs754269420 Chr.11: 117363391-117363391 on Build GRCh38
rs552422073 Chr.11: 117363724-117363724 on Build GRCh38
rs905540523 Chr.11: 117363368-117363368 on Build GRCh38
rs774961179 Chr.11: 117363379-117363379 on Build GRCh38
rs1038077089 Chr.11: 117363659-117363659 on Build GRCh38
rs755237805 Chr.11: 117363439-117363439 on Build GRCh38
rs751715778 Chr.11: 117363523-117363523 on Build GRCh38
rs370034077 Chr.11: 117363427-117363427 on Build GRCh38
rs754919251 Chr.11: 117363687-117363687 on Build GRCh38
rs146501105 Chr.11: 117363489-117363489 on Build GRCh38
rs549905287 Chr.11: 117363467-117363467 on Build GRCh38
rs750854769 Chr.11: 117363501-117363501 on Build GRCh38
rs934087216 Chr.11: 117363698-117363698 on Build GRCh38
rs994330234 Chr.11: 117363411-117363411 on Build GRCh38
rs376384904 Chr.11: 117363534-117363534 on Build GRCh38
rs768482761 Chr.11: 117363476-117363476 on Build GRCh38
rs772446890 Chr.11: 117363541-117363541 on Build GRCh38
rs1037252516 Chr.11: 117363380-117363380 on Build GRCh38
rs760189135 Chr.11: 117363475-117363475 on Build GRCh38
rs781435180 Chr.11: 117363535-117363535 on Build GRCh38
rs749146317 Chr.11: 117363447-117363447 on Build GRCh38
rs753611495 Chr.11: 117363542-117363542 on Build GRCh38
rs373403803 Chr.11: 117363521-117363521 on Build GRCh38
rs770827539 Chr.11: 117363449-117363449 on Build GRCh38
rs530136327 Chr.11: 117363456-117363456 on Build GRCh38
rs376432302 Chr.11: 117363405-117363405 on Build GRCh38
rs780214572 Chr.11: 117363431-117363431 on Build GRCh38
rs201838344 Chr.11: 117363383-117363383 on Build GRCh38
rs764300163 Chr.11: 117363386-117363386 on Build GRCh38
rs889867894 Chr.11: 117363735-117363735 on Build GRCh38
rs760406989 Chr.11: 117363385-117363385 on Build GRCh38
rs567606919 Chr.11: 117363451-117363451 on Build GRCh38
rs368668415 Chr.11: 117363490-117363490 on Build GRCh38
rs935444789 Chr.11: 117363500-117363500 on Build GRCh38
rs972603688 Chr.11: 117363679-117363679 on Build GRCh38
rs777103598 Chr.11: 117363481-117363481 on Build GRCh38
rs745843938 Chr.11: 117363455-117363455 on Build GRCh38
rs373565225 Chr.11: 117363486-117363486 on Build GRCh38
rs189584403 Chr.11: 117363646-117363646 on Build GRCh38
rs892024583 Chr.11: 117363453-117363453 on Build GRCh38
rs1044715213 Chr.11: 117363619-117363619 on Build GRCh38
rs781011398 Chr.11: 117363716-117363716 on Build GRCh38
rs970485689 Chr.11: 117363525-117363525 on Build GRCh38
rs762277597 Chr.11: 117363576-117363576 on Build GRCh38
rs1029865836 Chr.11: 117363425-117363425 on Build GRCh38
rs374465259 Chr.11: 117363429-117363429 on Build GRCh38
rs1005269066 Chr.11: 117363369-117363369 on Build GRCh38
rs777527578 Chr.11: 117363444-117363444 on Build GRCh38
rs752244904 Chr.11: 117363548-117363548 on Build GRCh38
rs939916960 Chr.11: 117363650-117363650 on Build GRCh38

View Full Product Details