Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 117371349-117371795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP164
Entrez Gene ID
22897
Gene Name
centrosomal protein 164
Gene Aliases
NPHP15
UniGene
Hs.504009
Gene Chromosome Location
Chr.11: 117316346-117413266 on Build GRCh38
Transcripts
NM_001271933.1 NM_014956.4 XM_005271453.1 XM_005271456.1 XM_005271457.1 XM_006718788.1 XM_006718794.1 XM_011542674.1 XM_017017364.1 XM_017017365.1 XM_017017366.1 XM_017017367.1 XM_017017368.1 XM_017017369.1 XM_017017370.1 XM_017017371.1 XM_017017372.1 XM_017017373.1 XM_017017374.1 XM_017017375.1 XM_017017376.1 XM_017017377.1 XM_017017378.1 XM_017017379.1 XM_017017380.1 XM_017017381.1 XM_017017382.1 XM_017017383.1 XM_017017384.1 XM_017017385.1 XM_017017386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5576528 c.1076C>T p.S359F Chr.11: 117371390-117371390 on Build GRCh38
4795626 c.1068G>A p.E356E Chr.11: 117371382-117371382 on Build GRCh38
5716634 c.1123C>T p.L375L Chr.11: 117371437-117371437 on Build GRCh38
686222 c.1043C>A p.P348Q Chr.11: 117371357-117371357 on Build GRCh38
6131283 c.1135A>G p.S379G Chr.11: 117371449-117371449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780364658 Chr.11: 117371458-117371458 on Build GRCh38
rs1036349313 Chr.11: 117371432-117371432 on Build GRCh38
rs1051046683 Chr.11: 117371581-117371581 on Build GRCh38
rs374832717 Chr.11: 117371450-117371450 on Build GRCh38
rs759540477 Chr.11: 117371505-117371505 on Build GRCh38
rs751458582 Chr.11: 117371495-117371495 on Build GRCh38
rs1025752062 Chr.11: 117371633-117371633 on Build GRCh38
rs891021697 Chr.11: 117371635-117371635 on Build GRCh38
rs973717966 Chr.11: 117371529-117371529 on Build GRCh38
rs371475400 Chr.11: 117371484-117371484 on Build GRCh38
rs771417207 Chr.11: 117371525-117371525 on Build GRCh38
rs942869444 Chr.11: 117371720-117371720 on Build GRCh38
rs183817288 Chr.11: 117371725-117371725 on Build GRCh38
rs190673622 Chr.11: 117371692-117371692 on Build GRCh38
rs1011635491 Chr.11: 117371685-117371685 on Build GRCh38
rs372037335 Chr.11: 117371479-117371479 on Build GRCh38
rs1003268770 Chr.11: 117371756-117371756 on Build GRCh38
rs962971618 Chr.11: 117371599-117371599 on Build GRCh38
rs550044351 Chr.11: 117371451-117371451 on Build GRCh38
rs748917124 Chr.11: 117371403-117371403 on Build GRCh38
rs776869429 Chr.11: 117371471-117371471 on Build GRCh38
rs368171178 Chr.11: 117371466-117371466 on Build GRCh38
rs759877601 Chr.11: 117371726-117371727 on Build GRCh38
rs749786253 Chr.11: 117371455-117371455 on Build GRCh38
rs760703377 Chr.11: 117371580-117371580 on Build GRCh38
rs140667494 Chr.11: 117371654-117371654 on Build GRCh38
rs537094175 Chr.11: 117371561-117371561 on Build GRCh38
rs981749795 Chr.11: 117371400-117371400 on Build GRCh38
rs973737870 Chr.11: 117371615-117371615 on Build GRCh38
rs1020375113 Chr.11: 117371710-117371710 on Build GRCh38
rs918287955 Chr.11: 117371555-117371555 on Build GRCh38
rs771288567 Chr.11: 117371433-117371433 on Build GRCh38
rs375703035 Chr.11: 117371482-117371482 on Build GRCh38
rs375647094 Chr.11: 117371457-117371457 on Build GRCh38
rs761942364 Chr.11: 117371480-117371480 on Build GRCh38
rs774584906 Chr.11: 117371443-117371443 on Build GRCh38
rs942560087 Chr.11: 117371641-117371641 on Build GRCh38
rs200122409 Chr.11: 117371449-117371449 on Build GRCh38
rs893230626 Chr.11: 117371668-117371668 on Build GRCh38
rs1034262977 Chr.11: 117371587-117371587 on Build GRCh38
rs903224242 Chr.11: 117371755-117371755 on Build GRCh38

View Full Product Details