Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118313615-118314116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
UniGene
Hs.3003
Gene Chromosome Location
Chr.11: 118304580-118316175 on Build GRCh38
Transcripts
NM_000733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2105209 c.490C>T p.R164* Chr.11: 118313844-118313844 on Build GRCh38
3868516 c.468G>A p.K156K Chr.11: 118313822-118313822 on Build GRCh38
4018266 c.443T>C p.V148A Chr.11: 118313797-118313797 on Build GRCh38
7092307 c.503C>T p.A168V Chr.11: 118313857-118313857 on Build GRCh38
77441 c.400G>C p.V134L Chr.11: 118313754-118313754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760176475 Chr.11: 118313703-118313703 on Build GRCh38
rs754912108 Chr.11: 118313761-118313761 on Build GRCh38
rs747019188 Chr.11: 118313685-118313685 on Build GRCh38
rs764211968 Chr.11: 118313729-118313729 on Build GRCh38
rs765464852 Chr.11: 118313879-118313879 on Build GRCh38
rs535137720 Chr.11: 118313933-118313933 on Build GRCh38
rs767365744 Chr.11: 118313884-118313884 on Build GRCh38
rs1019900729 Chr.11: 118313995-118313995 on Build GRCh38
rs201572053 Chr.11: 118313689-118313689 on Build GRCh38
rs374456909 Chr.11: 118313702-118313702 on Build GRCh38
rs951155693 Chr.11: 118313988-118313988 on Build GRCh38
rs199578380 Chr.11: 118313862-118313862 on Build GRCh38
rs944780357 Chr.11: 118313958-118313958 on Build GRCh38
rs143868906 Chr.11: 118313762-118313762 on Build GRCh38
rs528492194 Chr.11: 118313795-118313795 on Build GRCh38
rs140639753 Chr.11: 118313824-118313824 on Build GRCh38
rs746750290 Chr.11: 118313834-118313834 on Build GRCh38
rs563868055 Chr.11: 118313740-118313740 on Build GRCh38
rs376803194 Chr.11: 118313845-118313845 on Build GRCh38
rs745493004 Chr.11: 118313854-118313854 on Build GRCh38
rs772687199 Chr.11: 118313701-118313701 on Build GRCh38
rs775996414 Chr.11: 118313860-118313860 on Build GRCh38
rs747675748 Chr.11: 118313770-118313770 on Build GRCh38
rs747587896 Chr.11: 118313693-118313693 on Build GRCh38
rs749952590 Chr.11: 118313651-118313651 on Build GRCh38
rs142332884 Chr.11: 118313652-118313652 on Build GRCh38
rs200718457 Chr.11: 118313697-118313697 on Build GRCh38
rs200974160 Chr.11: 118313771-118313771 on Build GRCh38
rs2231447 Chr.11: 118313861-118313861 on Build GRCh38
rs180888533 Chr.11: 118314062-118314062 on Build GRCh38
rs530715111 Chr.11: 118313835-118313835 on Build GRCh38
rs772728734 Chr.11: 118313805-118313805 on Build GRCh38
rs765373016 Chr.11: 118313839-118313839 on Build GRCh38
rs200532850 Chr.11: 118313907-118313907 on Build GRCh38
rs763127370 Chr.11: 118313881-118313881 on Build GRCh38
rs146881885 Chr.11: 118313802-118313802 on Build GRCh38
rs748165299 Chr.11: 118314073-118314073 on Build GRCh38
rs2231446 Chr.11: 118313635-118313635 on Build GRCh38
rs368489854 Chr.11: 118313853-118313853 on Build GRCh38
rs1044339501 Chr.11: 118313681-118313681 on Build GRCh38
rs113606389 Chr.11: 118313980-118313980 on Build GRCh38
rs201004100 Chr.11: 118313672-118313672 on Build GRCh38
rs200744872 Chr.11: 118313866-118313866 on Build GRCh38
rs752522836 Chr.11: 118313891-118313891 on Build GRCh38
rs2231448 Chr.11: 118313920-118313920 on Build GRCh38
rs200193595 Chr.11: 118313741-118313741 on Build GRCh38
rs773756334 Chr.11: 118313696-118313697 on Build GRCh38
rs546397282 Chr.11: 118313910-118313910 on Build GRCh38
rs78373007 Chr.11: 118313852-118313852 on Build GRCh38
rs975763808 Chr.11: 118314029-118314029 on Build GRCh38
rs748959958 Chr.11: 118313793-118313793 on Build GRCh38
rs199933802 Chr.11: 118313696-118313696 on Build GRCh38
rs768570512 Chr.11: 118313687-118313687 on Build GRCh38
rs774861719 Chr.11: 118313800-118313800 on Build GRCh38
rs748743171 Chr.11: 118313660-118313660 on Build GRCh38
rs201496843 Chr.11: 118313896-118313896 on Build GRCh38
rs780420635 Chr.11: 118313728-118313728 on Build GRCh38
rs483352928 Chr.11: 118313876-118313876 on Build GRCh38
rs781262967 Chr.11: 118313994-118313994 on Build GRCh38
rs200258299 Chr.11: 118313812-118313812 on Build GRCh38
rs1023279888 Chr.11: 118313906-118313906 on Build GRCh38
rs73612248 Chr.11: 118313941-118313941 on Build GRCh38
rs369532194 Chr.11: 118313737-118313737 on Build GRCh38
rs568815647 Chr.11: 118314026-118314026 on Build GRCh38
rs955869745 Chr.11: 118313838-118313838 on Build GRCh38
rs201894935 Chr.11: 118313721-118313721 on Build GRCh38
rs202201983 Chr.11: 118313757-118313757 on Build GRCh38
rs552989886 Chr.11: 118313634-118313634 on Build GRCh38
rs772605914 Chr.11: 118313857-118313857 on Build GRCh38
rs995228474 Chr.11: 118313930-118313930 on Build GRCh38
rs372144734 Chr.11: 118313816-118313816 on Build GRCh38
rs530683848 Chr.11: 118313878-118313878 on Build GRCh38
rs537729890 Chr.11: 118314028-118314028 on Build GRCh38
rs768072020 Chr.11: 118313704-118313704 on Build GRCh38
rs779870558 Chr.11: 118313683-118313683 on Build GRCh38
rs60820882 Chr.11: 118314064-118314064 on Build GRCh38
rs201840561 Chr.11: 118313844-118313844 on Build GRCh38
rs764151020 Chr.11: 118313818-118313818 on Build GRCh38
rs199539988 Chr.11: 118313902-118313902 on Build GRCh38
rs757942732 Chr.11: 118313670-118313670 on Build GRCh38
rs777304757 Chr.11: 118313783-118313783 on Build GRCh38
rs200987721 Chr.11: 118313752-118313752 on Build GRCh38
rs368688587 Chr.11: 118313668-118313668 on Build GRCh38
rs770417767 Chr.11: 118313687-118313691 on Build GRCh38
rs751299903 Chr.11: 118313759-118313759 on Build GRCh38
rs200646622 Chr.11: 118313792-118313792 on Build GRCh38
rs971158613 Chr.11: 118314075-118314075 on Build GRCh38
rs780206260 Chr.11: 118313965-118313965 on Build GRCh38
rs144452564 Chr.11: 118313851-118313851 on Build GRCh38
rs201734738 Chr.11: 118313909-118313909 on Build GRCh38
rs946824844 Chr.11: 118313647-118313647 on Build GRCh38
rs752856900 Chr.11: 118313723-118313723 on Build GRCh38
rs201007974 Chr.11: 118313921-118313921 on Build GRCh38
rs758629254 Chr.11: 118313944-118313944 on Build GRCh38

View Full Product Details