Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119096414-119096910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPAGT1
Entrez Gene ID
1798
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Gene Aliases
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT
UniGene
Hs.524081
Gene Chromosome Location
Chr.11: 119096503-119102414 on Build GRCh38
Transcripts
NM_001382.3 XM_005271422.2 XM_011542648.1 XM_017017293.1 XM_017017294.1 XM_017017295.1

SNP Detail

SNP ID Chromosome Location
rs970406459 Chr.11: 119096820-119096820 on Build GRCh38
rs767052699 Chr.11: 119096460-119096460 on Build GRCh38
rs981745573 Chr.11: 119096829-119096829 on Build GRCh38
rs997793115 Chr.11: 119096514-119096514 on Build GRCh38
rs886047757 Chr.11: 119096514-119096514 on Build GRCh38
rs1044085763 Chr.11: 119096447-119096447 on Build GRCh38
rs942125614 Chr.11: 119096672-119096672 on Build GRCh38
rs559849249 Chr.11: 119096844-119096844 on Build GRCh38
rs8551 Chr.11: 119096814-119096814 on Build GRCh38
rs552234070 Chr.11: 119096785-119096785 on Build GRCh38
rs757149167 Chr.11: 119096749-119096749 on Build GRCh38
rs886047759 Chr.11: 119096629-119096629 on Build GRCh38
rs529508262 Chr.11: 119096636-119096636 on Build GRCh38
rs939858971 Chr.11: 119096883-119096883 on Build GRCh38
rs961755403 Chr.11: 119096722-119096722 on Build GRCh38
rs1032937642 Chr.11: 119096762-119096762 on Build GRCh38
rs577500688 Chr.11: 119096643-119096643 on Build GRCh38
rs748903893 Chr.11: 119096606-119096606 on Build GRCh38
rs866115351 Chr.11: 119096540-119096540 on Build GRCh38
rs374531996 Chr.11: 119096649-119096649 on Build GRCh38
rs953562445 Chr.11: 119096546-119096546 on Build GRCh38
rs951925000 Chr.11: 119096671-119096672 on Build GRCh38
rs982028493 Chr.11: 119096612-119096612 on Build GRCh38
rs752141819 Chr.11: 119096547-119096547 on Build GRCh38
rs970354016 Chr.11: 119096608-119096608 on Build GRCh38
rs549862935 Chr.11: 119096886-119096886 on Build GRCh38
rs958035813 Chr.11: 119096596-119096596 on Build GRCh38
rs28990976 Chr.11: 119096511-119096511 on Build GRCh38
rs926255174 Chr.11: 119096834-119096834 on Build GRCh38
rs28990975 Chr.11: 119096571-119096571 on Build GRCh38
rs923050764 Chr.11: 119096756-119096756 on Build GRCh38
rs878926920 Chr.11: 119096534-119096534 on Build GRCh38
rs997512016 Chr.11: 119096716-119096716 on Build GRCh38
rs886047758 Chr.11: 119096619-119096619 on Build GRCh38
rs74992671 Chr.11: 119096839-119096839 on Build GRCh38
rs7759 Chr.11: 119096581-119096581 on Build GRCh38
rs909729962 Chr.11: 119096882-119096882 on Build GRCh38
rs909685312 Chr.11: 119096671-119096671 on Build GRCh38
rs984029661 Chr.11: 119096863-119096863 on Build GRCh38
rs150240212 Chr.11: 119096631-119096631 on Build GRCh38
rs28990974 Chr.11: 119096733-119096733 on Build GRCh38
rs1013147344 Chr.11: 119096490-119096490 on Build GRCh38
rs557014624 Chr.11: 119096508-119096508 on Build GRCh38
rs926539216 Chr.11: 119096613-119096613 on Build GRCh38
rs34066457 Chr.11: 119096709-119096709 on Build GRCh38
rs541589857 Chr.11: 119096674-119096674 on Build GRCh38
rs1052058073 Chr.11: 119096719-119096719 on Build GRCh38
rs897858674 Chr.11: 119096710-119096710 on Build GRCh38
rs867673677 Chr.11: 119096828-119096828 on Build GRCh38
rs1036782503 Chr.11: 119096471-119096471 on Build GRCh38
rs958209954 Chr.11: 119096765-119096765 on Build GRCh38
rs74657539 Chr.11: 119096757-119096757 on Build GRCh38
rs536836557 Chr.11: 119096789-119096789 on Build GRCh38
rs937903557 Chr.11: 119096632-119096632 on Build GRCh38
rs999996483 Chr.11: 119096521-119096521 on Build GRCh38
rs987881369 Chr.11: 119096604-119096604 on Build GRCh38

View Full Product Details